Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 82358659-82359169 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NRG3
Entrez Gene ID
10718
Gene Name
neuregulin 3
Gene Aliases
HRG3, pro-NRG3
UniGene
Hs.125119
Gene Chromosome Location
Chr.10: 81875298-82989979 on Build GRCh38
Transcripts
NM_001010848.3 NM_001165972.1 NM_001165973.1 XM_005269444.4 XM_011539172.2 XM_011539173.2 XM_011539175.2 XM_017015573.1 XM_017015574.1 XM_017015575.1 XM_017015576.1 XM_017015577.1 XM_017015578.1 XM_017015579.1 XM_017015580.1 XM_017015581.1 XM_017015582.1 XM_017015583.1 XM_017015584.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4455724 c.875A>T p.D292V Chr.10: 82358790-82358790 on Build GRCh38
920842 c.190G>A p.E64K Chr.10: 82358768-82358768 on Build GRCh38
1638661 c.208C>T p.R70* Chr.10: 82358786-82358786 on Build GRCh38
1259498 c.873A>T p.R291R Chr.10: 82358788-82358788 on Build GRCh38
920846 c.258C>T p.I86I Chr.10: 82358836-82358836 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs984844666 Chr.10: 82358922-82358922 on Build GRCh38
rs911760005 Chr.10: 82358929-82358929 on Build GRCh38
rs768371489 Chr.10: 82358787-82358787 on Build GRCh38
rs755268774 Chr.10: 82358751-82358751 on Build GRCh38
rs997798748 Chr.10: 82358711-82358711 on Build GRCh38
rs771942584 Chr.10: 82358850-82358850 on Build GRCh38
rs148906289 Chr.10: 82358750-82358750 on Build GRCh38
rs112755047 Chr.10: 82358980-82358980 on Build GRCh38
rs779901106 Chr.10: 82358714-82358714 on Build GRCh38
rs993728638 Chr.10: 82359074-82359074 on Build GRCh38
rs747869353 Chr.10: 82358904-82358904 on Build GRCh38
rs896034017 Chr.10: 82359053-82359053 on Build GRCh38
rs902859001 Chr.10: 82358739-82358739 on Build GRCh38
rs551229180 Chr.10: 82358959-82358959 on Build GRCh38
rs764611896 Chr.10: 82358822-82358822 on Build GRCh38
rs978797133 Chr.10: 82358854-82358854 on Build GRCh38
rs764941110 Chr.10: 82358848-82358848 on Build GRCh38
rs765881380 Chr.10: 82358885-82358885 on Build GRCh38
rs185628605 Chr.10: 82358931-82358931 on Build GRCh38
rs548776537 Chr.10: 82358836-82358836 on Build GRCh38
rs745860661 Chr.10: 82358713-82358713 on Build GRCh38
rs753131620 Chr.10: 82358745-82358745 on Build GRCh38
rs759488158 Chr.10: 82358809-82358809 on Build GRCh38
rs772114036 Chr.10: 82358918-82358918 on Build GRCh38
rs756332932 Chr.10: 82358747-82358755 on Build GRCh38
rs780957321 Chr.10: 82358795-82358795 on Build GRCh38
rs754926761 Chr.10: 82358888-82358888 on Build GRCh38
rs999845042 Chr.10: 82358999-82358999 on Build GRCh38
rs1039975902 Chr.10: 82358953-82358953 on Build GRCh38
rs781602782 Chr.10: 82358816-82358816 on Build GRCh38
rs370478919 Chr.10: 82358805-82358805 on Build GRCh38
rs746740076 Chr.10: 82358716-82358716 on Build GRCh38
rs374849923 Chr.10: 82358849-82358849 on Build GRCh38
rs1024860967 Chr.10: 82359029-82359029 on Build GRCh38
rs775501942 Chr.10: 82359068-82359068 on Build GRCh38
rs1013079971 Chr.10: 82359021-82359021 on Build GRCh38
rs910657098 Chr.10: 82358924-82358924 on Build GRCh38
rs764492006 Chr.10: 82358749-82358749 on Build GRCh38
rs377542669 Chr.10: 82358778-82358778 on Build GRCh38
rs757893976 Chr.10: 82358860-82358860 on Build GRCh38
rs1024559755 Chr.10: 82358879-82358879 on Build GRCh38
rs773471294 Chr.10: 82358724-82358724 on Build GRCh38
rs564601376 Chr.10: 82358699-82358699 on Build GRCh38
rs766548381 Chr.10: 82358766-82358767 on Build GRCh38
rs563266860 Chr.10: 82358802-82358802 on Build GRCh38
rs761564574 Chr.10: 82358846-82358846 on Build GRCh38
rs999876279 Chr.10: 82358746-82358746 on Build GRCh38
rs756483335 Chr.10: 82358890-82358890 on Build GRCh38
rs756867808 Chr.10: 82358709-82358709 on Build GRCh38
rs905974055 Chr.10: 82359093-82359093 on Build GRCh38
rs1015810397 Chr.10: 82358759-82358759 on Build GRCh38
rs894253881 Chr.10: 82359017-82359017 on Build GRCh38
rs868605216 Chr.10: 82358915-82358915 on Build GRCh38
rs749884304 Chr.10: 82358763-82358763 on Build GRCh38
rs753522838 Chr.10: 82358705-82358705 on Build GRCh38
rs143302037 Chr.10: 82358997-82358997 on Build GRCh38
rs944551068 Chr.10: 82358930-82358930 on Build GRCh38
rs761240667 Chr.10: 82358820-82358820 on Build GRCh38
rs772173546 Chr.10: 82358818-82358818 on Build GRCh38
rs778580747 Chr.10: 82358762-82358762 on Build GRCh38
rs755470489 Chr.10: 82358693-82358693 on Build GRCh38
rs956224228 Chr.10: 82359144-82359144 on Build GRCh38
rs778089544 Chr.10: 82358962-82358962 on Build GRCh38
rs900227312 Chr.10: 82358682-82358682 on Build GRCh38
rs1054191678 Chr.10: 82359006-82359006 on Build GRCh38
rs767980524 Chr.10: 82358698-82358698 on Build GRCh38
rs533210959 Chr.10: 82358700-82358700 on Build GRCh38
rs774398423 Chr.10: 82358737-82358737 on Build GRCh38
rs749567041 Chr.10: 82358797-82358797 on Build GRCh38
rs779158215 Chr.10: 82358916-82358916 on Build GRCh38
rs766420708 Chr.10: 82358688-82358688 on Build GRCh38
rs1023059995 Chr.10: 82358837-82358837 on Build GRCh38
rs780975739 Chr.10: 82358896-82358896 on Build GRCh38
rs752071206 Chr.10: 82358685-82358685 on Build GRCh38
rs746943355 Chr.10: 82358779-82358779 on Build GRCh38
rs143196366 Chr.10: 82359107-82359107 on Build GRCh38
rs111753394 Chr.10: 82359089-82359089 on Build GRCh38
rs1033186176 Chr.10: 82358838-82358838 on Build GRCh38
rs201473623 Chr.10: 82358743-82358743 on Build GRCh38
rs771057615 Chr.10: 82358998-82358998 on Build GRCh38
rs752097883 Chr.10: 82358691-82358691 on Build GRCh38
rs149153629 Chr.10: 82358969-82358969 on Build GRCh38
rs201092412 Chr.10: 82358887-82358887 on Build GRCh38
rs556931925 Chr.10: 82359126-82359126 on Build GRCh38
rs543724200 Chr.10: 82359067-82359067 on Build GRCh38
rs576822436 Chr.10: 82359038-82359038 on Build GRCh38
rs377554965 Chr.10: 82358721-82358721 on Build GRCh38
rs753679022 Chr.10: 82358899-82358899 on Build GRCh38
rs763127200 Chr.10: 82358726-82358726 on Build GRCh38
rs778562404 Chr.10: 82358712-82358712 on Build GRCh38
rs771062511 Chr.10: 82358730-82358730 on Build GRCh38
rs760046822 Chr.10: 82358742-82358742 on Build GRCh38
rs757495719 Chr.10: 82358905-82358905 on Build GRCh38
rs758391265 Chr.10: 82358771-82358771 on Build GRCh38
rs552656708 Chr.10: 82358877-82358877 on Build GRCh38

View Full Product Details