Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 87753320-87753867 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATAD1
Entrez Gene ID
84896
Gene Name
ATPase family, AAA domain containing 1
Gene Aliases
AFDC1, FNP001, THORASE
UniGene
Hs.435948
Gene Chromosome Location
Chr.10: 87751512-87841345 on Build GRCh38
Transcripts
NM_001321967.1 NM_001321968.1 NM_001321969.1 NM_032810.3 NR_135914.1 XM_005270252.4 XM_011540302.1 XM_017016847.1 XM_017016848.1 XM_017016849.1 XM_017016850.1

SNP Detail

SNP ID Chromosome Location
rs1008044215 Chr.10: 87753372-87753372 on Build GRCh38
rs373457160 Chr.10: 87753458-87753458 on Build GRCh38
rs11546681 Chr.10: 87753365-87753365 on Build GRCh38
rs908684285 Chr.10: 87753531-87753531 on Build GRCh38
rs575292073 Chr.10: 87753749-87753749 on Build GRCh38
rs926853576 Chr.10: 87753620-87753620 on Build GRCh38
rs982302312 Chr.10: 87753604-87753604 on Build GRCh38
rs577589412 Chr.10: 87753817-87753817 on Build GRCh38
rs184646521 Chr.10: 87753685-87753685 on Build GRCh38
rs552525768 Chr.10: 87753595-87753595 on Build GRCh38
rs539213048 Chr.10: 87753742-87753742 on Build GRCh38
rs541763947 Chr.10: 87753832-87753832 on Build GRCh38
rs947128610 Chr.10: 87753705-87753705 on Build GRCh38
rs938268718 Chr.10: 87753636-87753636 on Build GRCh38
rs7481 Chr.10: 87753361-87753361 on Build GRCh38
rs7480 Chr.10: 87753695-87753695 on Build GRCh38
rs559227359 Chr.10: 87753787-87753787 on Build GRCh38
rs780455136 Chr.10: 87753470-87753470 on Build GRCh38
rs182044161 Chr.10: 87753603-87753603 on Build GRCh38
rs543985995 Chr.10: 87753763-87753763 on Build GRCh38
rs147644864 Chr.10: 87753552-87753552 on Build GRCh38
rs902963588 Chr.10: 87753837-87753837 on Build GRCh38
rs570400653 Chr.10: 87753482-87753482 on Build GRCh38
rs949520472 Chr.10: 87753560-87753560 on Build GRCh38
rs963970798 Chr.10: 87753434-87753434 on Build GRCh38
rs952788466 Chr.10: 87753486-87753486 on Build GRCh38
rs551996583 Chr.10: 87753419-87753419 on Build GRCh38
rs536659328 Chr.10: 87753842-87753843 on Build GRCh38
rs758503137 Chr.10: 87753446-87753446 on Build GRCh38
rs983291840 Chr.10: 87753530-87753530 on Build GRCh38
rs1019471084 Chr.10: 87753401-87753401 on Build GRCh38
rs776549475 Chr.10: 87753824-87753824 on Build GRCh38

View Full Product Details