Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 87770814-87771316 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATAD1
Entrez Gene ID
84896
Gene Name
ATPase family, AAA domain containing 1
Gene Aliases
AFDC1, FNP001, THORASE
UniGene
Hs.435948
Gene Chromosome Location
Chr.10: 87751512-87841345 on Build GRCh38
Transcripts
NM_001321967.1 NM_001321968.1 NM_001321969.1 NM_032810.3 NR_135914.1 XM_005270252.4 XM_011540302.1 XM_011540303.2 XM_017016847.1 XM_017016848.1 XM_017016849.1 XM_017016850.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1603822 c.734C>T p.S245L Chr.10: 87770998-87770998 on Build GRCh38
4442323 c.716G>A p.R239H Chr.10: 87771016-87771016 on Build GRCh38
6846793 c.713A>G p.N238S Chr.10: 87771019-87771019 on Build GRCh38
4663926 c.753G>A p.M251I Chr.10: 87770979-87770979 on Build GRCh38
5770768 c.715C>A p.R239S Chr.10: 87771017-87771017 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs562547031 Chr.10: 87771074-87771074 on Build GRCh38
rs765319610 Chr.10: 87770961-87770961 on Build GRCh38
rs561622231 Chr.10: 87770955-87770955 on Build GRCh38
rs1052384143 Chr.10: 87771143-87771143 on Build GRCh38
rs11202549 Chr.10: 87770948-87770948 on Build GRCh38
rs1016849773 Chr.10: 87770857-87770857 on Build GRCh38
rs192969409 Chr.10: 87770975-87770975 on Build GRCh38
rs772526467 Chr.10: 87770921-87770921 on Build GRCh38
rs889316566 Chr.10: 87771195-87771195 on Build GRCh38
rs770722824 Chr.10: 87771032-87771032 on Build GRCh38
rs550857030 Chr.10: 87771217-87771217 on Build GRCh38
rs886981600 Chr.10: 87771155-87771155 on Build GRCh38
rs374836218 Chr.10: 87770938-87770938 on Build GRCh38
rs761157221 Chr.10: 87770945-87770945 on Build GRCh38
rs368928987 Chr.10: 87770902-87770902 on Build GRCh38
rs769795343 Chr.10: 87771082-87771082 on Build GRCh38
rs766137546 Chr.10: 87770979-87770979 on Build GRCh38
rs922348297 Chr.10: 87771112-87771112 on Build GRCh38
rs374506570 Chr.10: 87770901-87770901 on Build GRCh38
rs11202550 Chr.10: 87771177-87771177 on Build GRCh38
rs114428714 Chr.10: 87770952-87770952 on Build GRCh38
rs1023689843 Chr.10: 87770875-87770875 on Build GRCh38
rs769112951 Chr.10: 87771051-87771051 on Build GRCh38
rs748970834 Chr.10: 87770912-87770912 on Build GRCh38
rs776954010 Chr.10: 87771058-87771058 on Build GRCh38
rs113578732 Chr.10: 87770869-87770869 on Build GRCh38
rs1039362964 Chr.10: 87771094-87771094 on Build GRCh38
rs371287150 Chr.10: 87770927-87770927 on Build GRCh38
rs898162396 Chr.10: 87771226-87771226 on Build GRCh38
rs933700800 Chr.10: 87771136-87771136 on Build GRCh38
rs368947548 Chr.10: 87770998-87770998 on Build GRCh38
rs747610694 Chr.10: 87770905-87770905 on Build GRCh38
rs1008209714 Chr.10: 87771197-87771197 on Build GRCh38
rs371502486 Chr.10: 87771066-87771066 on Build GRCh38
rs772980658 Chr.10: 87770909-87770909 on Build GRCh38
rs995135818 Chr.10: 87771291-87771291 on Build GRCh38
rs933477508 Chr.10: 87771118-87771118 on Build GRCh38
rs769473509 Chr.10: 87770907-87770907 on Build GRCh38
rs1012190489 Chr.10: 87770872-87770872 on Build GRCh38
rs533069872 Chr.10: 87771150-87771150 on Build GRCh38
rs747302499 Chr.10: 87771042-87771042 on Build GRCh38
rs780964486 Chr.10: 87771016-87771016 on Build GRCh38
rs777457295 Chr.10: 87771024-87771024 on Build GRCh38
rs748884882 Chr.10: 87771030-87771030 on Build GRCh38
rs942355325 Chr.10: 87771081-87771081 on Build GRCh38
rs951178065 Chr.10: 87770842-87770842 on Build GRCh38
rs770242821 Chr.10: 87770997-87770997 on Build GRCh38
rs752733088 Chr.10: 87771017-87771017 on Build GRCh38
rs552459814 Chr.10: 87771182-87771182 on Build GRCh38
rs144419191 Chr.10: 87770965-87770965 on Build GRCh38
rs936696254 Chr.10: 87770967-87770967 on Build GRCh38
rs544266071 Chr.10: 87771023-87771023 on Build GRCh38
rs1005916023 Chr.10: 87770852-87770852 on Build GRCh38

View Full Product Details