Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 88283981-88284521 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNLS
Entrez Gene ID
55328
Gene Name
renalase, FAD dependent amine oxidase
Gene Aliases
C10orf59, RENALASE
UniGene
Hs.149849
Gene Chromosome Location
Chr.10: 88176458-88583860 on Build GRCh38
Transcripts
NM_001031709.2 NM_018363.3 XM_005269950.3 XM_011539924.2 XM_017016380.1 XM_017016382.1

SNP Detail

SNP ID Chromosome Location
rs896891919 Chr.10: 88284018-88284018 on Build GRCh38
rs888098214 Chr.10: 88284049-88284049 on Build GRCh38
rs796309780 Chr.10: 88284342-88284342 on Build GRCh38
rs768415066 Chr.10: 88284087-88284087 on Build GRCh38
rs958028555 Chr.10: 88284426-88284426 on Build GRCh38
rs1011520416 Chr.10: 88284032-88284032 on Build GRCh38
rs80230329 Chr.10: 88284239-88284239 on Build GRCh38
rs949595047 Chr.10: 88284300-88284300 on Build GRCh38
rs540090485 Chr.10: 88284136-88284136 on Build GRCh38
rs773782292 Chr.10: 88284156-88284156 on Build GRCh38
rs1014040459 Chr.10: 88284187-88284187 on Build GRCh38
rs912685207 Chr.10: 88284101-88284101 on Build GRCh38
rs976907469 Chr.10: 88284475-88284475 on Build GRCh38
rs892832277 Chr.10: 88284416-88284416 on Build GRCh38
rs987279203 Chr.10: 88284092-88284092 on Build GRCh38
rs1011489164 Chr.10: 88284453-88284453 on Build GRCh38
rs944212850 Chr.10: 88284462-88284462 on Build GRCh38
rs7069968 Chr.10: 88284067-88284067 on Build GRCh38
rs763418196 Chr.10: 88284460-88284460 on Build GRCh38
rs779397501 Chr.10: 88284021-88284021 on Build GRCh38
rs966778534 Chr.10: 88284246-88284246 on Build GRCh38
rs931540549 Chr.10: 88284211-88284211 on Build GRCh38
rs112465465 Chr.10: 88284012-88284013 on Build GRCh38
rs918732606 Chr.10: 88284482-88284482 on Build GRCh38
rs895687681 Chr.10: 88284128-88284128 on Build GRCh38
rs559238572 Chr.10: 88284260-88284260 on Build GRCh38
rs1047004121 Chr.10: 88284315-88284315 on Build GRCh38
rs919940833 Chr.10: 88284155-88284155 on Build GRCh38
rs1027487270 Chr.10: 88284402-88284402 on Build GRCh38
rs540742725 Chr.10: 88284395-88284395 on Build GRCh38
rs185503816 Chr.10: 88284390-88284390 on Build GRCh38
rs199883970 Chr.10: 88284429-88284429 on Build GRCh38
rs964475998 Chr.10: 88284123-88284123 on Build GRCh38
rs906917585 Chr.10: 88284337-88284337 on Build GRCh38
rs192800917 Chr.10: 88284102-88284102 on Build GRCh38
rs866671128 Chr.10: 88284264-88284264 on Build GRCh38
rs1048551290 Chr.10: 88284047-88284047 on Build GRCh38

View Full Product Details