Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 89226939-89227487 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LIPA
Entrez Gene ID
3988
Gene Name
lipase A, lysosomal acid type
Gene Aliases
CESD, LAL
UniGene
Hs.643030
Gene Chromosome Location
Chr.10: 89213569-89252039 on Build GRCh38
Transcripts
NM_000235.3 NM_001127605.2 NM_001288979.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
428103 c.488C>T p.T163I Chr.10: 89226945-89226945 on Build GRCh38
4813002 c.494C>T p.T165I Chr.10: 89226945-89226945 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs964558319 Chr.10: 89227453-89227453 on Build GRCh38
rs549676436 Chr.10: 89227424-89227424 on Build GRCh38
rs771231905 Chr.10: 89227305-89227305 on Build GRCh38
rs911396027 Chr.10: 89227228-89227228 on Build GRCh38
rs560782412 Chr.10: 89227329-89227329 on Build GRCh38
rs762646420 Chr.10: 89226974-89226974 on Build GRCh38
rs758610920 Chr.10: 89227020-89227020 on Build GRCh38
rs748676985 Chr.10: 89227246-89227246 on Build GRCh38
rs776719147 Chr.10: 89227125-89227125 on Build GRCh38
rs1054718745 Chr.10: 89227234-89227234 on Build GRCh38
rs987290550 Chr.10: 89227422-89227422 on Build GRCh38
rs964498639 Chr.10: 89227238-89227238 on Build GRCh38
rs748072374 Chr.10: 89227060-89227060 on Build GRCh38
rs72468815 Chr.10: 89227100-89227101 on Build GRCh38
rs920225536 Chr.10: 89227077-89227077 on Build GRCh38
rs1025610260 Chr.10: 89227394-89227394 on Build GRCh38
rs372722864 Chr.10: 89227007-89227007 on Build GRCh38
rs962625611 Chr.10: 89227362-89227362 on Build GRCh38
rs571469230 Chr.10: 89227115-89227115 on Build GRCh38
rs768380059 Chr.10: 89227059-89227059 on Build GRCh38
rs377215072 Chr.10: 89227011-89227011 on Build GRCh38
rs1037330222 Chr.10: 89227069-89227069 on Build GRCh38
rs751257648 Chr.10: 89226984-89226984 on Build GRCh38
rs571984022 Chr.10: 89227233-89227233 on Build GRCh38
rs764569202 Chr.10: 89227000-89227000 on Build GRCh38
rs556376809 Chr.10: 89227091-89227091 on Build GRCh38
rs2254636 Chr.10: 89227250-89227250 on Build GRCh38
rs57350665 Chr.10: 89227321-89227321 on Build GRCh38
rs553651230 Chr.10: 89227158-89227158 on Build GRCh38
rs536329622 Chr.10: 89227425-89227426 on Build GRCh38
rs193022876 Chr.10: 89227399-89227399 on Build GRCh38
rs112248349 Chr.10: 89227012-89227012 on Build GRCh38
rs575813188 Chr.10: 89227253-89227253 on Build GRCh38
rs551345546 Chr.10: 89227105-89227105 on Build GRCh38
rs1047379974 Chr.10: 89227272-89227272 on Build GRCh38
rs760901300 Chr.10: 89226998-89226998 on Build GRCh38
rs1000654288 Chr.10: 89227140-89227140 on Build GRCh38
rs1016803149 Chr.10: 89227298-89227298 on Build GRCh38
rs986956307 Chr.10: 89227227-89227227 on Build GRCh38
rs567902552 Chr.10: 89227426-89227426 on Build GRCh38
rs1042107507 Chr.10: 89227267-89227267 on Build GRCh38
rs1044581348 Chr.10: 89227312-89227312 on Build GRCh38
rs778443906 Chr.10: 89227165-89227165 on Build GRCh38
rs756870829 Chr.10: 89227151-89227151 on Build GRCh38
rs184538935 Chr.10: 89227197-89227197 on Build GRCh38
rs911463408 Chr.10: 89227032-89227032 on Build GRCh38
rs747275421 Chr.10: 89227052-89227052 on Build GRCh38
rs994115209 Chr.10: 89227373-89227373 on Build GRCh38
rs749574811 Chr.10: 89227291-89227291 on Build GRCh38
rs191390336 Chr.10: 89227093-89227093 on Build GRCh38
rs902289974 Chr.10: 89227109-89227109 on Build GRCh38
rs774990016 Chr.10: 89227425-89227425 on Build GRCh38
rs2254640 Chr.10: 89227137-89227137 on Build GRCh38
rs755509339 Chr.10: 89227203-89227203 on Build GRCh38
rs772747557 Chr.10: 89227068-89227068 on Build GRCh38
rs576696638 Chr.10: 89227201-89227201 on Build GRCh38
rs75001468 Chr.10: 89227273-89227273 on Build GRCh38
rs780096608 Chr.10: 89227046-89227046 on Build GRCh38
rs956002266 Chr.10: 89227155-89227155 on Build GRCh38
rs894980826 Chr.10: 89227287-89227287 on Build GRCh38
rs34088667 Chr.10: 89227243-89227244 on Build GRCh38
rs962677026 Chr.10: 89227187-89227187 on Build GRCh38
rs748267444 Chr.10: 89226978-89226978 on Build GRCh38
rs936088755 Chr.10: 89227419-89227419 on Build GRCh38
rs189472160 Chr.10: 89227284-89227284 on Build GRCh38
rs557181411 Chr.10: 89227313-89227313 on Build GRCh38
rs942956204 Chr.10: 89227055-89227055 on Build GRCh38
rs370263738 Chr.10: 89227292-89227292 on Build GRCh38
rs769772927 Chr.10: 89227067-89227067 on Build GRCh38
rs542430662 Chr.10: 89227247-89227247 on Build GRCh38
rs762331015 Chr.10: 89227071-89227071 on Build GRCh38
rs754332616 Chr.10: 89227001-89227001 on Build GRCh38
rs567402009 Chr.10: 89227409-89227409 on Build GRCh38
rs564820135 Chr.10: 89227278-89227278 on Build GRCh38
rs1044958170 Chr.10: 89227135-89227135 on Build GRCh38

View Full Product Details