Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 89717310-89717842 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF20B
Entrez Gene ID
9585
Gene Name
kinesin family member 20B
Gene Aliases
CT90, KRMP1, MPHOSPH1, MPP-1, MPP1
UniGene
Hs.240
Gene Chromosome Location
Chr.10: 89701590-89774943 on Build GRCh38
Transcripts
NM_001284259.1 NM_016195.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6130880 c.1146T>C p.A382A Chr.10: 89717597-89717597 on Build GRCh38
5814800 c.1207A>G p.I403V Chr.10: 89717658-89717658 on Build GRCh38
4413462 c.1102T>C p.S368P Chr.10: 89717473-89717473 on Build GRCh38
4695990 c.1075A>C p.I359L Chr.10: 89717446-89717446 on Build GRCh38
6130879 c.1146T>C p.A382A Chr.10: 89717597-89717597 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746116898 Chr.10: 89717536-89717536 on Build GRCh38
rs958717434 Chr.10: 89717436-89717436 on Build GRCh38
rs112173887 Chr.10: 89717789-89717789 on Build GRCh38
rs139360783 Chr.10: 89717709-89717709 on Build GRCh38
rs76520569 Chr.10: 89717366-89717366 on Build GRCh38
rs754334159 Chr.10: 89717646-89717646 on Build GRCh38
rs761525225 Chr.10: 89717517-89717517 on Build GRCh38
rs548202480 Chr.10: 89717710-89717710 on Build GRCh38
rs150319905 Chr.10: 89717618-89717618 on Build GRCh38
rs758421430 Chr.10: 89717765-89717765 on Build GRCh38
rs750640636 Chr.10: 89717678-89717678 on Build GRCh38
rs551267859 Chr.10: 89717557-89717557 on Build GRCh38
rs751298392 Chr.10: 89717511-89717516 on Build GRCh38
rs771228993 Chr.10: 89717757-89717758 on Build GRCh38
rs771340403 Chr.10: 89717380-89717380 on Build GRCh38
rs371676289 Chr.10: 89717485-89717485 on Build GRCh38
rs777257304 Chr.10: 89717744-89717744 on Build GRCh38
rs761438044 Chr.10: 89717451-89717451 on Build GRCh38
rs375692649 Chr.10: 89717745-89717745 on Build GRCh38
rs879239534 Chr.10: 89717683-89717683 on Build GRCh38
rs556397412 Chr.10: 89717348-89717348 on Build GRCh38
rs149456198 Chr.10: 89717697-89717697 on Build GRCh38
rs765402722 Chr.10: 89717748-89717748 on Build GRCh38
rs769901683 Chr.10: 89717393-89717397 on Build GRCh38
rs976245285 Chr.10: 89717415-89717415 on Build GRCh38
rs891683724 Chr.10: 89717555-89717555 on Build GRCh38
rs767442477 Chr.10: 89717535-89717536 on Build GRCh38
rs769322639 Chr.10: 89717572-89717572 on Build GRCh38
rs762636055 Chr.10: 89717607-89717607 on Build GRCh38
rs776271223 Chr.10: 89717437-89717437 on Build GRCh38
rs767771862 Chr.10: 89717608-89717608 on Build GRCh38
rs756715290 Chr.10: 89717486-89717486 on Build GRCh38
rs763868862 Chr.10: 89717508-89717510 on Build GRCh38
rs760964846 Chr.10: 89717632-89717632 on Build GRCh38
rs764390114 Chr.10: 89717739-89717739 on Build GRCh38
rs768336681 Chr.10: 89717705-89717705 on Build GRCh38
rs759618504 Chr.10: 89717391-89717393 on Build GRCh38
rs779937913 Chr.10: 89717548-89717548 on Build GRCh38
rs775667770 Chr.10: 89717471-89717472 on Build GRCh38
rs759737602 Chr.10: 89717603-89717603 on Build GRCh38
rs370206377 Chr.10: 89717490-89717490 on Build GRCh38
rs531074538 Chr.10: 89717559-89717559 on Build GRCh38
rs758996295 Chr.10: 89717491-89717494 on Build GRCh38
rs369009574 Chr.10: 89717722-89717722 on Build GRCh38
rs556780625 Chr.10: 89717337-89717337 on Build GRCh38
rs150890799 Chr.10: 89717477-89717477 on Build GRCh38
rs555767778 Chr.10: 89717474-89717474 on Build GRCh38
rs769265565 Chr.10: 89717721-89717721 on Build GRCh38
rs754745441 Chr.10: 89717577-89717582 on Build GRCh38
rs763478746 Chr.10: 89717431-89717431 on Build GRCh38
rs773880743 Chr.10: 89717728-89717728 on Build GRCh38
rs1036132325 Chr.10: 89717410-89717410 on Build GRCh38
rs976781356 Chr.10: 89717752-89717752 on Build GRCh38
rs753897421 Chr.10: 89717505-89717505 on Build GRCh38
rs747342685 Chr.10: 89717702-89717704 on Build GRCh38
rs573176470 Chr.10: 89717426-89717426 on Build GRCh38
rs535558600 Chr.10: 89717432-89717432 on Build GRCh38
rs757418454 Chr.10: 89717510-89717510 on Build GRCh38
rs774728750 Chr.10: 89717384-89717384 on Build GRCh38
rs771969542 Chr.10: 89717542-89717542 on Build GRCh38
rs778436486 Chr.10: 89717619-89717620 on Build GRCh38
rs750361808 Chr.10: 89717544-89717545 on Build GRCh38
rs372350190 Chr.10: 89717661-89717661 on Build GRCh38
rs140162687 Chr.10: 89717580-89717580 on Build GRCh38
rs761061127 Chr.10: 89717729-89717729 on Build GRCh38
rs758759265 Chr.10: 89717689-89717689 on Build GRCh38
rs576724456 Chr.10: 89717341-89717341 on Build GRCh38
rs745592820 Chr.10: 89717404-89717404 on Build GRCh38
rs887758942 Chr.10: 89717687-89717687 on Build GRCh38
rs755658196 Chr.10: 89717549-89717550 on Build GRCh38
rs568056517 Chr.10: 89717782-89717782 on Build GRCh38
rs1031873378 Chr.10: 89717794-89717794 on Build GRCh38
rs1015151717 Chr.10: 89717409-89717409 on Build GRCh38
rs985243153 Chr.10: 89717466-89717466 on Build GRCh38
rs753528635 Chr.10: 89717556-89717557 on Build GRCh38
rs772026781 Chr.10: 89717411-89717411 on Build GRCh38
rs757365232 Chr.10: 89717657-89717657 on Build GRCh38
rs542233023 Chr.10: 89717463-89717463 on Build GRCh38
rs767920353 Chr.10: 89717476-89717476 on Build GRCh38
rs750587546 Chr.10: 89717763-89717763 on Build GRCh38
rs762584708 Chr.10: 89717725-89717725 on Build GRCh38
rs184665158 Chr.10: 89717563-89717563 on Build GRCh38
rs545697061 Chr.10: 89717344-89717344 on Build GRCh38
rs777705827 Chr.10: 89717488-89717488 on Build GRCh38
rs751756468 Chr.10: 89717550-89717550 on Build GRCh38
rs983652184 Chr.10: 89717399-89717399 on Build GRCh38
rs770563193 Chr.10: 89717726-89717726 on Build GRCh38
rs753166649 Chr.10: 89717611-89717611 on Build GRCh38
rs374218650 Chr.10: 89717640-89717640 on Build GRCh38
rs764809501 Chr.10: 89717454-89717454 on Build GRCh38
rs779238742 Chr.10: 89717531-89717531 on Build GRCh38
rs760714022 Chr.10: 89717429-89717429 on Build GRCh38
rs111940771 Chr.10: 89717696-89717696 on Build GRCh38

View Full Product Details