Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 96009819-96010361 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CC2D2B
Entrez Gene ID
387707
Gene Name
coiled-coil and C2 domain containing 2B
Gene Aliases
C10orf130, bA248J23.4
UniGene
Hs.538374
Gene Chromosome Location
Chr.10: 95974029-96035167 on Build GRCh38
Transcripts
NM_001001732.3 NM_001159747.1 XM_011539789.2 XM_011539790.2 XM_011539791.2 XM_011539792.2 XM_011539793.2 XM_011539795.2 XM_017016240.1 NR_038444.1

Gene Symbol
ENTPD1-AS1
Entrez Gene ID
728558
Gene Name
ENTPD1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.538374
Gene Chromosome Location
Chr.10: 95753206-96090235 on Build GRCh38
Transcripts
NM_001001732.3 NM_001159747.1 XM_011539789.2 XM_011539790.2 XM_011539791.2 XM_011539792.2 XM_011539793.2 XM_011539795.2 XM_017016240.1 NR_038444.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
921846 c.62G>A p.R21K Chr.10: 96009865-96009865 on Build GRCh38
921845 c.34A>T p.K12* Chr.10: 96009837-96009837 on Build GRCh38
6067133 c.389C>A p.S130Y Chr.10: 96009919-96009919 on Build GRCh38
6067131 c.116C>A p.S39Y Chr.10: 96009919-96009919 on Build GRCh38
4512637 c.90C>T p.F30F Chr.10: 96009893-96009893 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750326613 Chr.10: 96009883-96009883 on Build GRCh38
rs957009083 Chr.10: 96010176-96010176 on Build GRCh38
rs188469521 Chr.10: 96010256-96010256 on Build GRCh38
rs927055264 Chr.10: 96010253-96010253 on Build GRCh38
rs185488997 Chr.10: 96009961-96009961 on Build GRCh38
rs943931926 Chr.10: 96010091-96010092 on Build GRCh38
rs758314888 Chr.10: 96009900-96009900 on Build GRCh38
rs914361841 Chr.10: 96010028-96010028 on Build GRCh38
rs796487781 Chr.10: 96009973-96009973 on Build GRCh38
rs563463457 Chr.10: 96010116-96010116 on Build GRCh38
rs760951615 Chr.10: 96009847-96009847 on Build GRCh38
rs180813926 Chr.10: 96010174-96010174 on Build GRCh38
rs780761124 Chr.10: 96009931-96009931 on Build GRCh38
rs758817294 Chr.10: 96009972-96009973 on Build GRCh38
rs988878249 Chr.10: 96010185-96010185 on Build GRCh38
rs911061930 Chr.10: 96010062-96010062 on Build GRCh38
rs975208398 Chr.10: 96010129-96010142 on Build GRCh38
rs371240013 Chr.10: 96009955-96009955 on Build GRCh38
rs748857699 Chr.10: 96009931-96009931 on Build GRCh38
rs779916719 Chr.10: 96009857-96009860 on Build GRCh38
rs1006308072 Chr.10: 96010264-96010264 on Build GRCh38
rs747083753 Chr.10: 96010316-96010316 on Build GRCh38
rs779167220 Chr.10: 96009882-96009882 on Build GRCh38
rs754611444 Chr.10: 96009914-96009914 on Build GRCh38
rs1042848573 Chr.10: 96010245-96010245 on Build GRCh38
rs779866116 Chr.10: 96009910-96009910 on Build GRCh38
rs947093511 Chr.10: 96010221-96010221 on Build GRCh38
rs970674426 Chr.10: 96009941-96009942 on Build GRCh38
rs185617424 Chr.10: 96010214-96010214 on Build GRCh38
rs1056998549 Chr.10: 96010320-96010320 on Build GRCh38
rs376825763 Chr.10: 96009850-96009850 on Build GRCh38
rs987679792 Chr.10: 96010094-96010094 on Build GRCh38
rs539459005 Chr.10: 96009858-96009858 on Build GRCh38
rs994889893 Chr.10: 96010045-96010045 on Build GRCh38
rs188345915 Chr.10: 96010055-96010055 on Build GRCh38
rs552645367 Chr.10: 96009861-96009861 on Build GRCh38
rs887898036 Chr.10: 96010232-96010232 on Build GRCh38
rs1023530002 Chr.10: 96010297-96010297 on Build GRCh38
rs11188555 Chr.10: 96009996-96009996 on Build GRCh38
rs746846359 Chr.10: 96009911-96009911 on Build GRCh38
rs556814325 Chr.10: 96010311-96010311 on Build GRCh38
rs753032467 Chr.10: 96010097-96010097 on Build GRCh38
rs988539463 Chr.10: 96010016-96010016 on Build GRCh38
rs895719199 Chr.10: 96010331-96010331 on Build GRCh38
rs1004805310 Chr.10: 96010336-96010336 on Build GRCh38
rs747799779 Chr.10: 96009939-96009939 on Build GRCh38
rs4145209 Chr.10: 96010099-96010099 on Build GRCh38
rs139913395 Chr.10: 96009962-96009962 on Build GRCh38
rs867853994 Chr.10: 96010083-96010083 on Build GRCh38
rs953563694 Chr.10: 96010059-96010059 on Build GRCh38

View Full Product Details