Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 96203717-96204247 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BLNK
Entrez Gene ID
29760
Gene Name
B-cell linker
Gene Aliases
AGM4, BASH, BLNK-S, LY57, SLP-65, SLP65, bca
UniGene
Hs.665244
Gene Chromosome Location
Chr.10: 96189171-96271576 on Build GRCh38
Transcripts
NM_001114094.1 NM_001258440.1 NM_001258441.1 NM_001258442.1 NM_013314.3 NR_047680.1 NR_047681.1 NR_047682.1 NR_047683.1 XM_011539728.2 XM_017016159.1

SNP Detail

SNP ID Chromosome Location
rs781893863 Chr.10: 96204061-96204061 on Build GRCh38
rs376660776 Chr.10: 96204046-96204046 on Build GRCh38
rs796721264 Chr.10: 96203795-96203795 on Build GRCh38
rs1011793712 Chr.10: 96203813-96203813 on Build GRCh38
rs943669594 Chr.10: 96204099-96204099 on Build GRCh38
rs782631227 Chr.10: 96204006-96204006 on Build GRCh38
rs782569622 Chr.10: 96204107-96204107 on Build GRCh38
rs782739127 Chr.10: 96204070-96204070 on Build GRCh38
rs1015409404 Chr.10: 96203988-96203988 on Build GRCh38
rs144266674 Chr.10: 96204068-96204068 on Build GRCh38
rs1004945837 Chr.10: 96203986-96203986 on Build GRCh38
rs782545842 Chr.10: 96204139-96204139 on Build GRCh38
rs368840919 Chr.10: 96203987-96203987 on Build GRCh38
rs587680786 Chr.10: 96203899-96203899 on Build GRCh38
rs782201721 Chr.10: 96204161-96204161 on Build GRCh38
rs943805410 Chr.10: 96203750-96203750 on Build GRCh38
rs1043246199 Chr.10: 96204183-96204183 on Build GRCh38
rs782084331 Chr.10: 96204043-96204043 on Build GRCh38
rs782510147 Chr.10: 96204095-96204095 on Build GRCh38
rs905686861 Chr.10: 96204198-96204198 on Build GRCh38
rs199750156 Chr.10: 96203795-96203795 on Build GRCh38
rs771572053 Chr.10: 96203954-96203954 on Build GRCh38
rs963592131 Chr.10: 96204017-96204017 on Build GRCh38
rs1056958425 Chr.10: 96204209-96204209 on Build GRCh38
rs376056176 Chr.10: 96204029-96204029 on Build GRCh38
rs960027693 Chr.10: 96203969-96203969 on Build GRCh38
rs773027640 Chr.10: 96204059-96204059 on Build GRCh38
rs375361757 Chr.10: 96204045-96204045 on Build GRCh38
rs145125336 Chr.10: 96204016-96204016 on Build GRCh38
rs937297634 Chr.10: 96204203-96204203 on Build GRCh38
rs370164443 Chr.10: 96203783-96203783 on Build GRCh38
rs782351521 Chr.10: 96204175-96204175 on Build GRCh38
rs953725305 Chr.10: 96204048-96204048 on Build GRCh38
rs891652089 Chr.10: 96203767-96203767 on Build GRCh38
rs587689028 Chr.10: 96204047-96204047 on Build GRCh38
rs781847873 Chr.10: 96204074-96204074 on Build GRCh38
rs5787172 Chr.10: 96203783-96203783 on Build GRCh38
rs373842995 Chr.10: 96203754-96203754 on Build GRCh38
rs138963334 Chr.10: 96204125-96204125 on Build GRCh38
rs1001496981 Chr.10: 96203955-96203955 on Build GRCh38
rs782397728 Chr.10: 96204019-96204019 on Build GRCh38
rs782276368 Chr.10: 96203947-96203947 on Build GRCh38
rs987715589 Chr.10: 96204054-96204054 on Build GRCh38
rs782803552 Chr.10: 96204057-96204057 on Build GRCh38
rs1035686594 Chr.10: 96203966-96203966 on Build GRCh38
rs1042995032 Chr.10: 96203886-96203886 on Build GRCh38
rs781936696 Chr.10: 96204036-96204036 on Build GRCh38
rs35679287 Chr.10: 96203885-96203886 on Build GRCh38

View Full Product Details