Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 98252273-98252768 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOXL4
Entrez Gene ID
84171
Gene Name
lysyl oxidase like 4
Gene Aliases
LOXC
UniGene
Hs.306814
Gene Chromosome Location
Chr.10: 98247686-98268250 on Build GRCh38
Transcripts
NM_032211.6 XM_005270216.1 XM_006718020.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4010866 c.1945C>T p.P649S Chr.10: 98252359-98252359 on Build GRCh38
7351882 c.1951+2T>C p.? Chr.10: 98252351-98252351 on Build GRCh38
6551225 c.1887T>A p.N629K Chr.10: 98252417-98252417 on Build GRCh38
913674 c.1884C>T p.L628L Chr.10: 98252420-98252420 on Build GRCh38
28340 c.1848C>T p.S616S Chr.10: 98252456-98252456 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs529649800 Chr.10: 98252703-98252703 on Build GRCh38
rs940566892 Chr.10: 98252546-98252546 on Build GRCh38
rs776897971 Chr.10: 98252331-98252331 on Build GRCh38
rs932502946 Chr.10: 98252697-98252697 on Build GRCh38
rs749832699 Chr.10: 98252368-98252368 on Build GRCh38
rs778632063 Chr.10: 98252422-98252422 on Build GRCh38
rs372115263 Chr.10: 98252441-98252441 on Build GRCh38
rs201540369 Chr.10: 98252498-98252498 on Build GRCh38
rs748041604 Chr.10: 98252307-98252307 on Build GRCh38
rs367592797 Chr.10: 98252435-98252435 on Build GRCh38
rs775937122 Chr.10: 98252447-98252447 on Build GRCh38
rs200257522 Chr.10: 98252469-98252469 on Build GRCh38
rs901063273 Chr.10: 98252688-98252688 on Build GRCh38
rs764709809 Chr.10: 98252358-98252358 on Build GRCh38
rs35225911 Chr.10: 98252518-98252519 on Build GRCh38
rs754658874 Chr.10: 98252388-98252388 on Build GRCh38
rs753548741 Chr.10: 98252678-98252678 on Build GRCh38
rs773110000 Chr.10: 98252311-98252311 on Build GRCh38
rs1048538911 Chr.10: 98252617-98252617 on Build GRCh38
rs1034103529 Chr.10: 98252744-98252744 on Build GRCh38
rs915631380 Chr.10: 98252304-98252304 on Build GRCh38
rs767868821 Chr.10: 98252384-98252384 on Build GRCh38
rs778544111 Chr.10: 98252467-98252467 on Build GRCh38
rs564664927 Chr.10: 98252625-98252625 on Build GRCh38
rs375663705 Chr.10: 98252638-98252638 on Build GRCh38
rs770983737 Chr.10: 98252321-98252321 on Build GRCh38
rs777477009 Chr.10: 98252481-98252481 on Build GRCh38
rs745955149 Chr.10: 98252493-98252493 on Build GRCh38
rs1006925899 Chr.10: 98252664-98252664 on Build GRCh38
rs547749113 Chr.10: 98252729-98252729 on Build GRCh38
rs752087449 Chr.10: 98252457-98252457 on Build GRCh38
rs770070818 Chr.10: 98252494-98252494 on Build GRCh38
rs531948250 Chr.10: 98252626-98252626 on Build GRCh38
rs762271467 Chr.10: 98252458-98252458 on Build GRCh38
rs1019888648 Chr.10: 98252328-98252328 on Build GRCh38
rs779483867 Chr.10: 98252385-98252385 on Build GRCh38
rs375147335 Chr.10: 98252443-98252443 on Build GRCh38
rs753142849 Chr.10: 98252351-98252351 on Build GRCh38
rs757172743 Chr.10: 98252490-98252490 on Build GRCh38
rs931398369 Chr.10: 98252615-98252615 on Build GRCh38
rs959543917 Chr.10: 98252609-98252609 on Build GRCh38
rs375909179 Chr.10: 98252566-98252566 on Build GRCh38
rs746931431 Chr.10: 98252486-98252486 on Build GRCh38
rs931008629 Chr.10: 98252390-98252390 on Build GRCh38
rs754568734 Chr.10: 98252465-98252465 on Build GRCh38
rs755640664 Chr.10: 98252374-98252374 on Build GRCh38
rs927034310 Chr.10: 98252512-98252512 on Build GRCh38
rs901428781 Chr.10: 98252679-98252679 on Build GRCh38
rs765664092 Chr.10: 98252339-98252339 on Build GRCh38
rs545993261 Chr.10: 98252605-98252605 on Build GRCh38
rs1010933400 Chr.10: 98252533-98252533 on Build GRCh38
rs752414658 Chr.10: 98252473-98252473 on Build GRCh38
rs577419445 Chr.10: 98252558-98252558 on Build GRCh38
rs763410153 Chr.10: 98252452-98252452 on Build GRCh38
rs1036199263 Chr.10: 98252548-98252548 on Build GRCh38
rs776612272 Chr.10: 98252440-98252440 on Build GRCh38
rs763502058 Chr.10: 98252357-98252357 on Build GRCh38
rs992903342 Chr.10: 98252532-98252532 on Build GRCh38
rs966699391 Chr.10: 98252567-98252567 on Build GRCh38
rs765875135 Chr.10: 98252459-98252459 on Build GRCh38
rs140271302 Chr.10: 98252462-98252462 on Build GRCh38
rs759724953 Chr.10: 98252334-98252334 on Build GRCh38
rs937034115 Chr.10: 98252524-98252524 on Build GRCh38
rs968889379 Chr.10: 98252726-98252726 on Build GRCh38
rs562718897 Chr.10: 98252699-98252699 on Build GRCh38
rs367930549 Chr.10: 98252504-98252504 on Build GRCh38
rs34558912 Chr.10: 98252450-98252450 on Build GRCh38
rs200136133 Chr.10: 98252518-98252518 on Build GRCh38
rs1010882457 Chr.10: 98252683-98252683 on Build GRCh38
rs1048516824 Chr.10: 98252404-98252404 on Build GRCh38
rs771929604 Chr.10: 98252431-98252431 on Build GRCh38
rs371493970 Chr.10: 98252434-98252434 on Build GRCh38
rs552527561 Chr.10: 98252620-98252620 on Build GRCh38
rs939664757 Chr.10: 98252675-98252675 on Build GRCh38
rs760301990 Chr.10: 98252496-98252496 on Build GRCh38
rs773628452 Chr.10: 98252427-98252427 on Build GRCh38
rs747951884 Chr.10: 98252428-98252428 on Build GRCh38
rs772021262 Chr.10: 98252309-98252309 on Build GRCh38
rs893937307 Chr.10: 98252708-98252708 on Build GRCh38
rs747875708 Chr.10: 98252438-98252438 on Build GRCh38
rs73327609 Chr.10: 98252676-98252676 on Build GRCh38
rs908213518 Chr.10: 98252673-98252681 on Build GRCh38
rs1000304674 Chr.10: 98252727-98252727 on Build GRCh38
rs931061250 Chr.10: 98252650-98252650 on Build GRCh38
rs760814532 Chr.10: 98252317-98252317 on Build GRCh38
rs764429110 Chr.10: 98252454-98252454 on Build GRCh38
rs917205131 Chr.10: 98252542-98252542 on Build GRCh38
rs779124573 Chr.10: 98252326-98252326 on Build GRCh38
rs781284045 Chr.10: 98252492-98252492 on Build GRCh38
rs531494961 Chr.10: 98252718-98252718 on Build GRCh38

View Full Product Details