Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 101585842-101586346 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLL
Entrez Gene ID
27343
Gene Name
polymerase (DNA) lambda
Gene Aliases
BETAN, POLKAPPA
UniGene
Hs.523230
Gene Chromosome Location
Chr.10: 101578882-101588270 on Build GRCh38
Transcripts
NM_001174084.1 NM_001174085.1 NM_001308382.1 NM_013274.3 NR_033406.1 XM_006717775.1 XM_006717776.1 XM_006717777.1 XM_011539650.1 XM_011539651.1 XM_011539652.1 XM_011539653.1 XM_011539654.1 XM_011539655.1 XM_011539656.1 XM_011539657.1 XM_011539659.1 XM_011539660.1 XM_011539662.1 XM_011539663.1 XM_011539664.1 XM_011539665.2 XM_011539666.1 XM_011539667.1 XM_017016084.1 XM_017016085.1 XM_017016086.1 XM_017016087.1 XM_017016088.1 XM_017016089.1 XM_017016090.1 XM_017016091.1 XM_017016092.1 XM_017016093.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6752155 c.319C>T p.R107C Chr.10: 101585986-101585986 on Build GRCh38
5128152 c.314delC p.P105fs*6 Chr.10: 101585958-101585958 on Build GRCh38
6752175 c.178G>A p.V60M Chr.10: 101586127-101586127 on Build GRCh38
6752169 c.150G>A p.E50E Chr.10: 101586155-101586155 on Build GRCh38
6128188 c.167G>T p.R56L Chr.10: 101586138-101586138 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763387363 Chr.10: 101586199-101586199 on Build GRCh38
rs781504846 Chr.10: 101585997-101585997 on Build GRCh38
rs774622498 Chr.10: 101586070-101586070 on Build GRCh38
rs766683013 Chr.10: 101586263-101586263 on Build GRCh38
rs961306716 Chr.10: 101586144-101586144 on Build GRCh38
rs749708860 Chr.10: 101585968-101585968 on Build GRCh38
rs750178236 Chr.10: 101586040-101586042 on Build GRCh38
rs773579916 Chr.10: 101586078-101586078 on Build GRCh38
rs561526465 Chr.10: 101586055-101586055 on Build GRCh38
rs764072990 Chr.10: 101586026-101586029 on Build GRCh38
rs759853859 Chr.10: 101586185-101586185 on Build GRCh38
rs777078663 Chr.10: 101585976-101585976 on Build GRCh38
rs775992787 Chr.10: 101586194-101586194 on Build GRCh38
rs557827248 Chr.10: 101586156-101586156 on Build GRCh38
rs763128731 Chr.10: 101585909-101585909 on Build GRCh38
rs770553846 Chr.10: 101585985-101585985 on Build GRCh38
rs952520721 Chr.10: 101586157-101586157 on Build GRCh38
rs749316921 Chr.10: 101586058-101586058 on Build GRCh38
rs201767388 Chr.10: 101586139-101586139 on Build GRCh38
rs1044834686 Chr.10: 101585945-101585945 on Build GRCh38
rs761277868 Chr.10: 101586253-101586253 on Build GRCh38
rs57926494 Chr.10: 101586185-101586189 on Build GRCh38
rs369264701 Chr.10: 101586109-101586109 on Build GRCh38
rs895939021 Chr.10: 101585867-101585867 on Build GRCh38
rs755545668 Chr.10: 101586155-101586155 on Build GRCh38
rs775476715 Chr.10: 101585889-101585889 on Build GRCh38
rs934558312 Chr.10: 101586135-101586135 on Build GRCh38
rs751863141 Chr.10: 101586032-101586033 on Build GRCh38
rs3730462 Chr.10: 101586184-101586188 on Build GRCh38
rs751044136 Chr.10: 101586047-101586047 on Build GRCh38
rs201211517 Chr.10: 101586147-101586147 on Build GRCh38
rs759523385 Chr.10: 101585879-101585879 on Build GRCh38
rs148170530 Chr.10: 101585984-101585984 on Build GRCh38
rs775523053 Chr.10: 101585995-101585995 on Build GRCh38
rs753165243 Chr.10: 101586120-101586120 on Build GRCh38
rs757937376 Chr.10: 101585972-101585972 on Build GRCh38
rs548269958 Chr.10: 101586123-101586123 on Build GRCh38
rs370176524 Chr.10: 101586323-101586323 on Build GRCh38
rs762424614 Chr.10: 101586020-101586020 on Build GRCh38
rs775135336 Chr.10: 101585994-101585994 on Build GRCh38
rs771644362 Chr.10: 101585960-101585960 on Build GRCh38
rs770713461 Chr.10: 101585868-101585868 on Build GRCh38
rs375810045 Chr.10: 101586252-101586252 on Build GRCh38
rs779016004 Chr.10: 101586141-101586141 on Build GRCh38
rs1014344318 Chr.10: 101585870-101585870 on Build GRCh38
rs936781415 Chr.10: 101586307-101586307 on Build GRCh38
rs755028193 Chr.10: 101586100-101586100 on Build GRCh38
rs762743137 Chr.10: 101585998-101585998 on Build GRCh38
rs146124362 Chr.10: 101586071-101586071 on Build GRCh38
rs61757734 Chr.10: 101586103-101586103 on Build GRCh38
rs772229382 Chr.10: 101586072-101586072 on Build GRCh38
rs571075146 Chr.10: 101586264-101586264 on Build GRCh38
rs768787696 Chr.10: 101586001-101586001 on Build GRCh38
rs747311935 Chr.10: 101586162-101586162 on Build GRCh38
rs1017202998 Chr.10: 101585991-101585991 on Build GRCh38
rs377188876 Chr.10: 101586102-101586102 on Build GRCh38
rs779648167 Chr.10: 101586056-101586056 on Build GRCh38
rs1040632848 Chr.10: 101586043-101586043 on Build GRCh38
rs767081756 Chr.10: 101586206-101586206 on Build GRCh38
rs771032597 Chr.10: 101586169-101586169 on Build GRCh38
rs767816030 Chr.10: 101586019-101586019 on Build GRCh38
rs202208553 Chr.10: 101586186-101586186 on Build GRCh38
rs756068002 Chr.10: 101586089-101586090 on Build GRCh38
rs765141796 Chr.10: 101585888-101585888 on Build GRCh38
rs141683033 Chr.10: 101586138-101586138 on Build GRCh38
rs1042633908 Chr.10: 101586301-101586301 on Build GRCh38
rs773418506 Chr.10: 101586158-101586158 on Build GRCh38
rs1013707264 Chr.10: 101586126-101586126 on Build GRCh38
rs745476164 Chr.10: 101586054-101586054 on Build GRCh38
rs774291652 Chr.10: 101586007-101586007 on Build GRCh38
rs1000630474 Chr.10: 101586108-101586108 on Build GRCh38
rs923277556 Chr.10: 101586255-101586255 on Build GRCh38
rs751822184 Chr.10: 101585913-101585913 on Build GRCh38
rs754372306 Chr.10: 101586036-101586036 on Build GRCh38
rs777396875 Chr.10: 101586041-101586041 on Build GRCh38
rs779958111 Chr.10: 101586136-101586136 on Build GRCh38
rs755432831 Chr.10: 101586037-101586037 on Build GRCh38
rs368661800 Chr.10: 101586178-101586178 on Build GRCh38
rs762184144 Chr.10: 101586017-101586017 on Build GRCh38
rs201624181 Chr.10: 101585890-101585890 on Build GRCh38
rs755719141 Chr.10: 101586134-101586134 on Build GRCh38
rs369946628 Chr.10: 101585957-101585957 on Build GRCh38
rs746604600 Chr.10: 101585866-101585866 on Build GRCh38
rs964822629 Chr.10: 101586248-101586248 on Build GRCh38
rs776454975 Chr.10: 101585986-101585986 on Build GRCh38
rs987192163 Chr.10: 101586084-101586084 on Build GRCh38
rs201676435 Chr.10: 101585912-101585912 on Build GRCh38
rs757411742 Chr.10: 101586036-101586037 on Build GRCh38
rs925446914 Chr.10: 101586302-101586302 on Build GRCh38
rs372459718 Chr.10: 101585958-101585958 on Build GRCh38
rs143640062 Chr.10: 101585963-101585963 on Build GRCh38
rs767302525 Chr.10: 101586037-101586038 on Build GRCh38
rs756946598 Chr.10: 101586053-101586053 on Build GRCh38
rs745634557 Chr.10: 101585992-101585992 on Build GRCh38
rs750082153 Chr.10: 101586127-101586127 on Build GRCh38
rs766593391 Chr.10: 101586033-101586033 on Build GRCh38
rs760740139 Chr.10: 101586090-101586090 on Build GRCh38
rs761141737 Chr.10: 101586028-101586028 on Build GRCh38
rs776401875 Chr.10: 101585873-101585873 on Build GRCh38
rs143021263 Chr.10: 101586064-101586064 on Build GRCh38
rs746365299 Chr.10: 101586124-101586124 on Build GRCh38
rs147739279 Chr.10: 101586188-101586188 on Build GRCh38
rs780079927 Chr.10: 101586182-101586183 on Build GRCh38
rs757489920 Chr.10: 101585931-101585931 on Build GRCh38
rs1042601321 Chr.10: 101586076-101586076 on Build GRCh38

View Full Product Details