Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 102450065-102450563 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf95
Entrez Gene ID
79946
Gene Name
chromosome 10 open reading frame 95
Gene Aliases
-
UniGene
Hs.225084
Gene Chromosome Location
Chr.10: 102449837-102451543 on Build GRCh38
Transcripts
NM_024886.2 NR_038937.1 NR_038938.1

Gene Symbol
RPARP-AS1
Entrez Gene ID
100505761
Gene Name
RPARP antisense RNA 1
Gene Aliases
-
UniGene
Hs.656457
Gene Chromosome Location
Chr.10: 102449817-102456293 on Build GRCh38
Transcripts
NM_024886.2 NR_038937.1 NR_038938.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6051069 c.675C>A p.T225T Chr.10: 102450556-102450556 on Build GRCh38
5685562 c.678G>T p.A226A Chr.10: 102450553-102450553 on Build GRCh38
6435459 c.730G>T p.G244W Chr.10: 102450501-102450501 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1043593390 Chr.10: 102450178-102450178 on Build GRCh38
rs1009112424 Chr.10: 102450226-102450226 on Build GRCh38
rs913585129 Chr.10: 102450418-102450418 on Build GRCh38
rs898849894 Chr.10: 102450146-102450146 on Build GRCh38
rs763619520 Chr.10: 102450497-102450497 on Build GRCh38
rs774949295 Chr.10: 102450353-102450353 on Build GRCh38
rs946272090 Chr.10: 102450442-102450442 on Build GRCh38
rs573283657 Chr.10: 102450324-102450324 on Build GRCh38
rs764742984 Chr.10: 102450454-102450454 on Build GRCh38
rs753994369 Chr.10: 102450505-102450505 on Build GRCh38
rs1037398819 Chr.10: 102450141-102450141 on Build GRCh38
rs987733192 Chr.10: 102450396-102450396 on Build GRCh38
rs904713287 Chr.10: 102450204-102450204 on Build GRCh38
rs1000311109 Chr.10: 102450330-102450330 on Build GRCh38
rs1014939937 Chr.10: 102450306-102450306 on Build GRCh38
rs775216292 Chr.10: 102450484-102450484 on Build GRCh38
rs182460447 Chr.10: 102450088-102450088 on Build GRCh38
rs890718953 Chr.10: 102450207-102450208 on Build GRCh38
rs762388641 Chr.10: 102450395-102450395 on Build GRCh38
rs993175529 Chr.10: 102450177-102450177 on Build GRCh38
rs1010699104 Chr.10: 102450531-102450531 on Build GRCh38
rs937530253 Chr.10: 102450475-102450475 on Build GRCh38
rs768886883 Chr.10: 102450345-102450345 on Build GRCh38
rs937827623 Chr.10: 102450394-102450394 on Build GRCh38
rs1024693523 Chr.10: 102450373-102450373 on Build GRCh38
rs761160887 Chr.10: 102450445-102450445 on Build GRCh38
rs946331724 Chr.10: 102450133-102450133 on Build GRCh38
rs1048481904 Chr.10: 102450196-102450196 on Build GRCh38
rs1033082379 Chr.10: 102450346-102450346 on Build GRCh38
rs950128676 Chr.10: 102450509-102450509 on Build GRCh38
rs1014223132 Chr.10: 102450293-102450293 on Build GRCh38
rs1024328929 Chr.10: 102450532-102450532 on Build GRCh38
rs1001606031 Chr.10: 102450249-102450249 on Build GRCh38
rs755011561 Chr.10: 102450511-102450512 on Build GRCh38
rs575922128 Chr.10: 102450124-102450124 on Build GRCh38
rs751105898 Chr.10: 102450505-102450505 on Build GRCh38
rs1036749612 Chr.10: 102450529-102450529 on Build GRCh38
rs911681596 Chr.10: 102450381-102450381 on Build GRCh38
rs981964897 Chr.10: 102450383-102450383 on Build GRCh38
rs926106104 Chr.10: 102450473-102450473 on Build GRCh38
rs1017802791 Chr.10: 102450297-102450297 on Build GRCh38
rs1055920633 Chr.10: 102450262-102450262 on Build GRCh38
rs956276345 Chr.10: 102450352-102450352 on Build GRCh38
rs895815128 Chr.10: 102450273-102450273 on Build GRCh38
rs970675090 Chr.10: 102450379-102450379 on Build GRCh38
rs953269050 Chr.10: 102450361-102450361 on Build GRCh38
rs944955003 Chr.10: 102450450-102450450 on Build GRCh38
rs540584995 Chr.10: 102450444-102450444 on Build GRCh38
rs890092445 Chr.10: 102450448-102450448 on Build GRCh38
rs773445370 Chr.10: 102450429-102450429 on Build GRCh38
rs772747254 Chr.10: 102450414-102450414 on Build GRCh38
rs985835980 Chr.10: 102450363-102450363 on Build GRCh38
rs750124455 Chr.10: 102450520-102450520 on Build GRCh38
rs762412077 Chr.10: 102450496-102450496 on Build GRCh38
rs867283896 Chr.10: 102450237-102450237 on Build GRCh38
rs757179522 Chr.10: 102450508-102450508 on Build GRCh38
rs767544515 Chr.10: 102450517-102450517 on Build GRCh38
rs536564455 Chr.10: 102450143-102450143 on Build GRCh38
rs555209399 Chr.10: 102450219-102450219 on Build GRCh38

View Full Product Details