Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 114976216-114976761 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRUB1
Entrez Gene ID
142940
Gene Name
TruB pseudouridine synthase family member 1
Gene Aliases
PUS4
UniGene
Hs.21187
Gene Chromosome Location
Chr.10: 114938193-114977680 on Build GRCh38
Transcripts
NM_139169.4

SNP Detail

SNP ID Chromosome Location
rs908897577 Chr.10: 114976549-114976549 on Build GRCh38
rs974460826 Chr.10: 114976344-114976344 on Build GRCh38
rs777018847 Chr.10: 114976441-114976441 on Build GRCh38
rs972228628 Chr.10: 114976568-114976568 on Build GRCh38
rs775395402 Chr.10: 114976314-114976314 on Build GRCh38
rs987010727 Chr.10: 114976460-114976460 on Build GRCh38
rs544968508 Chr.10: 114976271-114976271 on Build GRCh38
rs41317246 Chr.10: 114976447-114976447 on Build GRCh38
rs564111883 Chr.10: 114976494-114976494 on Build GRCh38
rs1012088381 Chr.10: 114976288-114976288 on Build GRCh38
rs931369865 Chr.10: 114976646-114976646 on Build GRCh38
rs73372680 Chr.10: 114976614-114976614 on Build GRCh38
rs937927728 Chr.10: 114976665-114976665 on Build GRCh38
rs1055159354 Chr.10: 114976731-114976731 on Build GRCh38
rs530205033 Chr.10: 114976637-114976637 on Build GRCh38
rs563544592 Chr.10: 114976289-114976289 on Build GRCh38
rs141439536 Chr.10: 114976255-114976255 on Build GRCh38
rs542124190 Chr.10: 114976277-114976277 on Build GRCh38
rs941673013 Chr.10: 114976561-114976561 on Build GRCh38
rs566125081 Chr.10: 114976624-114976624 on Build GRCh38
rs560922674 Chr.10: 114976414-114976414 on Build GRCh38
rs1053674730 Chr.10: 114976668-114976668 on Build GRCh38
rs769621091 Chr.10: 114976292-114976292 on Build GRCh38
rs138352069 Chr.10: 114976593-114976593 on Build GRCh38
rs141707421 Chr.10: 114976729-114976729 on Build GRCh38
rs548390883 Chr.10: 114976701-114976701 on Build GRCh38
rs999734766 Chr.10: 114976300-114976300 on Build GRCh38
rs986322114 Chr.10: 114976538-114976538 on Build GRCh38
rs774097513 Chr.10: 114976735-114976735 on Build GRCh38
rs1028603309 Chr.10: 114976363-114976363 on Build GRCh38
rs530839902 Chr.10: 114976302-114976302 on Build GRCh38
rs958004930 Chr.10: 114976319-114976319 on Build GRCh38

View Full Product Details