Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 116460930-116461429 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PNLIPRP3
Entrez Gene ID
119548
Gene Name
pancreatic lipase related protein 3
Gene Aliases
-
UniGene
Hs.276724
Gene Chromosome Location
Chr.10: 116427879-116477961 on Build GRCh38
Transcripts
NM_001011709.2 XM_011539276.1 XM_011539277.1 XM_011539278.1 XM_011539279.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5630468 c.719A>G p.D240G Chr.10: 116461201-116461201 on Build GRCh38
1221354 c.721T>G p.F241V Chr.10: 116461203-116461203 on Build GRCh38
4559531 c.808G>A p.E270K Chr.10: 116461290-116461290 on Build GRCh38
683198 c.685+2T>A p.? Chr.10: 116461087-116461087 on Build GRCh38
6128433 c.566G>T p.G189V Chr.10: 116460966-116460966 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748719716 Chr.10: 116460994-116460994 on Build GRCh38
rs757723005 Chr.10: 116461224-116461224 on Build GRCh38
rs781704331 Chr.10: 116461229-116461229 on Build GRCh38
rs544377195 Chr.10: 116460983-116460983 on Build GRCh38
rs773291815 Chr.10: 116461152-116461152 on Build GRCh38
rs758748610 Chr.10: 116461089-116461089 on Build GRCh38
rs750646819 Chr.10: 116461217-116461217 on Build GRCh38
rs199970109 Chr.10: 116461012-116461012 on Build GRCh38
rs561083050 Chr.10: 116461131-116461131 on Build GRCh38
rs778594583 Chr.10: 116461067-116461067 on Build GRCh38
rs114674677 Chr.10: 116461046-116461046 on Build GRCh38
rs749440768 Chr.10: 116461187-116461187 on Build GRCh38
rs754546541 Chr.10: 116461062-116461062 on Build GRCh38
rs755781497 Chr.10: 116461216-116461216 on Build GRCh38
rs746136260 Chr.10: 116461124-116461124 on Build GRCh38
rs192845889 Chr.10: 116461333-116461333 on Build GRCh38
rs138106260 Chr.10: 116461051-116461051 on Build GRCh38
rs781085047 Chr.10: 116461121-116461121 on Build GRCh38
rs973051060 Chr.10: 116461122-116461122 on Build GRCh38
rs552112581 Chr.10: 116461178-116461178 on Build GRCh38
rs764855654 Chr.10: 116461180-116461180 on Build GRCh38
rs141556677 Chr.10: 116461030-116461030 on Build GRCh38
rs779916775 Chr.10: 116460975-116460975 on Build GRCh38
rs976446689 Chr.10: 116461284-116461285 on Build GRCh38
rs776616971 Chr.10: 116461251-116461251 on Build GRCh38
rs58155228 Chr.10: 116461298-116461298 on Build GRCh38
rs369900742 Chr.10: 116460995-116460995 on Build GRCh38
rs749460143 Chr.10: 116461235-116461235 on Build GRCh38
rs764738281 Chr.10: 116461276-116461276 on Build GRCh38
rs372620371 Chr.10: 116461165-116461165 on Build GRCh38
rs760638424 Chr.10: 116461162-116461162 on Build GRCh38
rs772010729 Chr.10: 116461239-116461239 on Build GRCh38
rs910931919 Chr.10: 116461169-116461169 on Build GRCh38
rs780165597 Chr.10: 116461331-116461331 on Build GRCh38
rs759033961 Chr.10: 116461253-116461253 on Build GRCh38
rs773980851 Chr.10: 116461032-116461032 on Build GRCh38
rs373999631 Chr.10: 116461010-116461010 on Build GRCh38
rs537229443 Chr.10: 116461289-116461289 on Build GRCh38
rs201251164 Chr.10: 116461045-116461045 on Build GRCh38
rs778053849 Chr.10: 116461209-116461209 on Build GRCh38
rs200485970 Chr.10: 116461058-116461058 on Build GRCh38
rs111998308 Chr.10: 116461009-116461009 on Build GRCh38
rs767970806 Chr.10: 116461308-116461308 on Build GRCh38
rs377105460 Chr.10: 116461050-116461050 on Build GRCh38
rs778231333 Chr.10: 116460997-116460997 on Build GRCh38
rs369099975 Chr.10: 116461104-116461104 on Build GRCh38
rs762545146 Chr.10: 116461293-116461293 on Build GRCh38
rs775764250 Chr.10: 116461135-116461135 on Build GRCh38
rs768836080 Chr.10: 116461315-116461315 on Build GRCh38
rs747148207 Chr.10: 116461002-116461002 on Build GRCh38
rs377433450 Chr.10: 116461215-116461215 on Build GRCh38
rs922299834 Chr.10: 116461290-116461290 on Build GRCh38
rs752389651 Chr.10: 116461195-116461195 on Build GRCh38
rs999761523 Chr.10: 116460989-116460989 on Build GRCh38
rs543246754 Chr.10: 116461142-116461142 on Build GRCh38
rs548386035 Chr.10: 116461245-116461245 on Build GRCh38
rs200957257 Chr.10: 116461141-116461141 on Build GRCh38
rs568576537 Chr.10: 116461247-116461247 on Build GRCh38
rs894369343 Chr.10: 116461139-116461139 on Build GRCh38
rs565944175 Chr.10: 116461230-116461230 on Build GRCh38
rs929675282 Chr.10: 116461021-116461021 on Build GRCh38
rs200148572 Chr.10: 116461278-116461278 on Build GRCh38
rs201998457 Chr.10: 116461237-116461237 on Build GRCh38
rs768519982 Chr.10: 116461024-116461024 on Build GRCh38
rs528534828 Chr.10: 116461233-116461233 on Build GRCh38
rs903518394 Chr.10: 116461228-116461228 on Build GRCh38
rs747450447 Chr.10: 116461112-116461112 on Build GRCh38
rs373615090 Chr.10: 116461114-116461114 on Build GRCh38
rs775853093 Chr.10: 116461112-116461113 on Build GRCh38
rs761639643 Chr.10: 116461036-116461036 on Build GRCh38
rs774605588 Chr.10: 116461060-116461060 on Build GRCh38
rs957546861 Chr.10: 116461187-116461187 on Build GRCh38
rs760262395 Chr.10: 116461052-116461052 on Build GRCh38
rs749550155 Chr.10: 116461337-116461337 on Build GRCh38
rs750939853 Chr.10: 116461329-116461329 on Build GRCh38
rs755378827 Chr.10: 116460982-116460982 on Build GRCh38
rs770335870 Chr.10: 116461037-116461037 on Build GRCh38
rs758767183 Chr.10: 116461206-116461206 on Build GRCh38
rs757598389 Chr.10: 116461302-116461302 on Build GRCh38
rs753051705 Chr.10: 116461068-116461068 on Build GRCh38
rs187436022 Chr.10: 116461126-116461126 on Build GRCh38
rs1024102263 Chr.10: 116461016-116461016 on Build GRCh38
rs1040702560 Chr.10: 116461007-116461007 on Build GRCh38
rs896657219 Chr.10: 116461263-116461263 on Build GRCh38
rs149530976 Chr.10: 116461177-116461177 on Build GRCh38
rs755666360 Chr.10: 116460974-116460974 on Build GRCh38

View Full Product Details