Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 117130299-117130801 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VAX1
Entrez Gene ID
11023
Gene Name
ventral anterior homeobox 1
Gene Aliases
MCOPS11
UniGene
Hs.441536
Gene Chromosome Location
Chr.10: 117128521-117138301 on Build GRCh38
Transcripts
NM_199131.2

SNP Detail

SNP ID Chromosome Location
rs1045824080 Chr.10: 117130345-117130345 on Build GRCh38
rs913785937 Chr.10: 117130469-117130469 on Build GRCh38
rs771031239 Chr.10: 117130697-117130697 on Build GRCh38
rs975200230 Chr.10: 117130329-117130329 on Build GRCh38
rs982954259 Chr.10: 117130516-117130516 on Build GRCh38
rs1046620379 Chr.10: 117130479-117130479 on Build GRCh38
rs935132069 Chr.10: 117130724-117130724 on Build GRCh38
rs12777713 Chr.10: 117130507-117130507 on Build GRCh38
rs906359406 Chr.10: 117130397-117130397 on Build GRCh38
rs376788253 Chr.10: 117130466-117130466 on Build GRCh38
rs1045792491 Chr.10: 117130638-117130638 on Build GRCh38
rs76598728 Chr.10: 117130495-117130497 on Build GRCh38
rs893691789 Chr.10: 117130761-117130761 on Build GRCh38
rs1020200154 Chr.10: 117130696-117130696 on Build GRCh38
rs875354 Chr.10: 117130704-117130704 on Build GRCh38
rs201256530 Chr.10: 117130496-117130505 on Build GRCh38
rs924735386 Chr.10: 117130653-117130653 on Build GRCh38
rs140332986 Chr.10: 117130497-117130507 on Build GRCh38
rs989161246 Chr.10: 117130441-117130441 on Build GRCh38
rs75602873 Chr.10: 117130499-117130509 on Build GRCh38
rs554574427 Chr.10: 117130475-117130475 on Build GRCh38
rs528211724 Chr.10: 117130347-117130347 on Build GRCh38
rs910093668 Chr.10: 117130520-117130520 on Build GRCh38
rs1028049603 Chr.10: 117130751-117130751 on Build GRCh38
rs201620036 Chr.10: 117130495-117130505 on Build GRCh38
rs1052222559 Chr.10: 117130744-117130744 on Build GRCh38
rs75502930 Chr.10: 117130501-117130506 on Build GRCh38
rs995438638 Chr.10: 117130510-117130510 on Build GRCh38
rs145213789 Chr.10: 117130552-117130552 on Build GRCh38
rs906399594 Chr.10: 117130481-117130481 on Build GRCh38
rs963010403 Chr.10: 117130524-117130524 on Build GRCh38
rs1055827876 Chr.10: 117130489-117130489 on Build GRCh38
rs887317501 Chr.10: 117130585-117130585 on Build GRCh38
rs182585787 Chr.10: 117130482-117130482 on Build GRCh38
rs975147935 Chr.10: 117130739-117130739 on Build GRCh38
rs936353149 Chr.10: 117130420-117130420 on Build GRCh38
rs767728142 Chr.10: 117130505-117130506 on Build GRCh38
rs772556817 Chr.10: 117130734-117130734 on Build GRCh38
rs148741605 Chr.10: 117130629-117130629 on Build GRCh38
rs920914078 Chr.10: 117130387-117130387 on Build GRCh38
rs915903057 Chr.10: 117130566-117130566 on Build GRCh38

View Full Product Details