Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 119033633-119034178 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NANOS1
Entrez Gene ID
340719
Gene Name
nanos C2HC-type zinc finger 1
Gene Aliases
EC_Rep1a, NOS-1, NOS1, SPGF12, ZC2HC12A
UniGene
Hs.591918
Gene Chromosome Location
Chr.10: 119029716-119033732 on Build GRCh38
Transcripts
NM_199461.3

SNP Detail

SNP ID Chromosome Location
rs909412171 Chr.10: 119033746-119033749 on Build GRCh38
rs938155208 Chr.10: 119033778-119033778 on Build GRCh38
rs895104302 Chr.10: 119034016-119034016 on Build GRCh38
rs983846259 Chr.10: 119033741-119033744 on Build GRCh38
rs1042405010 Chr.10: 119033883-119033884 on Build GRCh38
rs891860963 Chr.10: 119033900-119033900 on Build GRCh38
rs950522444 Chr.10: 119033679-119033679 on Build GRCh38
rs974997238 Chr.10: 119033883-119033883 on Build GRCh38
rs990526996 Chr.10: 119034129-119034129 on Build GRCh38
rs3061097 Chr.10: 119033873-119033873 on Build GRCh38
rs946165146 Chr.10: 119033913-119033913 on Build GRCh38
rs983608574 Chr.10: 119033820-119033820 on Build GRCh38
rs796201669 Chr.10: 119033872-119033873 on Build GRCh38
rs991519604 Chr.10: 119033834-119033838 on Build GRCh38
rs142128350 Chr.10: 119034149-119034149 on Build GRCh38
rs771586418 Chr.10: 119034084-119034084 on Build GRCh38
rs983219846 Chr.10: 119034061-119034061 on Build GRCh38
rs994460504 Chr.10: 119034030-119034030 on Build GRCh38
rs868485028 Chr.10: 119033886-119033886 on Build GRCh38
rs1027354301 Chr.10: 119034018-119034018 on Build GRCh38
rs778286189 Chr.10: 119034092-119034092 on Build GRCh38
rs546469511 Chr.10: 119033856-119033856 on Build GRCh38
rs35295352 Chr.10: 119034071-119034072 on Build GRCh38
rs562127934 Chr.10: 119034017-119034017 on Build GRCh38
rs577428406 Chr.10: 119033706-119033706 on Build GRCh38
rs1042179521 Chr.10: 119033927-119033927 on Build GRCh38
rs138984118 Chr.10: 119033990-119033990 on Build GRCh38
rs548264785 Chr.10: 119033876-119033876 on Build GRCh38
rs377707161 Chr.10: 119034012-119034012 on Build GRCh38
rs118174828 Chr.10: 119033948-119033948 on Build GRCh38
rs974944936 Chr.10: 119034102-119034102 on Build GRCh38
rs952347381 Chr.10: 119034060-119034060 on Build GRCh38
rs1019825966 Chr.10: 119033993-119033993 on Build GRCh38
rs869202862 Chr.10: 119033886-119033887 on Build GRCh38
rs1054483374 Chr.10: 119033894-119033894 on Build GRCh38
rs958406722 Chr.10: 119034144-119034144 on Build GRCh38
rs796365261 Chr.10: 119033873-119033876 on Build GRCh38
rs950492958 Chr.10: 119034028-119034028 on Build GRCh38
rs1025324557 Chr.10: 119034036-119034036 on Build GRCh38
rs1053785026 Chr.10: 119033928-119033928 on Build GRCh38
rs559844227 Chr.10: 119033861-119033861 on Build GRCh38
rs778479391 Chr.10: 119033835-119033835 on Build GRCh38
rs182166435 Chr.10: 119034121-119034121 on Build GRCh38
rs532849292 Chr.10: 119034125-119034125 on Build GRCh38
rs906378190 Chr.10: 119033945-119033945 on Build GRCh38
rs935996373 Chr.10: 119033890-119033890 on Build GRCh38
rs138749530 Chr.10: 119033884-119033887 on Build GRCh38
rs756766874 Chr.10: 119034126-119034127 on Build GRCh38
rs879543564 Chr.10: 119034006-119034008 on Build GRCh38
rs918212239 Chr.10: 119033680-119033684 on Build GRCh38
rs570735040 Chr.10: 119034090-119034090 on Build GRCh38

View Full Product Details