Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 122838932-122839433 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUZD1
Entrez Gene ID
50624
Gene Name
CUB and zona pellucida like domains 1
Gene Aliases
ERG-1, ERG1, ITMAP1, UO-44
UniGene
Hs.647182
Gene Chromosome Location
Chr.10: 122832155-122846175 on Build GRCh38
Transcripts
NM_022034.5 NR_037912.1 NR_037915.1

Gene Symbol
FAM24B-CUZD1
Entrez Gene ID
100533195
Gene Name
FAM24B-CUZD1 readthrough
Gene Aliases
-
UniGene
Hs.647182
Gene Chromosome Location
Chr.10: 122832155-122879641 on Build GRCh38
Transcripts
NM_022034.5 NR_037912.1 NR_037915.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
465382 c.406A>G p.R136G Chr.10: 122839059-122839059 on Build GRCh38
6861547 c.356C>T p.S119L Chr.10: 122839109-122839109 on Build GRCh38
2020818 c.330C>T p.N110N Chr.10: 122839135-122839135 on Build GRCh38
915719 c.407G>T p.R136I Chr.10: 122839058-122839058 on Build GRCh38
1346502 c.269T>G p.I90S Chr.10: 122839196-122839196 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771319979 Chr.10: 122839278-122839278 on Build GRCh38
rs776979000 Chr.10: 122839280-122839280 on Build GRCh38
rs145757808 Chr.10: 122839129-122839129 on Build GRCh38
rs143659470 Chr.10: 122839134-122839134 on Build GRCh38
rs781278916 Chr.10: 122839198-122839198 on Build GRCh38
rs764462252 Chr.10: 122839069-122839069 on Build GRCh38
rs767849604 Chr.10: 122839092-122839092 on Build GRCh38
rs563640546 Chr.10: 122839105-122839105 on Build GRCh38
rs746886583 Chr.10: 122839108-122839108 on Build GRCh38
rs746322795 Chr.10: 122839259-122839259 on Build GRCh38
rs146535754 Chr.10: 122839178-122839178 on Build GRCh38
rs374739388 Chr.10: 122839269-122839269 on Build GRCh38
rs539704104 Chr.10: 122839319-122839319 on Build GRCh38
rs766133030 Chr.10: 122839246-122839247 on Build GRCh38
rs750738667 Chr.10: 122839014-122839015 on Build GRCh38
rs371179428 Chr.10: 122839026-122839026 on Build GRCh38
rs754021931 Chr.10: 122839170-122839170 on Build GRCh38
rs757078432 Chr.10: 122839190-122839190 on Build GRCh38
rs1007879407 Chr.10: 122839327-122839327 on Build GRCh38
rs139643593 Chr.10: 122839117-122839117 on Build GRCh38
rs780911533 Chr.10: 122839127-122839127 on Build GRCh38
rs766165389 Chr.10: 122839158-122839158 on Build GRCh38
rs374258835 Chr.10: 122839192-122839192 on Build GRCh38
rs774825539 Chr.10: 122839075-122839075 on Build GRCh38
rs111772324 Chr.10: 122839392-122839392 on Build GRCh38
rs144877107 Chr.10: 122839281-122839281 on Build GRCh38
rs148941738 Chr.10: 122839130-122839130 on Build GRCh38
rs949201765 Chr.10: 122839188-122839188 on Build GRCh38
rs774718187 Chr.10: 122839141-122839141 on Build GRCh38
rs760945039 Chr.10: 122839159-122839159 on Build GRCh38
rs151179532 Chr.10: 122838960-122838960 on Build GRCh38
rs149807903 Chr.10: 122839120-122839120 on Build GRCh38
rs779646649 Chr.10: 122839221-122839221 on Build GRCh38
rs572884821 Chr.10: 122839007-122839007 on Build GRCh38
rs141440856 Chr.10: 122839183-122839183 on Build GRCh38
rs375496918 Chr.10: 122839008-122839008 on Build GRCh38
rs887709454 Chr.10: 122839324-122839324 on Build GRCh38
rs34089036 Chr.10: 122839182-122839183 on Build GRCh38
rs756148978 Chr.10: 122839003-122839003 on Build GRCh38
rs764372470 Chr.10: 122838983-122838983 on Build GRCh38
rs748771496 Chr.10: 122839135-122839135 on Build GRCh38
rs758363638 Chr.10: 122839102-122839102 on Build GRCh38
rs758273368 Chr.10: 122839016-122839016 on Build GRCh38
rs370935434 Chr.10: 122839260-122839260 on Build GRCh38
rs997277696 Chr.10: 122839346-122839346 on Build GRCh38
rs1031403528 Chr.10: 122839369-122839369 on Build GRCh38
rs1048834761 Chr.10: 122839310-122839310 on Build GRCh38
rs267602403 Chr.10: 122839203-122839203 on Build GRCh38
rs140178574 Chr.10: 122839020-122839020 on Build GRCh38
rs76764875 Chr.10: 122838997-122838997 on Build GRCh38
rs776141193 Chr.10: 122839031-122839031 on Build GRCh38
rs548655795 Chr.10: 122839165-122839165 on Build GRCh38
rs138976047 Chr.10: 122839222-122839222 on Build GRCh38
rs751809190 Chr.10: 122838986-122838986 on Build GRCh38
rs775978332 Chr.10: 122839054-122839054 on Build GRCh38
rs761100887 Chr.10: 122839277-122839277 on Build GRCh38
rs931320015 Chr.10: 122839296-122839296 on Build GRCh38
rs749725716 Chr.10: 122839376-122839376 on Build GRCh38
rs144481694 Chr.10: 122839034-122839034 on Build GRCh38
rs1054624187 Chr.10: 122838978-122838978 on Build GRCh38
rs149885016 Chr.10: 122839093-122839093 on Build GRCh38
rs766590573 Chr.10: 122839168-122839168 on Build GRCh38
rs760940694 Chr.10: 122839167-122839167 on Build GRCh38
rs267602402 Chr.10: 122839125-122839125 on Build GRCh38
rs748540906 Chr.10: 122839246-122839246 on Build GRCh38
rs780362446 Chr.10: 122839335-122839335 on Build GRCh38
rs763405638 Chr.10: 122839065-122839065 on Build GRCh38
rs752713645 Chr.10: 122839101-122839101 on Build GRCh38
rs376135910 Chr.10: 122839035-122839035 on Build GRCh38
rs762055108 Chr.10: 122839077-122839077 on Build GRCh38
rs760728510 Chr.10: 122839094-122839094 on Build GRCh38
rs766229922 Chr.10: 122839095-122839095 on Build GRCh38
rs953057883 Chr.10: 122839111-122839111 on Build GRCh38
rs750558552 Chr.10: 122839002-122839002 on Build GRCh38
rs747811930 Chr.10: 122839334-122839334 on Build GRCh38
rs948953931 Chr.10: 122839248-122839248 on Build GRCh38
rs543761437 Chr.10: 122839098-122839098 on Build GRCh38
rs145145827 Chr.10: 122839209-122839209 on Build GRCh38
rs550968134 Chr.10: 122839264-122839264 on Build GRCh38
rs368191862 Chr.10: 122839245-122839245 on Build GRCh38
rs202220182 Chr.10: 122839137-122839137 on Build GRCh38
rs147829911 Chr.10: 122839099-122839099 on Build GRCh38
rs189988968 Chr.10: 122838968-122838968 on Build GRCh38
rs757448158 Chr.10: 122838996-122838996 on Build GRCh38
rs762301050 Chr.10: 122839155-122839155 on Build GRCh38
rs528627329 Chr.10: 122839162-122839162 on Build GRCh38
rs975656118 Chr.10: 122839082-122839082 on Build GRCh38
rs770655210 Chr.10: 122839022-122839022 on Build GRCh38
rs764182845 Chr.10: 122839182-122839182 on Build GRCh38
rs767689270 Chr.10: 122838979-122838981 on Build GRCh38

View Full Product Details