Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 122993595-122994092 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IKZF5
Entrez Gene ID
64376
Gene Name
IKAROS family zinc finger 5
Gene Aliases
PEGASUS, ZNFN1A5
UniGene
Hs.501289
Gene Chromosome Location
Chr.10: 122990806-123008850 on Build GRCh38
Transcripts
NM_001271840.1 NR_073475.1 NR_073476.1 NR_073477.1 NR_073478.1 NR_073479.1 XM_006717947.3 XM_017016550.1 XM_017016551.1 XM_017016552.1 XM_017016553.1 XM_017016554.1 XM_017016555.1

Gene Symbol
PSTK
Entrez Gene ID
118672
Gene Name
phosphoseryl-tRNA kinase
Gene Aliases
C10orf89
UniGene
Hs.281004
Gene Chromosome Location
Chr.10: 122980040-122997519 on Build GRCh38
Transcripts
NM_001271840.1 NR_073475.1 NR_073476.1 NR_073477.1 NR_073478.1 NR_073479.1 XM_006717947.3 XM_017016550.1 XM_017016551.1 XM_017016552.1 XM_017016553.1 XM_017016554.1 XM_017016555.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4012362 c.1244A>G p.Q415R Chr.10: 122993796-122993796 on Build GRCh38
1470296 c.1048A>C p.T350P Chr.10: 122993992-122993992 on Build GRCh38
3358674 c.1247A>G p.H416R Chr.10: 122993793-122993793 on Build GRCh38
5559416 c.1088T>A p.L363Q Chr.10: 122993952-122993952 on Build GRCh38
4165413 c.1228C>G p.H410D Chr.10: 122993812-122993812 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs546901184 Chr.10: 122993912-122993912 on Build GRCh38
rs911291891 Chr.10: 122993795-122993795 on Build GRCh38
rs142260324 Chr.10: 122993929-122993929 on Build GRCh38
rs748397909 Chr.10: 122993995-122993995 on Build GRCh38
rs768471670 Chr.10: 122994034-122994034 on Build GRCh38
rs749973001 Chr.10: 122993968-122993968 on Build GRCh38
rs771174517 Chr.10: 122994026-122994026 on Build GRCh38
rs761592322 Chr.10: 122993960-122993960 on Build GRCh38
rs548855309 Chr.10: 122993666-122993666 on Build GRCh38
rs1045620 Chr.10: 122993724-122993724 on Build GRCh38
rs780490982 Chr.10: 122993793-122993793 on Build GRCh38
rs756835502 Chr.10: 122993766-122993766 on Build GRCh38
rs528335452 Chr.10: 122993880-122993880 on Build GRCh38
rs373833325 Chr.10: 122993871-122993871 on Build GRCh38
rs749294146 Chr.10: 122994027-122994027 on Build GRCh38
rs1032456569 Chr.10: 122993723-122993723 on Build GRCh38
rs376998630 Chr.10: 122993892-122993892 on Build GRCh38
rs918602309 Chr.10: 122993707-122993707 on Build GRCh38
rs974094497 Chr.10: 122994029-122994029 on Build GRCh38
rs773081271 Chr.10: 122993952-122993954 on Build GRCh38
rs765601336 Chr.10: 122994069-122994069 on Build GRCh38
rs376306904 Chr.10: 122993972-122993972 on Build GRCh38
rs780811487 Chr.10: 122993772-122993772 on Build GRCh38
rs752550125 Chr.10: 122993733-122993733 on Build GRCh38
rs35178103 Chr.10: 122993928-122993928 on Build GRCh38
rs370403522 Chr.10: 122994051-122994051 on Build GRCh38
rs755483547 Chr.10: 122994056-122994056 on Build GRCh38
rs972642927 Chr.10: 122993706-122993706 on Build GRCh38
rs767021729 Chr.10: 122993962-122993962 on Build GRCh38
rs151015331 Chr.10: 122993973-122993973 on Build GRCh38
rs760302154 Chr.10: 122994053-122994053 on Build GRCh38
rs772183136 Chr.10: 122993891-122993891 on Build GRCh38
rs751265429 Chr.10: 122993757-122993757 on Build GRCh38
rs962950480 Chr.10: 122993684-122993684 on Build GRCh38
rs113118844 Chr.10: 122993661-122993664 on Build GRCh38
rs755469062 Chr.10: 122993980-122993980 on Build GRCh38
rs756628262 Chr.10: 122993784-122993784 on Build GRCh38
rs955352936 Chr.10: 122993724-122993725 on Build GRCh38
rs771842420 Chr.10: 122993742-122993742 on Build GRCh38
rs189186734 Chr.10: 122993756-122993756 on Build GRCh38
rs1016892345 Chr.10: 122993679-122993679 on Build GRCh38
rs769871385 Chr.10: 122993925-122993925 on Build GRCh38
rs919932411 Chr.10: 122994062-122994062 on Build GRCh38
rs536303734 Chr.10: 122994035-122994035 on Build GRCh38
rs79165702 Chr.10: 122993791-122993791 on Build GRCh38
rs75613220 Chr.10: 122994009-122994009 on Build GRCh38
rs762784377 Chr.10: 122993739-122993739 on Build GRCh38
rs755379194 Chr.10: 122993828-122993828 on Build GRCh38
rs749837077 Chr.10: 122993776-122993776 on Build GRCh38
rs764029447 Chr.10: 122993930-122993930 on Build GRCh38
rs774001644 Chr.10: 122993948-122993948 on Build GRCh38
rs765935858 Chr.10: 122994055-122994055 on Build GRCh38
rs767229956 Chr.10: 122994046-122994046 on Build GRCh38
rs747112822 Chr.10: 122994006-122994006 on Build GRCh38
rs755581220 Chr.10: 122993971-122993971 on Build GRCh38
rs777805669 Chr.10: 122993999-122993999 on Build GRCh38
rs1034743402 Chr.10: 122993641-122993641 on Build GRCh38
rs749685530 Chr.10: 122993796-122993796 on Build GRCh38
rs902786653 Chr.10: 122993635-122993635 on Build GRCh38
rs779404645 Chr.10: 122993992-122993992 on Build GRCh38
rs768859957 Chr.10: 122993976-122993977 on Build GRCh38
rs758657670 Chr.10: 122993997-122993997 on Build GRCh38
rs761468259 Chr.10: 122994037-122994037 on Build GRCh38

View Full Product Details