Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 125770233-125770730 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MMP21
Entrez Gene ID
118856
Gene Name
matrix metallopeptidase 21
Gene Aliases
HTX7, MMP-21
UniGene
Hs.314141
Gene Chromosome Location
Chr.10: 125766453-125775821 on Build GRCh38
Transcripts
NM_147191.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3685975 c.980-4delT p.? Chr.10: 125770595-125770595 on Build GRCh38
6245226 c.1205C>A p.T402K Chr.10: 125770366-125770366 on Build GRCh38
6713846 c.1181T>C p.I394T Chr.10: 125770390-125770390 on Build GRCh38
6713843 c.1233delT p.Q412fs*28 Chr.10: 125770338-125770338 on Build GRCh38
7288766 c.1047G>T p.E349D Chr.10: 125770524-125770524 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138944733 Chr.10: 125770448-125770448 on Build GRCh38
rs764834246 Chr.10: 125770453-125770453 on Build GRCh38
rs887949790 Chr.10: 125770292-125770292 on Build GRCh38
rs755053134 Chr.10: 125770581-125770581 on Build GRCh38
rs751441192 Chr.10: 125770488-125770488 on Build GRCh38
rs191317268 Chr.10: 125770264-125770264 on Build GRCh38
rs762230950 Chr.10: 125770431-125770431 on Build GRCh38
rs984121903 Chr.10: 125770513-125770513 on Build GRCh38
rs769792888 Chr.10: 125770286-125770286 on Build GRCh38
rs374636376 Chr.10: 125770283-125770283 on Build GRCh38
rs991866203 Chr.10: 125770468-125770468 on Build GRCh38
rs921865127 Chr.10: 125770700-125770700 on Build GRCh38
rs544983541 Chr.10: 125770608-125770608 on Build GRCh38
rs983202396 Chr.10: 125770645-125770645 on Build GRCh38
rs372562000 Chr.10: 125770348-125770348 on Build GRCh38
rs749441754 Chr.10: 125770390-125770390 on Build GRCh38
rs772412908 Chr.10: 125770518-125770518 on Build GRCh38
rs1030377022 Chr.10: 125770374-125770374 on Build GRCh38
rs774755082 Chr.10: 125770406-125770406 on Build GRCh38
rs139649048 Chr.10: 125770549-125770549 on Build GRCh38
rs762318902 Chr.10: 125770327-125770327 on Build GRCh38
rs17153528 Chr.10: 125770620-125770620 on Build GRCh38
rs991879382 Chr.10: 125770685-125770685 on Build GRCh38
rs149458277 Chr.10: 125770371-125770371 on Build GRCh38
rs767594023 Chr.10: 125770626-125770627 on Build GRCh38
rs17153531 Chr.10: 125770642-125770642 on Build GRCh38
rs767318046 Chr.10: 125770564-125770564 on Build GRCh38
rs764747628 Chr.10: 125770534-125770534 on Build GRCh38
rs757119224 Chr.10: 125770550-125770550 on Build GRCh38
rs1039546531 Chr.10: 125770367-125770367 on Build GRCh38
rs752375826 Chr.10: 125770463-125770463 on Build GRCh38
rs752696006 Chr.10: 125770640-125770640 on Build GRCh38
rs777159889 Chr.10: 125770527-125770527 on Build GRCh38
rs764481266 Chr.10: 125770314-125770314 on Build GRCh38
rs150320323 Chr.10: 125770349-125770349 on Build GRCh38
rs979315676 Chr.10: 125770434-125770434 on Build GRCh38
rs368286076 Chr.10: 125770293-125770293 on Build GRCh38
rs749348073 Chr.10: 125770497-125770497 on Build GRCh38
rs202135492 Chr.10: 125770636-125770636 on Build GRCh38
rs1051813562 Chr.10: 125770594-125770594 on Build GRCh38
rs774847046 Chr.10: 125770318-125770318 on Build GRCh38
rs542840001 Chr.10: 125770630-125770630 on Build GRCh38
rs778105877 Chr.10: 125770602-125770602 on Build GRCh38
rs779230567 Chr.10: 125770508-125770508 on Build GRCh38
rs781243308 Chr.10: 125770495-125770495 on Build GRCh38
rs17153524 Chr.10: 125770492-125770492 on Build GRCh38
rs373501274 Chr.10: 125770258-125770258 on Build GRCh38
rs779995692 Chr.10: 125770389-125770389 on Build GRCh38
rs758535896 Chr.10: 125770600-125770600 on Build GRCh38
rs199999352 Chr.10: 125770353-125770353 on Build GRCh38
rs959161510 Chr.10: 125770656-125770656 on Build GRCh38
rs756113664 Chr.10: 125770634-125770634 on Build GRCh38
rs761159121 Chr.10: 125770523-125770523 on Build GRCh38
rs929597864 Chr.10: 125770259-125770259 on Build GRCh38
rs772346896 Chr.10: 125770437-125770437 on Build GRCh38
rs761089839 Chr.10: 125770447-125770447 on Build GRCh38
rs758137866 Chr.10: 125770470-125770470 on Build GRCh38
rs758225850 Chr.10: 125770379-125770379 on Build GRCh38
rs751578472 Chr.10: 125770607-125770610 on Build GRCh38
rs768106807 Chr.10: 125770329-125770329 on Build GRCh38
rs771709677 Chr.10: 125770599-125770602 on Build GRCh38
rs577517876 Chr.10: 125770607-125770607 on Build GRCh38
rs748304289 Chr.10: 125770591-125770591 on Build GRCh38
rs774970525 Chr.10: 125770613-125770613 on Build GRCh38
rs1012490527 Chr.10: 125770606-125770606 on Build GRCh38
rs762577062 Chr.10: 125770619-125770619 on Build GRCh38
rs748396381 Chr.10: 125770509-125770509 on Build GRCh38
rs750409809 Chr.10: 125770567-125770567 on Build GRCh38
rs746938693 Chr.10: 125770385-125770385 on Build GRCh38
rs974661188 Chr.10: 125770698-125770698 on Build GRCh38
rs765822061 Chr.10: 125770329-125770330 on Build GRCh38
rs147129223 Chr.10: 125770442-125770442 on Build GRCh38
rs750926499 Chr.10: 125770343-125770343 on Build GRCh38
rs763088203 Chr.10: 125770595-125770595 on Build GRCh38
rs1047935654 Chr.10: 125770281-125770281 on Build GRCh38
rs775945817 Chr.10: 125770594-125770595 on Build GRCh38
rs933393411 Chr.10: 125770588-125770588 on Build GRCh38
rs137955225 Chr.10: 125770368-125770368 on Build GRCh38
rs142536039 Chr.10: 125770612-125770613 on Build GRCh38
rs563864855 Chr.10: 125770612-125770612 on Build GRCh38
rs767435496 Chr.10: 125770631-125770631 on Build GRCh38
rs199919494 Chr.10: 125770308-125770308 on Build GRCh38
rs762510689 Chr.10: 125770542-125770542 on Build GRCh38
rs28381302 Chr.10: 125770525-125770525 on Build GRCh38
rs768888911 Chr.10: 125770503-125770503 on Build GRCh38
rs753274145 Chr.10: 125770449-125770449 on Build GRCh38
rs754621641 Chr.10: 125770357-125770357 on Build GRCh38
rs968306482 Chr.10: 125770408-125770408 on Build GRCh38
rs775062496 Chr.10: 125770536-125770536 on Build GRCh38
rs375237734 Chr.10: 125770304-125770304 on Build GRCh38
rs773443397 Chr.10: 125770519-125770519 on Build GRCh38
rs759002305 Chr.10: 125770533-125770533 on Build GRCh38
rs1022627441 Chr.10: 125770263-125770263 on Build GRCh38
rs777724439 Chr.10: 125770381-125770381 on Build GRCh38
rs564984415 Chr.10: 125770397-125770397 on Build GRCh38
rs142863684 Chr.10: 125770452-125770452 on Build GRCh38
rs779136644 Chr.10: 125770590-125770590 on Build GRCh38
rs753462694 Chr.10: 125770354-125770354 on Build GRCh38
rs946722250 Chr.10: 125770493-125770493 on Build GRCh38
rs1018714573 Chr.10: 125770664-125770664 on Build GRCh38
rs538574499 Chr.10: 125770679-125770679 on Build GRCh38
rs759773945 Chr.10: 125770290-125770290 on Build GRCh38

View Full Product Details