Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 127982191-127982705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPRE
Entrez Gene ID
5791
Gene Name
protein tyrosine phosphatase, receptor type E
Gene Aliases
HPTPE, PTPE, R-PTP-EPSILON
UniGene
Hs.127022
Gene Chromosome Location
Chr.10: 127907061-128085855 on Build GRCh38
Transcripts
NM_001316676.1 NM_001323355.1 NM_001323356.1 NM_006504.5 XM_011539994.2 XM_011539995.1 XM_011539996.1 XM_011539998.1 XM_017016467.1 XM_017016468.1 XM_017016469.1

SNP Detail

SNP ID Chromosome Location
rs72323848 Chr.10: 127982490-127982491 on Build GRCh38
rs140772206 Chr.10: 127982570-127982570 on Build GRCh38
rs1033546717 Chr.10: 127982675-127982675 on Build GRCh38
rs1046940707 Chr.10: 127982486-127982486 on Build GRCh38
rs947784685 Chr.10: 127982374-127982374 on Build GRCh38
rs980846186 Chr.10: 127982395-127982395 on Build GRCh38
rs1024519300 Chr.10: 127982348-127982348 on Build GRCh38
rs187446189 Chr.10: 127982661-127982661 on Build GRCh38
rs897066570 Chr.10: 127982301-127982301 on Build GRCh38
rs558619785 Chr.10: 127982490-127982490 on Build GRCh38
rs759660355 Chr.10: 127982649-127982649 on Build GRCh38
rs572981786 Chr.10: 127982465-127982465 on Build GRCh38
rs886965440 Chr.10: 127982489-127982490 on Build GRCh38
rs760713352 Chr.10: 127982501-127982502 on Build GRCh38
rs150897918 Chr.10: 127982490-127982505 on Build GRCh38
rs145544146 Chr.10: 127982622-127982622 on Build GRCh38
rs922062921 Chr.10: 127982557-127982557 on Build GRCh38
rs1001143291 Chr.10: 127982595-127982595 on Build GRCh38
rs970181118 Chr.10: 127982403-127982403 on Build GRCh38
rs944922613 Chr.10: 127982535-127982535 on Build GRCh38
rs377509566 Chr.10: 127982673-127982673 on Build GRCh38
rs773519462 Chr.10: 127982233-127982233 on Build GRCh38
rs769564058 Chr.10: 127982504-127982515 on Build GRCh38
rs59170572 Chr.10: 127982529-127982530 on Build GRCh38
rs1031247214 Chr.10: 127982603-127982603 on Build GRCh38
rs774932915 Chr.10: 127982443-127982443 on Build GRCh38
rs980623572 Chr.10: 127982422-127982422 on Build GRCh38
rs577580866 Chr.10: 127982662-127982662 on Build GRCh38
rs897076682 Chr.10: 127982628-127982628 on Build GRCh38
rs1033080754 Chr.10: 127982451-127982451 on Build GRCh38
rs149719270 Chr.10: 127982580-127982580 on Build GRCh38
rs554315124 Chr.10: 127982382-127982382 on Build GRCh38
rs976292893 Chr.10: 127982546-127982546 on Build GRCh38
rs982692006 Chr.10: 127982636-127982636 on Build GRCh38
rs3886573 Chr.10: 127982426-127982426 on Build GRCh38
rs535707143 Chr.10: 127982606-127982606 on Build GRCh38
rs536365025 Chr.10: 127982346-127982346 on Build GRCh38
rs184257479 Chr.10: 127982529-127982529 on Build GRCh38
rs952393701 Chr.10: 127982632-127982632 on Build GRCh38
rs3891272 Chr.10: 127982511-127982511 on Build GRCh38
rs144391777 Chr.10: 127982490-127982501 on Build GRCh38
rs931236046 Chr.10: 127982470-127982470 on Build GRCh38
rs150961087 Chr.10: 127982287-127982287 on Build GRCh38
rs763356223 Chr.10: 127982246-127982246 on Build GRCh38
rs771149903 Chr.10: 127982328-127982328 on Build GRCh38
rs7921013 Chr.10: 127982625-127982625 on Build GRCh38
rs905716484 Chr.10: 127982663-127982663 on Build GRCh38
rs980517135 Chr.10: 127982626-127982626 on Build GRCh38
rs1014103693 Chr.10: 127982305-127982305 on Build GRCh38
rs71035510 Chr.10: 127982530-127982531 on Build GRCh38
rs908465118 Chr.10: 127982639-127982639 on Build GRCh38
rs996241750 Chr.10: 127982541-127982541 on Build GRCh38
rs574556503 Chr.10: 127982607-127982607 on Build GRCh38
rs541972490 Chr.10: 127982612-127982612 on Build GRCh38
rs774996556 Chr.10: 127982296-127982296 on Build GRCh38
rs1040476338 Chr.10: 127982521-127982521 on Build GRCh38
rs764560888 Chr.10: 127982295-127982295 on Build GRCh38
rs115040045 Chr.10: 127982469-127982469 on Build GRCh38
rs533069009 Chr.10: 127982249-127982249 on Build GRCh38

View Full Product Details