Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 497923-498430 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNH1
Entrez Gene ID
6050
Gene Name
ribonuclease/angiogenin inhibitor 1
Gene Aliases
RAI, RNH
UniGene
Hs.530687
Gene Chromosome Location
Chr.11: 494512-507283 on Build GRCh38
Transcripts
NM_002939.3 NM_203383.1 NM_203384.1 NM_203385.1 NM_203386.2 NM_203387.2 NM_203388.2 NM_203389.2 XM_011520255.1 XM_011520257.2 XM_011520258.2 XM_011520259.2 XM_011520260.2 XM_011520261.2 XM_011520262.2 XM_011520263.1 XM_017018106.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6055604 c.1109C>T p.S370F Chr.11: 497989-497989 on Build GRCh38
6768078 c.1085G>A p.C362Y Chr.11: 498013-498013 on Build GRCh38
3383531 c.981C>T p.A327A Chr.11: 498117-498117 on Build GRCh38
3687353 c.966C>A p.S322S Chr.11: 498132-498132 on Build GRCh38
5800132 c.1027C>T p.L343F Chr.11: 498071-498071 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1037250043 Chr.11: 498377-498377 on Build GRCh38
rs143037442 Chr.11: 497995-497995 on Build GRCh38
rs972082962 Chr.11: 498264-498264 on Build GRCh38
rs924788943 Chr.11: 498277-498277 on Build GRCh38
rs748514042 Chr.11: 498132-498132 on Build GRCh38
rs752556323 Chr.11: 498072-498072 on Build GRCh38
rs760118083 Chr.11: 497973-497973 on Build GRCh38
rs747554530 Chr.11: 497960-497961 on Build GRCh38
rs769373159 Chr.11: 498381-498381 on Build GRCh38
rs200489835 Chr.11: 497994-497994 on Build GRCh38
rs369604821 Chr.11: 498159-498159 on Build GRCh38
rs991759511 Chr.11: 497965-497965 on Build GRCh38
rs11825121 Chr.11: 498383-498383 on Build GRCh38
rs564863081 Chr.11: 498169-498169 on Build GRCh38
rs752539935 Chr.11: 498000-498000 on Build GRCh38
rs190556300 Chr.11: 498321-498321 on Build GRCh38
rs764029487 Chr.11: 498173-498173 on Build GRCh38
rs774256528 Chr.11: 498176-498176 on Build GRCh38
rs934692134 Chr.11: 498320-498320 on Build GRCh38
rs746709036 Chr.11: 498153-498153 on Build GRCh38
rs745312635 Chr.11: 498164-498164 on Build GRCh38
rs556843557 Chr.11: 497958-497958 on Build GRCh38
rs746696140 Chr.11: 498026-498026 on Build GRCh38
rs772764579 Chr.11: 497961-497961 on Build GRCh38
rs767884083 Chr.11: 498084-498084 on Build GRCh38
rs759267447 Chr.11: 497981-497981 on Build GRCh38
rs780449071 Chr.11: 498018-498018 on Build GRCh38
rs549572853 Chr.11: 498348-498348 on Build GRCh38
rs750626202 Chr.11: 498116-498116 on Build GRCh38
rs748497432 Chr.11: 498027-498027 on Build GRCh38
rs1016516757 Chr.11: 498218-498218 on Build GRCh38
rs372334175 Chr.11: 498165-498165 on Build GRCh38
rs899783880 Chr.11: 498146-498146 on Build GRCh38
rs566178389 Chr.11: 498378-498378 on Build GRCh38
rs900436782 Chr.11: 498071-498071 on Build GRCh38
rs777422128 Chr.11: 498151-498151 on Build GRCh38
rs76149492 Chr.11: 498402-498402 on Build GRCh38
rs781354565 Chr.11: 498148-498149 on Build GRCh38
rs755565668 Chr.11: 498125-498125 on Build GRCh38
rs1023321090 Chr.11: 498220-498220 on Build GRCh38
rs771723035 Chr.11: 498029-498029 on Build GRCh38
rs1039469169 Chr.11: 498373-498373 on Build GRCh38
rs760206062 Chr.11: 498036-498036 on Build GRCh38
rs779333627 Chr.11: 498126-498126 on Build GRCh38
rs374635782 Chr.11: 498023-498023 on Build GRCh38
rs375372486 Chr.11: 498102-498102 on Build GRCh38
rs774556629 Chr.11: 497952-497952 on Build GRCh38
rs201867248 Chr.11: 497972-497972 on Build GRCh38
rs577836583 Chr.11: 498022-498022 on Build GRCh38
rs572871991 Chr.11: 498052-498052 on Build GRCh38
rs148207047 Chr.11: 498117-498117 on Build GRCh38
rs886829805 Chr.11: 498183-498183 on Build GRCh38
rs756223796 Chr.11: 498006-498006 on Build GRCh38
rs563978526 Chr.11: 498327-498327 on Build GRCh38
rs914691816 Chr.11: 498357-498357 on Build GRCh38
rs948888750 Chr.11: 497984-497984 on Build GRCh38
rs769016598 Chr.11: 497951-497951 on Build GRCh38
rs771283899 Chr.11: 498044-498046 on Build GRCh38
rs1003935588 Chr.11: 498193-498193 on Build GRCh38
rs981622201 Chr.11: 498276-498276 on Build GRCh38
rs943379230 Chr.11: 498359-498359 on Build GRCh38
rs765075092 Chr.11: 498069-498069 on Build GRCh38
rs772383277 Chr.11: 498138-498138 on Build GRCh38
rs770422036 Chr.11: 498042-498042 on Build GRCh38
rs750536501 Chr.11: 498003-498003 on Build GRCh38
rs766229039 Chr.11: 497976-497976 on Build GRCh38
rs370831864 Chr.11: 498347-498347 on Build GRCh38
rs761024081 Chr.11: 498178-498178 on Build GRCh38
rs762772127 Chr.11: 498075-498075 on Build GRCh38
rs201659542 Chr.11: 498002-498002 on Build GRCh38
rs372343001 Chr.11: 498055-498055 on Build GRCh38
rs1023217656 Chr.11: 498228-498228 on Build GRCh38
rs184699651 Chr.11: 498250-498250 on Build GRCh38
rs997468426 Chr.11: 498096-498096 on Build GRCh38
rs761469131 Chr.11: 498225-498225 on Build GRCh38
rs941987430 Chr.11: 498379-498379 on Build GRCh38
rs893143827 Chr.11: 498093-498093 on Build GRCh38
rs990326070 Chr.11: 497978-497978 on Build GRCh38
rs536946904 Chr.11: 497956-497956 on Build GRCh38
rs766495476 Chr.11: 498407-498407 on Build GRCh38
rs757613430 Chr.11: 498001-498001 on Build GRCh38
rs201027907 Chr.11: 497980-497980 on Build GRCh38
rs772782609 Chr.11: 498030-498030 on Build GRCh38
rs776419087 Chr.11: 498224-498224 on Build GRCh38

View Full Product Details