Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 790769-791258 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A22
Entrez Gene ID
79751
Gene Name
solute carrier family 25 member 22
Gene Aliases
EIEE3, GC-1, GC1, NET44
UniGene
Hs.99486
Gene Chromosome Location
Chr.11: 790475-798269 on Build GRCh38
Transcripts
NM_001191060.1 NM_001191061.1 NM_024698.5 XM_011520369.1 XM_011520370.1 XM_011520371.1

SNP Detail

SNP ID Chromosome Location
rs1030869137 Chr.11: 791143-791143 on Build GRCh38
rs923851069 Chr.11: 790914-790914 on Build GRCh38
rs1006279251 Chr.11: 790839-790839 on Build GRCh38
rs768779780 Chr.11: 790831-790831 on Build GRCh38
rs947342030 Chr.11: 791174-791174 on Build GRCh38
rs886048691 Chr.11: 791070-791070 on Build GRCh38
rs1024773402 Chr.11: 790869-790869 on Build GRCh38
rs1019728232 Chr.11: 791209-791209 on Build GRCh38
rs1024722451 Chr.11: 791121-791121 on Build GRCh38
rs938579581 Chr.11: 790918-790918 on Build GRCh38
rs570270747 Chr.11: 791221-791221 on Build GRCh38
rs952981529 Chr.11: 791223-791223 on Build GRCh38
rs549458341 Chr.11: 791233-791233 on Build GRCh38
rs894813059 Chr.11: 791070-791070 on Build GRCh38
rs577974181 Chr.11: 790967-790967 on Build GRCh38
rs999341429 Chr.11: 791138-791138 on Build GRCh38
rs908970706 Chr.11: 790862-790862 on Build GRCh38
rs978000475 Chr.11: 790817-790817 on Build GRCh38
rs544567754 Chr.11: 791162-791162 on Build GRCh38
rs928445753 Chr.11: 791146-791146 on Build GRCh38
rs1021756897 Chr.11: 790795-790796 on Build GRCh38
rs375467519 Chr.11: 791222-791222 on Build GRCh38
rs977953428 Chr.11: 790908-790908 on Build GRCh38
rs558106987 Chr.11: 790937-790937 on Build GRCh38
rs967500110 Chr.11: 790812-790812 on Build GRCh38
rs118051043 Chr.11: 791009-791009 on Build GRCh38
rs139397585 Chr.11: 791155-791155 on Build GRCh38
rs111723529 Chr.11: 791204-791204 on Build GRCh38
rs907095642 Chr.11: 790910-790910 on Build GRCh38
rs906320767 Chr.11: 791135-791135 on Build GRCh38
rs1020180656 Chr.11: 791022-791022 on Build GRCh38
rs886048692 Chr.11: 791105-791105 on Build GRCh38
rs570129087 Chr.11: 790799-790799 on Build GRCh38
rs940496738 Chr.11: 790874-790874 on Build GRCh38
rs1036618990 Chr.11: 790878-790878 on Build GRCh38
rs573754741 Chr.11: 791019-791019 on Build GRCh38
rs534970957 Chr.11: 790933-790933 on Build GRCh38
rs908937740 Chr.11: 791117-791117 on Build GRCh38
rs543726213 Chr.11: 790975-790975 on Build GRCh38
rs970556206 Chr.11: 790896-790896 on Build GRCh38
rs139577104 Chr.11: 791209-791210 on Build GRCh38
rs942308652 Chr.11: 791047-791047 on Build GRCh38
rs1047977298 Chr.11: 790800-790800 on Build GRCh38
rs1021683339 Chr.11: 791014-791014 on Build GRCh38
rs943425806 Chr.11: 791010-791010 on Build GRCh38
rs766005705 Chr.11: 791144-791144 on Build GRCh38
rs923409569 Chr.11: 791020-791020 on Build GRCh38
rs901337284 Chr.11: 791210-791221 on Build GRCh38
rs953012195 Chr.11: 791102-791102 on Build GRCh38
rs550204204 Chr.11: 791205-791205 on Build GRCh38
rs763350462 Chr.11: 791112-791112 on Build GRCh38
rs1037748447 Chr.11: 791063-791063 on Build GRCh38
rs965600937 Chr.11: 791029-791029 on Build GRCh38
rs886728603 Chr.11: 790811-790811 on Build GRCh38
rs886675681 Chr.11: 791040-791040 on Build GRCh38
rs17156066 Chr.11: 791002-791002 on Build GRCh38
rs375049082 Chr.11: 790858-790858 on Build GRCh38
rs886048690 Chr.11: 790874-790874 on Build GRCh38
rs542750433 Chr.11: 791036-791036 on Build GRCh38
rs762429573 Chr.11: 790850-790850 on Build GRCh38
rs976061263 Chr.11: 791045-791045 on Build GRCh38
rs34712290 Chr.11: 791009-791010 on Build GRCh38
rs114476401 Chr.11: 791203-791203 on Build GRCh38
rs886048693 Chr.11: 791135-791135 on Build GRCh38
rs572412088 Chr.11: 790893-790893 on Build GRCh38
rs966031353 Chr.11: 790834-790834 on Build GRCh38
rs894476789 Chr.11: 791004-791004 on Build GRCh38
rs542204237 Chr.11: 790902-790902 on Build GRCh38
rs576938612 Chr.11: 791169-791169 on Build GRCh38
rs959352369 Chr.11: 790856-790856 on Build GRCh38
rs933484658 Chr.11: 791034-791034 on Build GRCh38
rs974751122 Chr.11: 791181-791181 on Build GRCh38
rs971863478 Chr.11: 791137-791137 on Build GRCh38
rs752155405 Chr.11: 791230-791230 on Build GRCh38
rs1037964153 Chr.11: 791235-791235 on Build GRCh38
rs747118458 Chr.11: 790796-790796 on Build GRCh38
rs116673603 Chr.11: 791074-791074 on Build GRCh38
rs535992217 Chr.11: 790849-790849 on Build GRCh38

View Full Product Details