Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 1557714-1558221 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP8
Entrez Gene ID
1850
Gene Name
dual specificity phosphatase 8
Gene Aliases
C11orf81, HB5, HVH-5, HVH8
UniGene
Hs.41688
Gene Chromosome Location
Chr.11: 1554051-1572839 on Build GRCh38
Transcripts
NM_004420.2 XM_011519932.2 XM_011519933.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7119622 c.750C>G p.I250M Chr.11: 1557865-1557865 on Build GRCh38
3445677 c.729C>T p.I243I Chr.11: 1557886-1557886 on Build GRCh38
5958546 c.697G>T p.D233Y Chr.11: 1558112-1558112 on Build GRCh38
6426875 c.723A>C p.Q241H Chr.11: 1557892-1557892 on Build GRCh38
5913174 c.742G>A p.A248T Chr.11: 1557873-1557873 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs151020782 Chr.11: 1557996-1557996 on Build GRCh38
rs375664599 Chr.11: 1557937-1557937 on Build GRCh38
rs765537977 Chr.11: 1557885-1557885 on Build GRCh38
rs781269302 Chr.11: 1557808-1557808 on Build GRCh38
rs113161544 Chr.11: 1557788-1557788 on Build GRCh38
rs760317003 Chr.11: 1557878-1557878 on Build GRCh38
rs764560257 Chr.11: 1557927-1557927 on Build GRCh38
rs764933813 Chr.11: 1557847-1557852 on Build GRCh38
rs752362706 Chr.11: 1558166-1558167 on Build GRCh38
rs369977606 Chr.11: 1557963-1557963 on Build GRCh38
rs368533536 Chr.11: 1558101-1558101 on Build GRCh38
rs752077678 Chr.11: 1558167-1558167 on Build GRCh38
rs764378533 Chr.11: 1558162-1558162 on Build GRCh38
rs753062218 Chr.11: 1557886-1557886 on Build GRCh38
rs758940909 Chr.11: 1557889-1557889 on Build GRCh38
rs763305827 Chr.11: 1558157-1558157 on Build GRCh38
rs1026443743 Chr.11: 1557978-1557978 on Build GRCh38
rs779754427 Chr.11: 1558088-1558088 on Build GRCh38
rs767524832 Chr.11: 1558194-1558194 on Build GRCh38
rs756320305 Chr.11: 1558068-1558068 on Build GRCh38
rs373838996 Chr.11: 1557776-1557776 on Build GRCh38
rs777768655 Chr.11: 1557743-1557743 on Build GRCh38
rs748030374 Chr.11: 1558095-1558095 on Build GRCh38
rs760272806 Chr.11: 1558140-1558140 on Build GRCh38
rs201635880 Chr.11: 1557949-1557949 on Build GRCh38
rs200423798 Chr.11: 1557934-1557934 on Build GRCh38
rs768686376 Chr.11: 1557826-1557826 on Build GRCh38
rs370107291 Chr.11: 1557814-1557814 on Build GRCh38
rs750486625 Chr.11: 1558200-1558200 on Build GRCh38
rs761392997 Chr.11: 1557841-1557841 on Build GRCh38
rs917297083 Chr.11: 1557739-1557739 on Build GRCh38
rs773959979 Chr.11: 1557772-1557772 on Build GRCh38
rs533179997 Chr.11: 1557753-1557753 on Build GRCh38
rs144327546 Chr.11: 1558189-1558189 on Build GRCh38
rs749070851 Chr.11: 1558089-1558089 on Build GRCh38
rs775298923 Chr.11: 1557764-1557764 on Build GRCh38
rs779190227 Chr.11: 1557805-1557805 on Build GRCh38
rs753157729 Chr.11: 1557783-1557783 on Build GRCh38
rs898887776 Chr.11: 1557929-1557929 on Build GRCh38
rs774055480 Chr.11: 1557834-1557834 on Build GRCh38
rs778987415 Chr.11: 1558094-1558094 on Build GRCh38
rs771215213 Chr.11: 1558122-1558122 on Build GRCh38
rs1014551832 Chr.11: 1558016-1558016 on Build GRCh38
rs763720271 Chr.11: 1557770-1557770 on Build GRCh38
rs886603815 Chr.11: 1557981-1557981 on Build GRCh38
rs781278349 Chr.11: 1557962-1557962 on Build GRCh38
rs375567051 Chr.11: 1558091-1558091 on Build GRCh38
rs546651148 Chr.11: 1557819-1557819 on Build GRCh38
rs574421182 Chr.11: 1557740-1557740 on Build GRCh38
rs995422779 Chr.11: 1557960-1557960 on Build GRCh38
rs772729080 Chr.11: 1558113-1558113 on Build GRCh38
rs746039638 Chr.11: 1557809-1557809 on Build GRCh38
rs747250242 Chr.11: 1557752-1557752 on Build GRCh38
rs776448506 Chr.11: 1557754-1557754 on Build GRCh38
rs563099635 Chr.11: 1557757-1557757 on Build GRCh38
rs749215329 Chr.11: 1557821-1557821 on Build GRCh38
rs770583324 Chr.11: 1558146-1558146 on Build GRCh38
rs575190569 Chr.11: 1557763-1557763 on Build GRCh38
rs144147970 Chr.11: 1557853-1557853 on Build GRCh38
rs777858702 Chr.11: 1557799-1557799 on Build GRCh38
rs771720849 Chr.11: 1558180-1558180 on Build GRCh38
rs767343651 Chr.11: 1557847-1557847 on Build GRCh38
rs200008161 Chr.11: 1557773-1557773 on Build GRCh38
rs775764872 Chr.11: 1558152-1558152 on Build GRCh38
rs77411787 Chr.11: 1557802-1557802 on Build GRCh38
rs754286469 Chr.11: 1557774-1557774 on Build GRCh38

View Full Product Details