Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 1880003-1880496 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LSP1
Entrez Gene ID
4046
Gene Name
lymphocyte-specific protein 1
Gene Aliases
WP34, pp52
UniGene
Hs.56729
Gene Chromosome Location
Chr.11: 1852970-1892263 on Build GRCh38
Transcripts
NM_001013253.1 NM_001013254.1 NM_001013255.1 NM_001242932.1 NM_001289005.1 NM_002339.2 NR_107001.1

Gene Symbol
MIR7847
Entrez Gene ID
102465993
Gene Name
microRNA 7847
Gene Aliases
hsa-mir-7847
UniGene
Gene Chromosome Location
Chr.11: 1880045-1880147 on Build GRCh38
Transcripts
NM_001013253.1 NM_001013254.1 NM_001013255.1 NM_001242932.1 NM_001289005.1 NM_002339.2 NR_107001.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6704727 c.112C>T p.H38Y Chr.11: 1880145-1880145 on Build GRCh38
5480849 c.84G>A p.E28E Chr.11: 1880117-1880117 on Build GRCh38
5564820 c.172C>T p.R58W Chr.11: 1880205-1880205 on Build GRCh38
4793307 c.56C>T p.P19L Chr.11: 1880089-1880089 on Build GRCh38
6704728 c.150G>T p.E50D Chr.11: 1880183-1880183 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202243311 Chr.11: 1880046-1880046 on Build GRCh38
rs368052660 Chr.11: 1880105-1880105 on Build GRCh38
rs200764037 Chr.11: 1880114-1880114 on Build GRCh38
rs547274032 Chr.11: 1880304-1880304 on Build GRCh38
rs141540340 Chr.11: 1880180-1880180 on Build GRCh38
rs774187451 Chr.11: 1880190-1880190 on Build GRCh38
rs150172209 Chr.11: 1880132-1880132 on Build GRCh38
rs771869893 Chr.11: 1880195-1880195 on Build GRCh38
rs918237824 Chr.11: 1880417-1880417 on Build GRCh38
rs764553883 Chr.11: 1880148-1880148 on Build GRCh38
rs750861469 Chr.11: 1880234-1880234 on Build GRCh38
rs753356088 Chr.11: 1880137-1880137 on Build GRCh38
rs569794366 Chr.11: 1880466-1880466 on Build GRCh38
rs765597316 Chr.11: 1880060-1880060 on Build GRCh38
rs758943635 Chr.11: 1880148-1880151 on Build GRCh38
rs778193946 Chr.11: 1880091-1880091 on Build GRCh38
rs767118436 Chr.11: 1880129-1880129 on Build GRCh38
rs558867326 Chr.11: 1880191-1880191 on Build GRCh38
rs985208629 Chr.11: 1880356-1880356 on Build GRCh38
rs141767980 Chr.11: 1880126-1880126 on Build GRCh38
rs772062445 Chr.11: 1880244-1880244 on Build GRCh38
rs866657307 Chr.11: 1880471-1880471 on Build GRCh38
rs769199013 Chr.11: 1880262-1880262 on Build GRCh38
rs758887041 Chr.11: 1880068-1880068 on Build GRCh38
rs564153867 Chr.11: 1880289-1880289 on Build GRCh38
rs1049445128 Chr.11: 1880025-1880025 on Build GRCh38
rs144778074 Chr.11: 1880224-1880224 on Build GRCh38
rs753564099 Chr.11: 1880063-1880064 on Build GRCh38
rs922438967 Chr.11: 1880347-1880347 on Build GRCh38
rs866926158 Chr.11: 1880181-1880181 on Build GRCh38
rs751920485 Chr.11: 1880162-1880162 on Build GRCh38
rs1057054922 Chr.11: 1880246-1880246 on Build GRCh38
rs79902109 Chr.11: 1880250-1880250 on Build GRCh38
rs749887310 Chr.11: 1880051-1880051 on Build GRCh38
rs375928183 Chr.11: 1880198-1880198 on Build GRCh38
rs57352451 Chr.11: 1880205-1880205 on Build GRCh38
rs756341065 Chr.11: 1880235-1880235 on Build GRCh38
rs113444626 Chr.11: 1880085-1880085 on Build GRCh38
rs773560624 Chr.11: 1880329-1880329 on Build GRCh38
rs577178834 Chr.11: 1880202-1880202 on Build GRCh38
rs547972876 Chr.11: 1880364-1880364 on Build GRCh38
rs757725608 Chr.11: 1880175-1880175 on Build GRCh38
rs764747598 Chr.11: 1880237-1880237 on Build GRCh38
rs762382226 Chr.11: 1880267-1880267 on Build GRCh38
rs138090185 Chr.11: 1880363-1880363 on Build GRCh38
rs374180477 Chr.11: 1880059-1880059 on Build GRCh38
rs778767008 Chr.11: 1880243-1880243 on Build GRCh38
rs755019696 Chr.11: 1880242-1880242 on Build GRCh38
rs146975581 Chr.11: 1880268-1880268 on Build GRCh38
rs112538101 Chr.11: 1880462-1880462 on Build GRCh38
rs953866699 Chr.11: 1880354-1880354 on Build GRCh38
rs1036723198 Chr.11: 1880405-1880405 on Build GRCh38
rs535919851 Chr.11: 1880133-1880133 on Build GRCh38
rs2271439 Chr.11: 1880237-1880237 on Build GRCh38
rs574587041 Chr.11: 1880222-1880222 on Build GRCh38
rs747369818 Chr.11: 1880095-1880095 on Build GRCh38
rs570838125 Chr.11: 1880124-1880124 on Build GRCh38
rs181774507 Chr.11: 1880128-1880128 on Build GRCh38
rs924571497 Chr.11: 1880138-1880138 on Build GRCh38
rs542314446 Chr.11: 1880067-1880067 on Build GRCh38
rs757527171 Chr.11: 1880104-1880104 on Build GRCh38
rs770564830 Chr.11: 1880255-1880255 on Build GRCh38
rs775493053 Chr.11: 1880117-1880117 on Build GRCh38
rs1022297816 Chr.11: 1880300-1880300 on Build GRCh38
rs941077315 Chr.11: 1880380-1880380 on Build GRCh38
rs199983134 Chr.11: 1880266-1880266 on Build GRCh38
rs370864872 Chr.11: 1880050-1880050 on Build GRCh38
rs149491406 Chr.11: 1880206-1880206 on Build GRCh38
rs143537697 Chr.11: 1880093-1880093 on Build GRCh38
rs200822258 Chr.11: 1880241-1880241 on Build GRCh38
rs765897697 Chr.11: 1880135-1880135 on Build GRCh38
rs1030946437 Chr.11: 1880328-1880328 on Build GRCh38
rs1012294891 Chr.11: 1880293-1880293 on Build GRCh38
rs751382413 Chr.11: 1880049-1880049 on Build GRCh38
rs999516745 Chr.11: 1880314-1880315 on Build GRCh38
rs765942586 Chr.11: 1880059-1880059 on Build GRCh38
rs976672514 Chr.11: 1880344-1880344 on Build GRCh38
rs776175755 Chr.11: 1880260-1880260 on Build GRCh38
rs939613451 Chr.11: 1880461-1880461 on Build GRCh38
rs373858328 Chr.11: 1880140-1880140 on Build GRCh38
rs372884030 Chr.11: 1880189-1880189 on Build GRCh38
rs148042410 Chr.11: 1880106-1880106 on Build GRCh38
rs777512918 Chr.11: 1880249-1880249 on Build GRCh38
rs138504655 Chr.11: 1880178-1880178 on Build GRCh38
rs367677456 Chr.11: 1880177-1880177 on Build GRCh38
rs201512623 Chr.11: 1880232-1880232 on Build GRCh38
rs956172992 Chr.11: 1880340-1880340 on Build GRCh38
rs746854369 Chr.11: 1880253-1880253 on Build GRCh38
rs562100125 Chr.11: 1880313-1880313 on Build GRCh38
rs369993990 Chr.11: 1880200-1880200 on Build GRCh38
rs550150180 Chr.11: 1880292-1880292 on Build GRCh38
rs372030914 Chr.11: 1880209-1880209 on Build GRCh38
rs146194987 Chr.11: 1880201-1880201 on Build GRCh38
rs774174728 Chr.11: 1880127-1880127 on Build GRCh38

View Full Product Details