Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 2148264-2148773 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGF2
Entrez Gene ID
3481
Gene Name
insulin like growth factor 2
Gene Aliases
C11orf43, GRDF, IGF-II, PP9974
UniGene
Hs.272259
Gene Chromosome Location
Chr.11: 2129112-2149603 on Build GRCh38
Transcripts
NM_001007139.5 NR_028043.2 NR_133657.1 NM_001042376.2 NR_003512.3

Gene Symbol
IGF2-AS
Entrez Gene ID
51214
Gene Name
IGF2 antisense RNA
Gene Aliases
IGF2-AS1, IGF2AS, PEG8
UniGene
Hs.716962
Gene Chromosome Location
Chr.11: 2140512-2148666 on Build GRCh38
Transcripts
NM_001007139.5 NR_028043.2 NR_133657.1 NM_001042376.2 NR_003512.3

Gene Symbol
INS-IGF2
Entrez Gene ID
723961
Gene Name
INS-IGF2 readthrough
Gene Aliases
INSIGF
UniGene
Hs.272259
Gene Chromosome Location
Chr.11: 2129112-2161209 on Build GRCh38
Transcripts
NM_001007139.5 NR_028043.2 NR_133657.1 NM_001042376.2 NR_003512.3

SNP Detail

SNP ID Chromosome Location
rs543514383 Chr.11: 2148318-2148318 on Build GRCh38
rs917174515 Chr.11: 2148630-2148630 on Build GRCh38
rs185462585 Chr.11: 2148509-2148509 on Build GRCh38
rs1051934705 Chr.11: 2148562-2148562 on Build GRCh38
rs3741210 Chr.11: 2148310-2148310 on Build GRCh38
rs745984519 Chr.11: 2148745-2148745 on Build GRCh38
rs960116114 Chr.11: 2148643-2148643 on Build GRCh38
rs1045515963 Chr.11: 2148490-2148490 on Build GRCh38
rs3741206 Chr.11: 2148634-2148634 on Build GRCh38
rs551255045 Chr.11: 2148671-2148671 on Build GRCh38
rs919168516 Chr.11: 2148414-2148414 on Build GRCh38
rs17883152 Chr.11: 2148601-2148601 on Build GRCh38
rs949428769 Chr.11: 2148488-2148488 on Build GRCh38
rs993127174 Chr.11: 2148607-2148607 on Build GRCh38
rs940333019 Chr.11: 2148591-2148591 on Build GRCh38
rs1027180420 Chr.11: 2148633-2148633 on Build GRCh38
rs142242396 Chr.11: 2148379-2148379 on Build GRCh38
rs1009755706 Chr.11: 2148678-2148678 on Build GRCh38
rs181907014 Chr.11: 2148285-2148285 on Build GRCh38
rs954471665 Chr.11: 2148337-2148337 on Build GRCh38
rs897427598 Chr.11: 2148610-2148610 on Build GRCh38
rs911639977 Chr.11: 2148732-2148732 on Build GRCh38
rs980144736 Chr.11: 2148289-2148289 on Build GRCh38
rs987274766 Chr.11: 2148692-2148692 on Build GRCh38
rs878927927 Chr.11: 2148697-2148697 on Build GRCh38
rs1051453529 Chr.11: 2148424-2148424 on Build GRCh38
rs1036174346 Chr.11: 2148636-2148636 on Build GRCh38
rs910321326 Chr.11: 2148364-2148364 on Build GRCh38
rs1052945660 Chr.11: 2148330-2148330 on Build GRCh38
rs1027130255 Chr.11: 2148479-2148479 on Build GRCh38
rs4929963 Chr.11: 2148430-2148430 on Build GRCh38
rs3741207 Chr.11: 2148554-2148554 on Build GRCh38
rs17883285 Chr.11: 2148551-2148551 on Build GRCh38
rs940396253 Chr.11: 2148747-2148747 on Build GRCh38
rs116779517 Chr.11: 2148288-2148288 on Build GRCh38
rs1035717804 Chr.11: 2148524-2148524 on Build GRCh38
rs988722287 Chr.11: 2148534-2148534 on Build GRCh38
rs931806921 Chr.11: 2148682-2148682 on Build GRCh38
rs973210569 Chr.11: 2148587-2148587 on Build GRCh38
rs897363560 Chr.11: 2148447-2148447 on Build GRCh38
rs879920874 Chr.11: 2148543-2148544 on Build GRCh38
rs931701965 Chr.11: 2148412-2148412 on Build GRCh38
rs3741209 Chr.11: 2148525-2148525 on Build GRCh38
rs920015568 Chr.11: 2148286-2148286 on Build GRCh38
rs980581144 Chr.11: 2148513-2148513 on Build GRCh38
rs1015853063 Chr.11: 2148432-2148432 on Build GRCh38
rs921814710 Chr.11: 2148681-2148681 on Build GRCh38
rs3741205 Chr.11: 2148654-2148654 on Build GRCh38
rs1017309553 Chr.11: 2148716-2148716 on Build GRCh38
rs929267157 Chr.11: 2148419-2148419 on Build GRCh38
rs751102610 Chr.11: 2148635-2148635 on Build GRCh38
rs146404982 Chr.11: 2148420-2148420 on Build GRCh38
rs17883577 Chr.11: 2148543-2148543 on Build GRCh38
rs548456548 Chr.11: 2148522-2148522 on Build GRCh38
rs1026035746 Chr.11: 2148735-2148735 on Build GRCh38
rs4930042 Chr.11: 2148586-2148586 on Build GRCh38
rs772317002 Chr.11: 2148442-2148442 on Build GRCh38
rs552992329 Chr.11: 2148557-2148557 on Build GRCh38
rs905282665 Chr.11: 2148491-2148491 on Build GRCh38
rs3741204 Chr.11: 2148678-2148678 on Build GRCh38
rs748437577 Chr.11: 2148602-2148602 on Build GRCh38
rs182764161 Chr.11: 2148672-2148672 on Build GRCh38
rs530062133 Chr.11: 2148431-2148431 on Build GRCh38
rs4930041 Chr.11: 2148347-2148347 on Build GRCh38
rs372130188 Chr.11: 2148733-2148733 on Build GRCh38
rs17883084 Chr.11: 2148632-2148632 on Build GRCh38
rs768373789 Chr.11: 2148540-2148540 on Build GRCh38
rs973218973 Chr.11: 2148414-2148414 on Build GRCh38
rs3741208 Chr.11: 2148544-2148544 on Build GRCh38
rs995675006 Chr.11: 2148467-2148467 on Build GRCh38
rs557180918 Chr.11: 2148292-2148292 on Build GRCh38
rs559391350 Chr.11: 2148687-2148687 on Build GRCh38
rs754467283 Chr.11: 2148676-2148676 on Build GRCh38
rs892607946 Chr.11: 2148675-2148675 on Build GRCh38
rs573661046 Chr.11: 2148725-2148726 on Build GRCh38
rs558314833 Chr.11: 2148662-2148662 on Build GRCh38
rs17880764 Chr.11: 2148642-2148642 on Build GRCh38
rs1042838700 Chr.11: 2148358-2148358 on Build GRCh38
rs913104505 Chr.11: 2148334-2148334 on Build GRCh38
rs1001024405 Chr.11: 2148499-2148499 on Build GRCh38

View Full Product Details