Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 3797658-3798179 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUP98
Entrez Gene ID
4928
Gene Name
nucleoporin 98
Gene Aliases
ADIR2, NUP196, NUP96
UniGene
Hs.524750
Gene Chromosome Location
Chr.11: 3675010-3797792 on Build GRCh38
Transcripts
NM_005387.6 NM_016320.4 NM_139131.4 NM_139132.3 NM_001145438.2 NM_001256235.1 NM_001256236.1 NM_001256237.1 NM_001283038.1 NM_001283039.1 NM_001283040.1 NR_027017.2 NR_045923.1 NR_045925.1 NR_045926.1 NR_045927.1 NR_104270.1 NR_104271.1 XM_006718183.3 XM_006718185.2 XM_006718186.2 XM_006718190.3 XM_006718191.3 XM_006718192.1 XM_006718193.2 XM_011519992.1 XM_011519996.1 XM_011519997.1 XM_011519998.2 XM_011520000.1 XM_011520003.2 XM_011520004.2 XM_011520007.2 XM_017017557.1 XM_017017559.1 XM_017017560.1

Gene Symbol
PGAP2
Entrez Gene ID
27315
Gene Name
post-GPI attachment to proteins 2
Gene Aliases
CWH43-N, FRAG1, HPMRS3, MRT17, MRT21
UniGene
Hs.133968
Gene Chromosome Location
Chr.11: 3797724-3826371 on Build GRCh38
Transcripts
NM_005387.6 NM_016320.4 NM_139131.4 NM_139132.3 NM_001145438.2 NM_001256235.1 NM_001256236.1 NM_001256237.1 NM_001283038.1 NM_001283039.1 NM_001283040.1 NR_027017.2 NR_045923.1 NR_045925.1 NR_045926.1 NR_045927.1 NR_104270.1 NR_104271.1 XM_006718183.3 XM_006718185.2 XM_006718186.2 XM_006718190.3 XM_006718191.3 XM_006718192.1 XM_006718193.2 XM_011519992.1 XM_011519996.1 XM_011519997.1 XM_011519998.2 XM_011520000.1 XM_011520003.2 XM_011520004.2 XM_011520007.2 XM_017017557.1 XM_017017559.1 XM_017017560.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5957713 c.29T>G p.V10G Chr.11: 3797872-3797872 on Build GRCh38
3809193 c.106G>T p.G36W Chr.11: 3797949-3797949 on Build GRCh38
4544591 c.35G>A p.G12E Chr.11: 3797878-3797878 on Build GRCh38
5721634 c.38G>A p.R13K Chr.11: 3797881-3797881 on Build GRCh38
5497480 c.86G>A p.R29Q Chr.11: 3797929-3797929 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771803107 Chr.11: 3798058-3798058 on Build GRCh38
rs757468946 Chr.11: 3797964-3797964 on Build GRCh38
rs772780218 Chr.11: 3798014-3798014 on Build GRCh38
rs747979399 Chr.11: 3797998-3797998 on Build GRCh38
rs777601877 Chr.11: 3798154-3798154 on Build GRCh38
rs769586052 Chr.11: 3797732-3797732 on Build GRCh38
rs7116540 Chr.11: 3798037-3798037 on Build GRCh38
rs1019093202 Chr.11: 3797756-3797756 on Build GRCh38
rs1040416497 Chr.11: 3798079-3798079 on Build GRCh38
rs774100860 Chr.11: 3797864-3797864 on Build GRCh38
rs929436151 Chr.11: 3798117-3798117 on Build GRCh38
rs925134425 Chr.11: 3798049-3798049 on Build GRCh38
rs866855762 Chr.11: 3798064-3798064 on Build GRCh38
rs186502112 Chr.11: 3798030-3798030 on Build GRCh38
rs1033902298 Chr.11: 3798155-3798155 on Build GRCh38
rs771564057 Chr.11: 3798010-3798010 on Build GRCh38
rs781099258 Chr.11: 3797821-3797821 on Build GRCh38
rs764419154 Chr.11: 3797927-3797927 on Build GRCh38
rs747858326 Chr.11: 3798007-3798007 on Build GRCh38
rs896958082 Chr.11: 3797884-3797884 on Build GRCh38
rs1007675943 Chr.11: 3797754-3797754 on Build GRCh38
rs967696287 Chr.11: 3797840-3797840 on Build GRCh38
rs558998015 Chr.11: 3798061-3798061 on Build GRCh38
rs760098298 Chr.11: 3797915-3797915 on Build GRCh38
rs866937011 Chr.11: 3797912-3797912 on Build GRCh38
rs1040871921 Chr.11: 3797928-3797928 on Build GRCh38
rs903149310 Chr.11: 3797731-3797731 on Build GRCh38
rs1040632246 Chr.11: 3797721-3797721 on Build GRCh38
rs751660056 Chr.11: 3797818-3797818 on Build GRCh38
rs942091438 Chr.11: 3798071-3798071 on Build GRCh38
rs758619897 Chr.11: 3797802-3797802 on Build GRCh38
rs979192291 Chr.11: 3798016-3798016 on Build GRCh38
rs547916329 Chr.11: 3798104-3798104 on Build GRCh38
rs1006402317 Chr.11: 3798133-3798133 on Build GRCh38
rs925186703 Chr.11: 3797868-3797868 on Build GRCh38
rs61896926 Chr.11: 3797943-3797943 on Build GRCh38
rs1026986001 Chr.11: 3797790-3797790 on Build GRCh38
rs897839598 Chr.11: 3797934-3797934 on Build GRCh38
rs768458307 Chr.11: 3797863-3797863 on Build GRCh38
rs772580935 Chr.11: 3797886-3797886 on Build GRCh38
rs926320503 Chr.11: 3797804-3797804 on Build GRCh38
rs879347546 Chr.11: 3797710-3797710 on Build GRCh38
rs889194951 Chr.11: 3797744-3797744 on Build GRCh38
rs576607794 Chr.11: 3797940-3797940 on Build GRCh38
rs763253326 Chr.11: 3797923-3797923 on Build GRCh38
rs887966854 Chr.11: 3798128-3798128 on Build GRCh38
rs1006454438 Chr.11: 3798006-3798006 on Build GRCh38
rs917768598 Chr.11: 3797712-3797712 on Build GRCh38
rs901971824 Chr.11: 3797952-3797952 on Build GRCh38
rs180810726 Chr.11: 3797734-3797734 on Build GRCh38
rs1047868107 Chr.11: 3798126-3798126 on Build GRCh38
rs994071896 Chr.11: 3797976-3797976 on Build GRCh38
rs959099545 Chr.11: 3797806-3797806 on Build GRCh38
rs945212319 Chr.11: 3797862-3797862 on Build GRCh38
rs148297801 Chr.11: 3798073-3798073 on Build GRCh38
rs748970017 Chr.11: 3797853-3797853 on Build GRCh38
rs561479643 Chr.11: 3797919-3797919 on Build GRCh38
rs766644255 Chr.11: 3797807-3797807 on Build GRCh38
rs1000198150 Chr.11: 3797733-3797733 on Build GRCh38
rs778938135 Chr.11: 3797783-3797783 on Build GRCh38
rs543214203 Chr.11: 3797792-3797792 on Build GRCh38
rs990938951 Chr.11: 3797822-3797822 on Build GRCh38
rs1042711680 Chr.11: 3797730-3797730 on Build GRCh38
rs917659162 Chr.11: 3798035-3798035 on Build GRCh38
rs889144356 Chr.11: 3797911-3797911 on Build GRCh38
rs569729736 Chr.11: 3798113-3798113 on Build GRCh38
rs931835772 Chr.11: 3797896-3797896 on Build GRCh38
rs765592826 Chr.11: 3797917-3797917 on Build GRCh38
rs1006217155 Chr.11: 3797784-3797784 on Build GRCh38
rs537301662 Chr.11: 3798152-3798152 on Build GRCh38
rs543736143 Chr.11: 3797973-3797973 on Build GRCh38
rs756081998 Chr.11: 3797982-3797982 on Build GRCh38
rs777930856 Chr.11: 3797811-3797811 on Build GRCh38
rs755017795 Chr.11: 3797925-3797926 on Build GRCh38
rs1013127609 Chr.11: 3798002-3798002 on Build GRCh38
rs779913251 Chr.11: 3797837-3797837 on Build GRCh38
rs771751020 Chr.11: 3797878-3797878 on Build GRCh38
rs753897159 Chr.11: 3797825-3797825 on Build GRCh38
rs979342715 Chr.11: 3797690-3797690 on Build GRCh38
rs779819871 Chr.11: 3797987-3797987 on Build GRCh38
rs946369591 Chr.11: 3797681-3797681 on Build GRCh38
rs966628854 Chr.11: 3797759-3797759 on Build GRCh38
rs923886809 Chr.11: 3797705-3797705 on Build GRCh38
rs991891606 Chr.11: 3798034-3798034 on Build GRCh38
rs1050466264 Chr.11: 3797898-3797898 on Build GRCh38
rs761538212 Chr.11: 3797872-3797872 on Build GRCh38
rs1054444879 Chr.11: 3797907-3797907 on Build GRCh38
rs554269042 Chr.11: 3797724-3797724 on Build GRCh38
rs745886822 Chr.11: 3797828-3797828 on Build GRCh38
rs778821816 Chr.11: 3798001-3798001 on Build GRCh38
rs978319419 Chr.11: 3798039-3798039 on Build GRCh38
rs985952120 Chr.11: 3798056-3798056 on Build GRCh38
rs1023375231 Chr.11: 3798005-3798005 on Build GRCh38
rs943371673 Chr.11: 3798078-3798078 on Build GRCh38
rs953325164 Chr.11: 3798050-3798050 on Build GRCh38
rs945291430 Chr.11: 3797714-3797714 on Build GRCh38
rs762642892 Chr.11: 3797935-3797935 on Build GRCh38
rs749120967 Chr.11: 3797996-3797996 on Build GRCh38
rs932030773 Chr.11: 3797752-3797752 on Build GRCh38
rs191394026 Chr.11: 3798059-3798059 on Build GRCh38
rs769606483 Chr.11: 3797834-3797834 on Build GRCh38
rs935219037 Chr.11: 3797876-3797876 on Build GRCh38
rs937796536 Chr.11: 3797681-3797681 on Build GRCh38

View Full Product Details