Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 5323583-5324102 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR51B2
Entrez Gene ID
79345
Gene Name
olfactory receptor family 51 subfamily B member 2 (gene/pseudogene)
Gene Aliases
HOR5'Beta3, OR51B1P
UniGene
Hs.680163
Gene Chromosome Location
Chr.11: 5323298-5324352 on Build GRCh38
Transcripts
NM_033180.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4033532 c.454G>A p.V152I Chr.11: 5323844-5323844 on Build GRCh38
3448620 c.265G>A p.E89K Chr.11: 5324033-5324033 on Build GRCh38
1579770 c.466C>A p.P156T Chr.11: 5323832-5323832 on Build GRCh38
5098531 c.423G>A p.A141A Chr.11: 5323875-5323875 on Build GRCh38
3998450 c.400C>T p.L134F Chr.11: 5323898-5323898 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372875685 Chr.11: 5323961-5323961 on Build GRCh38
rs763944362 Chr.11: 5323836-5323836 on Build GRCh38
rs767837612 Chr.11: 5323607-5323607 on Build GRCh38
rs753522109 Chr.11: 5323683-5323684 on Build GRCh38
rs778916955 Chr.11: 5323788-5323788 on Build GRCh38
rs371761737 Chr.11: 5323622-5323622 on Build GRCh38
rs761315295 Chr.11: 5323618-5323618 on Build GRCh38
rs376970807 Chr.11: 5324069-5324069 on Build GRCh38
rs10837814 Chr.11: 5323898-5323898 on Build GRCh38
rs771610800 Chr.11: 5323888-5323888 on Build GRCh38
rs747270245 Chr.11: 5323862-5323862 on Build GRCh38
rs778070865 Chr.11: 5323794-5323794 on Build GRCh38
rs767181585 Chr.11: 5323831-5323831 on Build GRCh38
rs760520024 Chr.11: 5323643-5323644 on Build GRCh38
rs757354059 Chr.11: 5323787-5323787 on Build GRCh38
rs778432707 Chr.11: 5323873-5323873 on Build GRCh38
rs758517460 Chr.11: 5323908-5323908 on Build GRCh38
rs755533292 Chr.11: 5323825-5323825 on Build GRCh38
rs758948105 Chr.11: 5323885-5323885 on Build GRCh38
rs201051547 Chr.11: 5323879-5323879 on Build GRCh38
rs142847491 Chr.11: 5323698-5323698 on Build GRCh38
rs756074230 Chr.11: 5323676-5323676 on Build GRCh38
rs898308471 Chr.11: 5323840-5323840 on Build GRCh38
rs201379460 Chr.11: 5323738-5323738 on Build GRCh38
rs776436074 Chr.11: 5323967-5323967 on Build GRCh38
rs750420590 Chr.11: 5323907-5323907 on Build GRCh38
rs750189158 Chr.11: 5323626-5323627 on Build GRCh38
rs376302800 Chr.11: 5323820-5323820 on Build GRCh38
rs771557475 Chr.11: 5323954-5323954 on Build GRCh38
rs776021095 Chr.11: 5323710-5323710 on Build GRCh38
rs777666405 Chr.11: 5323780-5323780 on Build GRCh38
rs935770488 Chr.11: 5323757-5323757 on Build GRCh38
rs767614362 Chr.11: 5324004-5324004 on Build GRCh38
rs551907020 Chr.11: 5323695-5323695 on Build GRCh38
rs375582627 Chr.11: 5323686-5323686 on Build GRCh38
rs763001379 Chr.11: 5323973-5323973 on Build GRCh38
rs941819263 Chr.11: 5323838-5323838 on Build GRCh38
rs371532806 Chr.11: 5323945-5323945 on Build GRCh38
rs140971687 Chr.11: 5323777-5323777 on Build GRCh38
rs369276246 Chr.11: 5324073-5324073 on Build GRCh38
rs746117255 Chr.11: 5323708-5323708 on Build GRCh38
rs563433128 Chr.11: 5323981-5323981 on Build GRCh38
rs768326980 Chr.11: 5323966-5323966 on Build GRCh38
rs886444462 Chr.11: 5323852-5323852 on Build GRCh38
rs532742082 Chr.11: 5323674-5323674 on Build GRCh38
rs746596734 Chr.11: 5323963-5323963 on Build GRCh38
rs200974051 Chr.11: 5324028-5324028 on Build GRCh38
rs752158101 Chr.11: 5323909-5323910 on Build GRCh38
rs747396901 Chr.11: 5323809-5323809 on Build GRCh38
rs563063263 Chr.11: 5323606-5323606 on Build GRCh38
rs1030756953 Chr.11: 5324029-5324029 on Build GRCh38
rs375843480 Chr.11: 5324017-5324017 on Build GRCh38
rs34329019 Chr.11: 5323863-5323864 on Build GRCh38
rs189784581 Chr.11: 5324049-5324049 on Build GRCh38
rs766504415 Chr.11: 5323979-5323979 on Build GRCh38
rs75168207 Chr.11: 5323848-5323848 on Build GRCh38
rs369919059 Chr.11: 5323715-5323715 on Build GRCh38
rs751778194 Chr.11: 5323909-5323909 on Build GRCh38
rs145927879 Chr.11: 5323704-5323704 on Build GRCh38
rs781703701 Chr.11: 5323867-5323867 on Build GRCh38
rs775119687 Chr.11: 5323889-5323889 on Build GRCh38
rs762499259 Chr.11: 5323730-5323730 on Build GRCh38
rs980735117 Chr.11: 5323924-5323924 on Build GRCh38
rs367655935 Chr.11: 5323950-5323950 on Build GRCh38
rs765875773 Chr.11: 5323735-5323735 on Build GRCh38
rs780524233 Chr.11: 5323804-5323804 on Build GRCh38
rs761768145 Chr.11: 5323901-5323901 on Build GRCh38
rs752396679 Chr.11: 5323832-5323832 on Build GRCh38
rs754647046 Chr.11: 5323688-5323688 on Build GRCh38
rs773013986 Chr.11: 5323971-5323971 on Build GRCh38
rs267602944 Chr.11: 5323875-5323875 on Build GRCh38
rs749986741 Chr.11: 5323631-5323631 on Build GRCh38
rs964754176 Chr.11: 5323743-5323743 on Build GRCh38
rs200164732 Chr.11: 5324031-5324031 on Build GRCh38
rs777242723 Chr.11: 5323815-5323815 on Build GRCh38
rs754603872 Chr.11: 5323750-5323750 on Build GRCh38
rs368031685 Chr.11: 5323914-5323914 on Build GRCh38
rs749382965 Chr.11: 5323697-5323697 on Build GRCh38
rs761488654 Chr.11: 5323968-5323968 on Build GRCh38
rs7952293 Chr.11: 5323940-5323940 on Build GRCh38
rs907431317 Chr.11: 5323628-5323628 on Build GRCh38
rs766072373 Chr.11: 5323650-5323650 on Build GRCh38
rs755204482 Chr.11: 5323990-5323990 on Build GRCh38
rs770237512 Chr.11: 5323876-5323876 on Build GRCh38
rs760377882 Chr.11: 5323895-5323895 on Build GRCh38
rs753801014 Chr.11: 5323837-5323837 on Build GRCh38
rs764793730 Chr.11: 5323621-5323621 on Build GRCh38
rs7937237 Chr.11: 5323672-5323672 on Build GRCh38
rs202221282 Chr.11: 5323630-5323630 on Build GRCh38
rs766313727 Chr.11: 5323678-5323679 on Build GRCh38
rs900565653 Chr.11: 5324025-5324025 on Build GRCh38
rs764726412 Chr.11: 5323726-5323726 on Build GRCh38
rs772187246 Chr.11: 5323803-5323803 on Build GRCh38
rs986512316 Chr.11: 5323986-5323986 on Build GRCh38
rs750556277 Chr.11: 5323946-5323947 on Build GRCh38
rs767278785 Chr.11: 5323751-5323751 on Build GRCh38
rs778982907 Chr.11: 5323843-5323843 on Build GRCh38
rs763817910 Chr.11: 5323897-5323897 on Build GRCh38
rs759190687 Chr.11: 5323829-5323829 on Build GRCh38
rs201851252 Chr.11: 5323844-5323844 on Build GRCh38
rs867730593 Chr.11: 5323661-5323661 on Build GRCh38
rs758935334 Chr.11: 5323767-5323767 on Build GRCh38
rs757154370 Chr.11: 5323842-5323842 on Build GRCh38
rs10768793 Chr.11: 5323617-5323617 on Build GRCh38
rs746543267 Chr.11: 5324065-5324065 on Build GRCh38
rs370688356 Chr.11: 5323850-5323850 on Build GRCh38
rs138700784 Chr.11: 5323819-5323819 on Build GRCh38
rs144963209 Chr.11: 5323643-5323643 on Build GRCh38
rs866980907 Chr.11: 5324046-5324046 on Build GRCh38
rs144798172 Chr.11: 5323784-5323784 on Build GRCh38
rs747897739 Chr.11: 5324074-5324074 on Build GRCh38
rs374119453 Chr.11: 5323902-5323902 on Build GRCh38
rs777762331 Chr.11: 5323677-5323677 on Build GRCh38
rs140386840 Chr.11: 5323835-5323835 on Build GRCh38
rs145704673 Chr.11: 5323659-5323659 on Build GRCh38
rs373331835 Chr.11: 5323811-5323811 on Build GRCh38
rs949105971 Chr.11: 5323871-5323871 on Build GRCh38
rs149457927 Chr.11: 5323939-5323939 on Build GRCh38
rs983385820 Chr.11: 5324034-5324034 on Build GRCh38
rs942601462 Chr.11: 5323833-5323833 on Build GRCh38
rs377186037 Chr.11: 5323864-5323864 on Build GRCh38
rs746043607 Chr.11: 5323797-5323797 on Build GRCh38
rs772461820 Chr.11: 5323709-5323709 on Build GRCh38
rs779498626 Chr.11: 5324064-5324064 on Build GRCh38
rs373769005 Chr.11: 5323949-5323949 on Build GRCh38
rs765116418 Chr.11: 5323969-5323969 on Build GRCh38
rs754768270 Chr.11: 5323670-5323670 on Build GRCh38
rs376911782 Chr.11: 5323925-5323925 on Build GRCh38
rs893269189 Chr.11: 5323694-5323694 on Build GRCh38

View Full Product Details