Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 5754589-5755099 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR52N4
Entrez Gene ID
390072
Gene Name
olfactory receptor family 52 subfamily N member 4 (gene/pseudogene)
Gene Aliases
OR11-64
UniGene
Hs.553735
Gene Chromosome Location
Chr.11: 5722137-5755958 on Build GRCh38
Transcripts
NM_001005175.3 XM_017017711.1 XM_017017712.1 XM_017017713.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
467010 c.211A>G p.T71A Chr.11: 5754951-5754951 on Build GRCh38
3791604 c.14A>C p.N5T Chr.11: 5754754-5754754 on Build GRCh38
4795218 c.95C>T p.P32L Chr.11: 5754835-5754835 on Build GRCh38
283583 c.115G>A p.V39M Chr.11: 5754855-5754855 on Build GRCh38
3869571 c.84G>A p.W28* Chr.11: 5754824-5754824 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1000433196 Chr.11: 5754671-5754671 on Build GRCh38
rs771873459 Chr.11: 5755003-5755003 on Build GRCh38
rs765368851 Chr.11: 5755052-5755052 on Build GRCh38
rs267603021 Chr.11: 5754822-5754822 on Build GRCh38
rs956160207 Chr.11: 5754941-5754941 on Build GRCh38
rs768996708 Chr.11: 5754961-5754961 on Build GRCh38
rs560301831 Chr.11: 5754634-5754634 on Build GRCh38
rs780599104 Chr.11: 5754859-5754859 on Build GRCh38
rs756176720 Chr.11: 5754642-5754643 on Build GRCh38
rs376624755 Chr.11: 5754962-5754962 on Build GRCh38
rs778040330 Chr.11: 5754699-5754699 on Build GRCh38
rs549040408 Chr.11: 5754644-5754644 on Build GRCh38
rs75635340 Chr.11: 5754835-5754835 on Build GRCh38
rs749537367 Chr.11: 5754712-5754712 on Build GRCh38
rs371785567 Chr.11: 5754806-5754806 on Build GRCh38
rs754034580 Chr.11: 5754807-5754807 on Build GRCh38
rs993456510 Chr.11: 5754708-5754708 on Build GRCh38
rs369109501 Chr.11: 5754872-5754872 on Build GRCh38
rs146030201 Chr.11: 5755023-5755023 on Build GRCh38
rs745914968 Chr.11: 5754947-5754948 on Build GRCh38
rs766151093 Chr.11: 5754784-5754784 on Build GRCh38
rs527911027 Chr.11: 5754640-5754640 on Build GRCh38
rs531569145 Chr.11: 5754861-5754861 on Build GRCh38
rs748846231 Chr.11: 5754685-5754686 on Build GRCh38
rs868539171 Chr.11: 5755023-5755024 on Build GRCh38
rs374373349 Chr.11: 5754738-5754740 on Build GRCh38
rs776725509 Chr.11: 5754764-5754764 on Build GRCh38
rs779141741 Chr.11: 5754819-5754819 on Build GRCh38
rs377104220 Chr.11: 5754755-5754755 on Build GRCh38
rs752647270 Chr.11: 5754882-5754882 on Build GRCh38
rs868846018 Chr.11: 5755077-5755077 on Build GRCh38
rs751804062 Chr.11: 5755063-5755063 on Build GRCh38
rs375554405 Chr.11: 5754995-5754995 on Build GRCh38
rs775867412 Chr.11: 5754744-5754744 on Build GRCh38
rs185750636 Chr.11: 5754863-5754863 on Build GRCh38
rs930799860 Chr.11: 5755049-5755049 on Build GRCh38
rs567439185 Chr.11: 5754728-5754728 on Build GRCh38
rs547409437 Chr.11: 5754979-5754979 on Build GRCh38
rs369523700 Chr.11: 5754789-5754789 on Build GRCh38
rs997195395 Chr.11: 5754865-5754865 on Build GRCh38
rs750704734 Chr.11: 5754918-5754918 on Build GRCh38
rs761871718 Chr.11: 5755032-5755032 on Build GRCh38
rs145181208 Chr.11: 5754860-5754861 on Build GRCh38
rs772340611 Chr.11: 5754847-5754847 on Build GRCh38
rs769959782 Chr.11: 5755019-5755020 on Build GRCh38
rs762735356 Chr.11: 5754769-5754769 on Build GRCh38
rs757338261 Chr.11: 5754897-5754897 on Build GRCh38
rs920787752 Chr.11: 5754751-5754751 on Build GRCh38
rs368536720 Chr.11: 5754734-5754734 on Build GRCh38
rs7936512 Chr.11: 5755057-5755057 on Build GRCh38
rs1048188713 Chr.11: 5754779-5754779 on Build GRCh38
rs755382904 Chr.11: 5755068-5755068 on Build GRCh38
rs759390967 Chr.11: 5754873-5754873 on Build GRCh38
rs1000198692 Chr.11: 5754894-5754894 on Build GRCh38
rs199608836 Chr.11: 5755059-5755059 on Build GRCh38
rs746167351 Chr.11: 5754845-5754845 on Build GRCh38
rs762582198 Chr.11: 5754864-5754864 on Build GRCh38
rs767430015 Chr.11: 5754795-5754795 on Build GRCh38
rs373568225 Chr.11: 5754717-5754717 on Build GRCh38
rs772531595 Chr.11: 5754739-5754739 on Build GRCh38
rs756159640 Chr.11: 5754692-5754692 on Build GRCh38
rs756212038 Chr.11: 5754804-5754804 on Build GRCh38
rs865957835 Chr.11: 5755038-5755038 on Build GRCh38
rs764111958 Chr.11: 5754885-5754885 on Build GRCh38
rs761332651 Chr.11: 5754749-5754749 on Build GRCh38
rs397723036 Chr.11: 5754684-5754685 on Build GRCh38
rs917264333 Chr.11: 5754970-5754970 on Build GRCh38
rs1032963748 Chr.11: 5754673-5754673 on Build GRCh38
rs34221828 Chr.11: 5754674-5754675 on Build GRCh38
rs924592781 Chr.11: 5754615-5754615 on Build GRCh38
rs190345271 Chr.11: 5754973-5754973 on Build GRCh38
rs779051435 Chr.11: 5754908-5754908 on Build GRCh38
rs770640997 Chr.11: 5754870-5754870 on Build GRCh38
rs200603805 Chr.11: 5755029-5755029 on Build GRCh38
rs776564930 Chr.11: 5755031-5755031 on Build GRCh38
rs780324701 Chr.11: 5754923-5754923 on Build GRCh38
rs566832049 Chr.11: 5754673-5754674 on Build GRCh38
rs759299253 Chr.11: 5754994-5754994 on Build GRCh38
rs746257299 Chr.11: 5754738-5754738 on Build GRCh38
rs748747858 Chr.11: 5754967-5754967 on Build GRCh38
rs372632967 Chr.11: 5754754-5754754 on Build GRCh38
rs760575260 Chr.11: 5754884-5754884 on Build GRCh38
rs1003888889 Chr.11: 5754798-5754798 on Build GRCh38
rs751360439 Chr.11: 5754786-5754786 on Build GRCh38
rs201497778 Chr.11: 5755021-5755021 on Build GRCh38
rs753878461 Chr.11: 5754892-5754892 on Build GRCh38
rs199870684 Chr.11: 5754814-5754814 on Build GRCh38
rs752632287 Chr.11: 5754797-5754797 on Build GRCh38
rs373101849 Chr.11: 5754874-5754874 on Build GRCh38
rs968540852 Chr.11: 5754887-5754887 on Build GRCh38
rs759407480 Chr.11: 5754788-5754788 on Build GRCh38
rs770622555 Chr.11: 5754976-5754976 on Build GRCh38
rs747528380 Chr.11: 5754953-5754953 on Build GRCh38

View Full Product Details