Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 6484060-6484607 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TIMM10B
Entrez Gene ID
26515
Gene Name
translocase of inner mitochondrial membrane 10 homolog B (yeast)
Gene Aliases
FXC1, TIM10B, Tim9b
UniGene
Hs.54943
Gene Chromosome Location
Chr.11: 6481447-6484681 on Build GRCh38
Transcripts
NM_012192.3

SNP Detail

SNP ID Chromosome Location
rs752316708 Chr.11: 6484444-6484444 on Build GRCh38
rs1002259766 Chr.11: 6484191-6484191 on Build GRCh38
rs778698450 Chr.11: 6484440-6484440 on Build GRCh38
rs958394682 Chr.11: 6484124-6484124 on Build GRCh38
rs144413908 Chr.11: 6484574-6484574 on Build GRCh38
rs991771606 Chr.11: 6484238-6484238 on Build GRCh38
rs533211334 Chr.11: 6484353-6484353 on Build GRCh38
rs189336483 Chr.11: 6484229-6484229 on Build GRCh38
rs750815546 Chr.11: 6484350-6484350 on Build GRCh38
rs1140221 Chr.11: 6484294-6484294 on Build GRCh38
rs933395857 Chr.11: 6484112-6484113 on Build GRCh38
rs1008015439 Chr.11: 6484095-6484095 on Build GRCh38
rs562985432 Chr.11: 6484344-6484344 on Build GRCh38
rs1020804075 Chr.11: 6484237-6484237 on Build GRCh38
rs906126858 Chr.11: 6484355-6484355 on Build GRCh38
rs150061600 Chr.11: 6484527-6484527 on Build GRCh38
rs1011429237 Chr.11: 6484432-6484432 on Build GRCh38
rs991876533 Chr.11: 6484222-6484222 on Build GRCh38
rs59421287 Chr.11: 6484239-6484239 on Build GRCh38
rs1007716504 Chr.11: 6484374-6484374 on Build GRCh38
rs529421241 Chr.11: 6484245-6484245 on Build GRCh38
rs964762460 Chr.11: 6484317-6484317 on Build GRCh38
rs928438416 Chr.11: 6484181-6484181 on Build GRCh38
rs762429524 Chr.11: 6484283-6484283 on Build GRCh38
rs1055209220 Chr.11: 6484415-6484415 on Build GRCh38
rs1000681000 Chr.11: 6484558-6484558 on Build GRCh38
rs201233062 Chr.11: 6484214-6484214 on Build GRCh38
rs139464704 Chr.11: 6484388-6484388 on Build GRCh38
rs1035569774 Chr.11: 6484518-6484518 on Build GRCh38
rs375452183 Chr.11: 6484091-6484091 on Build GRCh38
rs112798999 Chr.11: 6484151-6484151 on Build GRCh38
rs561962225 Chr.11: 6484243-6484243 on Build GRCh38
rs556201921 Chr.11: 6484119-6484119 on Build GRCh38
rs977272225 Chr.11: 6484445-6484445 on Build GRCh38
rs975167236 Chr.11: 6484332-6484332 on Build GRCh38
rs879828500 Chr.11: 6484107-6484107 on Build GRCh38
rs549311429 Chr.11: 6484179-6484179 on Build GRCh38
rs966561815 Chr.11: 6484545-6484545 on Build GRCh38
rs960265219 Chr.11: 6484519-6484519 on Build GRCh38
rs890749002 Chr.11: 6484352-6484352 on Build GRCh38
rs1047897294 Chr.11: 6484127-6484127 on Build GRCh38
rs967246210 Chr.11: 6484174-6484174 on Build GRCh38
rs929510457 Chr.11: 6484338-6484338 on Build GRCh38
rs1013382572 Chr.11: 6484429-6484429 on Build GRCh38
rs941760523 Chr.11: 6484557-6484557 on Build GRCh38
rs1016464285 Chr.11: 6484143-6484143 on Build GRCh38
rs966530390 Chr.11: 6484256-6484256 on Build GRCh38
rs537961615 Chr.11: 6484096-6484096 on Build GRCh38
rs1006469244 Chr.11: 6484140-6484140 on Build GRCh38
rs886574559 Chr.11: 6484129-6484129 on Build GRCh38
rs942191209 Chr.11: 6484349-6484349 on Build GRCh38
rs974653228 Chr.11: 6484576-6484576 on Build GRCh38
rs527599549 Chr.11: 6484549-6484549 on Build GRCh38
rs1020707363 Chr.11: 6484515-6484515 on Build GRCh38
rs923267686 Chr.11: 6484334-6484334 on Build GRCh38
rs896116134 Chr.11: 6484426-6484426 on Build GRCh38
rs977322914 Chr.11: 6484175-6484175 on Build GRCh38
rs370812940 Chr.11: 6484555-6484555 on Build GRCh38
rs1033330276 Chr.11: 6484201-6484201 on Build GRCh38
rs960753520 Chr.11: 6484213-6484213 on Build GRCh38
rs989481089 Chr.11: 6484150-6484150 on Build GRCh38
rs763189199 Chr.11: 6484182-6484182 on Build GRCh38
rs1020885323 Chr.11: 6484441-6484441 on Build GRCh38
rs1018216262 Chr.11: 6484383-6484383 on Build GRCh38
rs1052019120 Chr.11: 6484343-6484343 on Build GRCh38
rs933414122 Chr.11: 6484341-6484341 on Build GRCh38
rs570275947 Chr.11: 6484093-6484093 on Build GRCh38

View Full Product Details