Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 31369723-31370228 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCDC1
Entrez Gene ID
341019
Gene Name
doublecortin domain containing 1
Gene Aliases
-
UniGene
Hs.127405
Gene Chromosome Location
Chr.11: 31262624-31369810 on Build GRCh38
Transcripts
NM_181807.3 NM_181706.4 XM_017017216.1

Gene Symbol
DNAJC24
Entrez Gene ID
120526
Gene Name
DnaJ heat shock protein family (Hsp40) member C24
Gene Aliases
DPH4, JJJ3, ZCSL3
UniGene
Hs.187269
Gene Chromosome Location
Chr.11: 31369829-31432835 on Build GRCh38
Transcripts
NM_181807.3 NM_181706.4 XM_017017216.1

SNP Detail

SNP ID Chromosome Location
rs553185165 Chr.11: 31370052-31370052 on Build GRCh38
rs530789876 Chr.11: 31370196-31370196 on Build GRCh38
rs940436319 Chr.11: 31369841-31369841 on Build GRCh38
rs997731973 Chr.11: 31369856-31369856 on Build GRCh38
rs541658258 Chr.11: 31370031-31370031 on Build GRCh38
rs952954176 Chr.11: 31370077-31370077 on Build GRCh38
rs570674089 Chr.11: 31369784-31369784 on Build GRCh38
rs1036528468 Chr.11: 31369848-31369848 on Build GRCh38
rs116391966 Chr.11: 31369790-31369790 on Build GRCh38
rs933479308 Chr.11: 31370091-31370091 on Build GRCh38
rs34335469 Chr.11: 31369852-31369853 on Build GRCh38
rs903144263 Chr.11: 31370137-31370137 on Build GRCh38
rs188456366 Chr.11: 31369792-31369792 on Build GRCh38
rs1022031962 Chr.11: 31369929-31369929 on Build GRCh38
rs574913830 Chr.11: 31370090-31370090 on Build GRCh38
rs949017280 Chr.11: 31369841-31369843 on Build GRCh38
rs563724642 Chr.11: 31370171-31370171 on Build GRCh38
rs1042024068 Chr.11: 31370120-31370120 on Build GRCh38
rs770602651 Chr.11: 31369819-31369819 on Build GRCh38
rs1001112604 Chr.11: 31369870-31369871 on Build GRCh38
rs889404326 Chr.11: 31370099-31370099 on Build GRCh38
rs889133983 Chr.11: 31369887-31369887 on Build GRCh38
rs960501442 Chr.11: 31369957-31369957 on Build GRCh38
rs569990399 Chr.11: 31369926-31369926 on Build GRCh38
rs541838409 Chr.11: 31370123-31370123 on Build GRCh38
rs965613260 Chr.11: 31369742-31369742 on Build GRCh38
rs1051014272 Chr.11: 31370093-31370093 on Build GRCh38
rs759897498 Chr.11: 31370040-31370040 on Build GRCh38
rs998947214 Chr.11: 31370157-31370157 on Build GRCh38
rs902423238 Chr.11: 31369862-31369862 on Build GRCh38
rs916246780 Chr.11: 31369801-31369801 on Build GRCh38
rs1043099318 Chr.11: 31370127-31370127 on Build GRCh38
rs756209806 Chr.11: 31369754-31369754 on Build GRCh38
rs573901171 Chr.11: 31369880-31369880 on Build GRCh38
rs181643106 Chr.11: 31369821-31369821 on Build GRCh38
rs776933081 Chr.11: 31369938-31369938 on Build GRCh38
rs1029217105 Chr.11: 31369879-31369879 on Build GRCh38
rs923453669 Chr.11: 31370078-31370078 on Build GRCh38
rs537368598 Chr.11: 31369928-31369928 on Build GRCh38
rs992505612 Chr.11: 31369966-31369966 on Build GRCh38
rs1030841006 Chr.11: 31369948-31369948 on Build GRCh38
rs931512408 Chr.11: 31369775-31369775 on Build GRCh38
rs947724302 Chr.11: 31369818-31369818 on Build GRCh38
rs568654927 Chr.11: 31369774-31369774 on Build GRCh38
rs967487205 Chr.11: 31369936-31369936 on Build GRCh38
rs765657123 Chr.11: 31370164-31370164 on Build GRCh38
rs1032135147 Chr.11: 31369886-31369886 on Build GRCh38
rs141305806 Chr.11: 31370008-31370008 on Build GRCh38
rs1042369315 Chr.11: 31369851-31369851 on Build GRCh38
rs556948635 Chr.11: 31369855-31369855 on Build GRCh38
rs949069217 Chr.11: 31370117-31370117 on Build GRCh38
rs1036038157 Chr.11: 31370076-31370076 on Build GRCh38
rs916527598 Chr.11: 31370005-31370005 on Build GRCh38
rs377611888 Chr.11: 31369755-31369755 on Build GRCh38
rs558686094 Chr.11: 31369959-31369959 on Build GRCh38
rs568194229 Chr.11: 31369794-31369794 on Build GRCh38
rs972987393 Chr.11: 31369753-31369753 on Build GRCh38
rs1048521174 Chr.11: 31369789-31369789 on Build GRCh38

View Full Product Details