Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 36575496-36576004 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAG1
Entrez Gene ID
5896
Gene Name
recombination activating gene 1
Gene Aliases
RAG-1, RNF74
UniGene
Hs.538979
Gene Chromosome Location
Chr.11: 36510366-36579762 on Build GRCh38
Transcripts
NM_000448.2 XM_005253041.4 XM_011520250.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1722592 c.2464C>T p.P822S Chr.11: 36575768-36575768 on Build GRCh38
7202525 c.2617A>G p.I873V Chr.11: 36575921-36575921 on Build GRCh38
1168721 c.2333G>A p.R778Q Chr.11: 36575637-36575637 on Build GRCh38
3809167 c.2486G>T p.R829M Chr.11: 36575790-36575790 on Build GRCh38
6218340 c.2327G>A p.R776Q Chr.11: 36575631-36575631 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771140517 Chr.11: 36575850-36575850 on Build GRCh38
rs200300629 Chr.11: 36575649-36575649 on Build GRCh38
rs764301645 Chr.11: 36575944-36575944 on Build GRCh38
rs758236480 Chr.11: 36575892-36575892 on Build GRCh38
rs199474690 Chr.11: 36575868-36575868 on Build GRCh38
rs762107952 Chr.11: 36575523-36575523 on Build GRCh38
rs186717025 Chr.11: 36575831-36575831 on Build GRCh38
rs754502950 Chr.11: 36575652-36575652 on Build GRCh38
rs772380086 Chr.11: 36575941-36575941 on Build GRCh38
rs760394954 Chr.11: 36575899-36575900 on Build GRCh38
rs769483414 Chr.11: 36575569-36575569 on Build GRCh38
rs374387337 Chr.11: 36575622-36575622 on Build GRCh38
rs999020322 Chr.11: 36575846-36575846 on Build GRCh38
rs753382928 Chr.11: 36575796-36575796 on Build GRCh38
rs754073191 Chr.11: 36575519-36575519 on Build GRCh38
rs563140654 Chr.11: 36575588-36575588 on Build GRCh38
rs776061406 Chr.11: 36575961-36575961 on Build GRCh38
rs745744137 Chr.11: 36575743-36575743 on Build GRCh38
rs752304188 Chr.11: 36575791-36575791 on Build GRCh38
rs773503251 Chr.11: 36575835-36575835 on Build GRCh38
rs370390229 Chr.11: 36575614-36575614 on Build GRCh38
rs748296558 Chr.11: 36575826-36575826 on Build GRCh38
rs770953505 Chr.11: 36575580-36575580 on Build GRCh38
rs142345523 Chr.11: 36575856-36575856 on Build GRCh38
rs773002320 Chr.11: 36575834-36575834 on Build GRCh38
rs141560248 Chr.11: 36575875-36575875 on Build GRCh38
rs764967774 Chr.11: 36575887-36575887 on Build GRCh38
rs147656090 Chr.11: 36575772-36575772 on Build GRCh38
rs2227973 Chr.11: 36575763-36575763 on Build GRCh38
rs1014281478 Chr.11: 36575534-36575534 on Build GRCh38
rs766907859 Chr.11: 36575629-36575629 on Build GRCh38
rs777644555 Chr.11: 36575917-36575917 on Build GRCh38
rs777272905 Chr.11: 36575705-36575705 on Build GRCh38
rs4151033 Chr.11: 36575942-36575942 on Build GRCh38
rs757133059 Chr.11: 36575810-36575810 on Build GRCh38
rs748827810 Chr.11: 36575545-36575545 on Build GRCh38
rs4151034 Chr.11: 36575963-36575963 on Build GRCh38
rs145772007 Chr.11: 36575930-36575930 on Build GRCh38
rs61758791 Chr.11: 36575755-36575755 on Build GRCh38
rs763202008 Chr.11: 36575628-36575628 on Build GRCh38
rs376140107 Chr.11: 36575873-36575873 on Build GRCh38
rs756228686 Chr.11: 36575915-36575915 on Build GRCh38
rs748162851 Chr.11: 36575749-36575749 on Build GRCh38
rs755551812 Chr.11: 36575792-36575792 on Build GRCh38
rs866862677 Chr.11: 36575925-36575925 on Build GRCh38
rs759008821 Chr.11: 36575546-36575546 on Build GRCh38
rs542851147 Chr.11: 36575549-36575549 on Build GRCh38
rs763838960 Chr.11: 36575793-36575793 on Build GRCh38
rs753534445 Chr.11: 36575874-36575874 on Build GRCh38
rs768860215 Chr.11: 36575746-36575746 on Build GRCh38
rs151322536 Chr.11: 36575708-36575708 on Build GRCh38
rs961233109 Chr.11: 36575647-36575647 on Build GRCh38
rs768260595 Chr.11: 36575612-36575612 on Build GRCh38
rs768001715 Chr.11: 36575782-36575782 on Build GRCh38
rs61752933 Chr.11: 36575732-36575732 on Build GRCh38
rs199474691 Chr.11: 36575958-36575958 on Build GRCh38
rs760869236 Chr.11: 36575590-36575590 on Build GRCh38
rs749410483 Chr.11: 36575836-36575836 on Build GRCh38
rs774757762 Chr.11: 36575853-36575853 on Build GRCh38
rs142419805 Chr.11: 36575617-36575617 on Build GRCh38
rs121918572 Chr.11: 36575630-36575630 on Build GRCh38
rs199474687 Chr.11: 36575562-36575562 on Build GRCh38
rs193922462 Chr.11: 36575907-36575907 on Build GRCh38
rs377690758 Chr.11: 36575615-36575615 on Build GRCh38
rs760255456 Chr.11: 36575858-36575858 on Build GRCh38
rs868819938 Chr.11: 36575576-36575576 on Build GRCh38
rs778785063 Chr.11: 36575820-36575820 on Build GRCh38
rs984404819 Chr.11: 36575610-36575610 on Build GRCh38
rs751115703 Chr.11: 36575554-36575554 on Build GRCh38
rs752020152 Chr.11: 36575636-36575636 on Build GRCh38
rs751400793 Chr.11: 36575911-36575911 on Build GRCh38
rs972558635 Chr.11: 36575714-36575714 on Build GRCh38
rs777005609 Chr.11: 36575608-36575608 on Build GRCh38
rs121918569 Chr.11: 36575637-36575637 on Build GRCh38
rs773703055 Chr.11: 36575730-36575730 on Build GRCh38
rs1022967472 Chr.11: 36575903-36575903 on Build GRCh38
rs749027430 Chr.11: 36575579-36575579 on Build GRCh38
rs752146257 Chr.11: 36575666-36575666 on Build GRCh38
rs749149886 Chr.11: 36575722-36575722 on Build GRCh38
rs909724257 Chr.11: 36575634-36575634 on Build GRCh38
rs104894282 Chr.11: 36575624-36575624 on Build GRCh38
rs866509921 Chr.11: 36575678-36575678 on Build GRCh38
rs531271076 Chr.11: 36575695-36575695 on Build GRCh38
rs768809293 Chr.11: 36575595-36575595 on Build GRCh38
rs567443823 Chr.11: 36575857-36575857 on Build GRCh38
rs745652258 Chr.11: 36575821-36575821 on Build GRCh38
rs200264827 Chr.11: 36575929-36575929 on Build GRCh38
rs770103028 Chr.11: 36575751-36575751 on Build GRCh38
rs970428951 Chr.11: 36575904-36575904 on Build GRCh38
rs765578408 Chr.11: 36575526-36575526 on Build GRCh38
rs777693786 Chr.11: 36575575-36575575 on Build GRCh38
rs758468024 Chr.11: 36575824-36575824 on Build GRCh38
rs104894287 Chr.11: 36575825-36575825 on Build GRCh38
rs761022716 Chr.11: 36575952-36575952 on Build GRCh38
rs1014725960 Chr.11: 36575882-36575882 on Build GRCh38
rs749360497 Chr.11: 36575919-36575919 on Build GRCh38
rs776915729 Chr.11: 36575747-36575747 on Build GRCh38
rs377252087 Chr.11: 36575541-36575541 on Build GRCh38
rs766950619 Chr.11: 36575665-36575665 on Build GRCh38
rs201313833 Chr.11: 36575909-36575909 on Build GRCh38
rs759876731 Chr.11: 36575855-36575855 on Build GRCh38
rs570885960 Chr.11: 36575756-36575756 on Build GRCh38
rs747746845 Chr.11: 36575563-36575563 on Build GRCh38

View Full Product Details