Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 44264791-44265292 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALX4
Entrez Gene ID
60529
Gene Name
ALX homeobox 4
Gene Aliases
CRS5, FND2
UniGene
Hs.436055
Gene Chromosome Location
Chr.11: 44260728-44310166 on Build GRCh38
Transcripts
NM_021926.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6598962 c.1102T>C p.F368L Chr.11: 44264988-44264988 on Build GRCh38
5595671 c.1230C>T p.A410A Chr.11: 44264860-44264860 on Build GRCh38
236982 c.918G>A p.P306P Chr.11: 44265172-44265172 on Build GRCh38
6598960 c.1181C>T p.A394V Chr.11: 44264909-44264909 on Build GRCh38
1182653 c.1091T>C p.M364T Chr.11: 44264999-44264999 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs186244229 Chr.11: 44265114-44265114 on Build GRCh38
rs768446841 Chr.11: 44265000-44265000 on Build GRCh38
rs762716140 Chr.11: 44265126-44265126 on Build GRCh38
rs190814077 Chr.11: 44265262-44265262 on Build GRCh38
rs752640074 Chr.11: 44265143-44265143 on Build GRCh38
rs755595641 Chr.11: 44264911-44264911 on Build GRCh38
rs748695742 Chr.11: 44265175-44265175 on Build GRCh38
rs904598017 Chr.11: 44264933-44264933 on Build GRCh38
rs559981237 Chr.11: 44264826-44264826 on Build GRCh38
rs868038992 Chr.11: 44264867-44264867 on Build GRCh38
rs200122905 Chr.11: 44264876-44264876 on Build GRCh38
rs369740288 Chr.11: 44264910-44264910 on Build GRCh38
rs148027225 Chr.11: 44264973-44264973 on Build GRCh38
rs756084826 Chr.11: 44265148-44265148 on Build GRCh38
rs372442372 Chr.11: 44264932-44264932 on Build GRCh38
rs761250353 Chr.11: 44265032-44265032 on Build GRCh38
rs891814573 Chr.11: 44265115-44265115 on Build GRCh38
rs764490498 Chr.11: 44265021-44265021 on Build GRCh38
rs355697 Chr.11: 44264944-44264944 on Build GRCh38
rs61737293 Chr.11: 44265005-44265005 on Build GRCh38
rs144198846 Chr.11: 44265078-44265078 on Build GRCh38
rs150706927 Chr.11: 44265154-44265154 on Build GRCh38
rs59194417 Chr.11: 44265255-44265255 on Build GRCh38
rs754671277 Chr.11: 44265131-44265131 on Build GRCh38
rs779876897 Chr.11: 44264994-44264994 on Build GRCh38
rs1019429549 Chr.11: 44264823-44264823 on Build GRCh38
rs1015314548 Chr.11: 44265159-44265159 on Build GRCh38
rs1039616189 Chr.11: 44265010-44265010 on Build GRCh38
rs745849255 Chr.11: 44264872-44264872 on Build GRCh38
rs375614713 Chr.11: 44265204-44265204 on Build GRCh38
rs952423649 Chr.11: 44265024-44265024 on Build GRCh38
rs750030599 Chr.11: 44265038-44265038 on Build GRCh38
rs780076792 Chr.11: 44265162-44265162 on Build GRCh38
rs147215488 Chr.11: 44265034-44265034 on Build GRCh38
rs780898515 Chr.11: 44264829-44264829 on Build GRCh38
rs1028561582 Chr.11: 44265147-44265147 on Build GRCh38
rs755790488 Chr.11: 44264831-44264831 on Build GRCh38
rs775689545 Chr.11: 44265015-44265015 on Build GRCh38
rs200498976 Chr.11: 44264821-44264821 on Build GRCh38
rs533211314 Chr.11: 44264830-44264830 on Build GRCh38
rs201245023 Chr.11: 44265004-44265004 on Build GRCh38
rs776002938 Chr.11: 44265033-44265035 on Build GRCh38
rs773423723 Chr.11: 44264897-44264897 on Build GRCh38
rs759404039 Chr.11: 44265212-44265212 on Build GRCh38
rs763307132 Chr.11: 44264940-44264940 on Build GRCh38
rs760748844 Chr.11: 44265218-44265218 on Build GRCh38
rs375068808 Chr.11: 44264884-44264884 on Build GRCh38
rs779972509 Chr.11: 44264934-44264934 on Build GRCh38
rs753201422 Chr.11: 44264950-44264950 on Build GRCh38
rs140652481 Chr.11: 44265055-44265055 on Build GRCh38
rs200431603 Chr.11: 44264930-44264930 on Build GRCh38
rs749052167 Chr.11: 44264836-44264836 on Build GRCh38
rs763248407 Chr.11: 44264900-44264900 on Build GRCh38
rs920979894 Chr.11: 44264832-44264832 on Build GRCh38
rs751941183 Chr.11: 44264905-44264905 on Build GRCh38
rs757361317 Chr.11: 44265075-44265075 on Build GRCh38
rs751290539 Chr.11: 44265054-44265054 on Build GRCh38
rs369085844 Chr.11: 44264995-44264995 on Build GRCh38
rs766809146 Chr.11: 44264901-44264901 on Build GRCh38
rs773825623 Chr.11: 44265211-44265211 on Build GRCh38
rs922576689 Chr.11: 44265244-44265244 on Build GRCh38
rs747416406 Chr.11: 44265163-44265163 on Build GRCh38
rs764119148 Chr.11: 44265224-44265224 on Build GRCh38
rs549463745 Chr.11: 44264938-44264938 on Build GRCh38
rs760004486 Chr.11: 44265007-44265007 on Build GRCh38
rs753270187 Chr.11: 44264970-44264970 on Build GRCh38
rs372163762 Chr.11: 44265110-44265110 on Build GRCh38
rs768153687 Chr.11: 44264949-44264949 on Build GRCh38
rs770265382 Chr.11: 44265187-44265187 on Build GRCh38
rs971229762 Chr.11: 44265132-44265132 on Build GRCh38
rs758129706 Chr.11: 44264992-44264992 on Build GRCh38
rs587777702 Chr.11: 44265105-44265114 on Build GRCh38
rs943508604 Chr.11: 44265001-44265001 on Build GRCh38
rs367915603 Chr.11: 44265109-44265109 on Build GRCh38
rs561018820 Chr.11: 44265127-44265127 on Build GRCh38
rs149365713 Chr.11: 44264908-44264908 on Build GRCh38
rs750817902 Chr.11: 44264818-44264818 on Build GRCh38
rs368387718 Chr.11: 44265217-44265217 on Build GRCh38
rs574187575 Chr.11: 44265130-44265130 on Build GRCh38
rs774043649 Chr.11: 44264909-44264909 on Build GRCh38
rs563686757 Chr.11: 44265243-44265243 on Build GRCh38
rs369302027 Chr.11: 44264834-44264834 on Build GRCh38
rs749390789 Chr.11: 44265056-44265056 on Build GRCh38
rs747334899 Chr.11: 44265079-44265079 on Build GRCh38
rs770666353 Chr.11: 44264838-44264838 on Build GRCh38
rs145904583 Chr.11: 44265172-44265172 on Build GRCh38
rs749632326 Chr.11: 44264825-44264825 on Build GRCh38
rs756876780 Chr.11: 44264927-44264927 on Build GRCh38
rs753493510 Chr.11: 44264917-44264917 on Build GRCh38
rs758355772 Chr.11: 44265230-44265230 on Build GRCh38
rs776186258 Chr.11: 44265028-44265028 on Build GRCh38
rs766238204 Chr.11: 44265049-44265049 on Build GRCh38
rs541605039 Chr.11: 44265151-44265151 on Build GRCh38
rs149897209 Chr.11: 44265173-44265173 on Build GRCh38
rs774924693 Chr.11: 44264947-44264947 on Build GRCh38
rs774306730 Chr.11: 44264863-44264863 on Build GRCh38
rs772768304 Chr.11: 44265201-44265201 on Build GRCh38
rs772445432 Chr.11: 44265008-44265008 on Build GRCh38
rs542338601 Chr.11: 44265118-44265118 on Build GRCh38
rs985033532 Chr.11: 44264976-44264976 on Build GRCh38
rs778529834 Chr.11: 44264978-44264978 on Build GRCh38
rs901924979 Chr.11: 44264837-44264837 on Build GRCh38
rs777315924 Chr.11: 44264912-44264912 on Build GRCh38
rs772158840 Chr.11: 44265077-44265077 on Build GRCh38
rs376675326 Chr.11: 44264941-44264941 on Build GRCh38
rs770630271 Chr.11: 44265210-44265210 on Build GRCh38
rs750221052 Chr.11: 44264984-44264984 on Build GRCh38
rs763267255 Chr.11: 44265211-44265212 on Build GRCh38
rs763704637 Chr.11: 44264961-44264961 on Build GRCh38
rs1015282003 Chr.11: 44265040-44265040 on Build GRCh38
rs778724337 Chr.11: 44264928-44264928 on Build GRCh38
rs767511619 Chr.11: 44265141-44265141 on Build GRCh38
rs980674490 Chr.11: 44265098-44265098 on Build GRCh38
rs369033845 Chr.11: 44265048-44265048 on Build GRCh38
rs760189256 Chr.11: 44265089-44265089 on Build GRCh38
rs748187496 Chr.11: 44264937-44264937 on Build GRCh38
rs770986777 Chr.11: 44264875-44264875 on Build GRCh38
rs144742417 Chr.11: 44264957-44264957 on Build GRCh38
rs372004471 Chr.11: 44265226-44265226 on Build GRCh38
rs1041001928 Chr.11: 44265122-44265122 on Build GRCh38
rs759914813 Chr.11: 44264948-44264948 on Build GRCh38
rs888252724 Chr.11: 44265150-44265150 on Build GRCh38
rs1040761259 Chr.11: 44265239-44265239 on Build GRCh38
rs3802805 Chr.11: 44265016-44265016 on Build GRCh38
rs774659166 Chr.11: 44265006-44265006 on Build GRCh38
rs953816581 Chr.11: 44265249-44265249 on Build GRCh38
rs200251401 Chr.11: 44265231-44265231 on Build GRCh38

View Full Product Details