Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 44764347-44764842 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSPAN18
Entrez Gene ID
90139
Gene Name
tetraspanin 18
Gene Aliases
TSPAN
UniGene
Hs.385634
Gene Chromosome Location
Chr.11: 44727148-44932427 on Build GRCh38
Transcripts
NM_130783.4 XM_005253217.3 XM_006718372.3 XM_006718373.3 XM_011520459.2 XM_017018530.1 XM_017018531.1

SNP Detail

SNP ID Chromosome Location
rs1015580645 Chr.11: 44764687-44764687 on Build GRCh38
rs1001350815 Chr.11: 44764702-44764702 on Build GRCh38
rs1024522862 Chr.11: 44764749-44764749 on Build GRCh38
rs112614792 Chr.11: 44764767-44764767 on Build GRCh38
rs925721079 Chr.11: 44764555-44764555 on Build GRCh38
rs772786670 Chr.11: 44764817-44764817 on Build GRCh38
rs1049941551 Chr.11: 44764617-44764617 on Build GRCh38
rs1052763849 Chr.11: 44764651-44764651 on Build GRCh38
rs768388515 Chr.11: 44764524-44764524 on Build GRCh38
rs920006896 Chr.11: 44764614-44764614 on Build GRCh38
rs1008809448 Chr.11: 44764596-44764596 on Build GRCh38
rs961749640 Chr.11: 44764558-44764558 on Build GRCh38
rs12273904 Chr.11: 44764673-44764673 on Build GRCh38
rs974486102 Chr.11: 44764772-44764772 on Build GRCh38
rs1044092804 Chr.11: 44764548-44764548 on Build GRCh38
rs542515043 Chr.11: 44764529-44764529 on Build GRCh38
rs182171278 Chr.11: 44764797-44764797 on Build GRCh38
rs1035510743 Chr.11: 44764717-44764717 on Build GRCh38
rs144539683 Chr.11: 44764728-44764728 on Build GRCh38
rs1004788602 Chr.11: 44764742-44764742 on Build GRCh38
rs547759207 Chr.11: 44764807-44764807 on Build GRCh38
rs1028381903 Chr.11: 44764646-44764646 on Build GRCh38
rs1040732593 Chr.11: 44764606-44764606 on Build GRCh38
rs200143130 Chr.11: 44764616-44764616 on Build GRCh38
rs552400902 Chr.11: 44764642-44764642 on Build GRCh38
rs1028413039 Chr.11: 44764368-44764368 on Build GRCh38
rs1011359962 Chr.11: 44764483-44764483 on Build GRCh38
rs1035541856 Chr.11: 44764516-44764516 on Build GRCh38
rs984150638 Chr.11: 44764380-44764380 on Build GRCh38
rs763541365 Chr.11: 44764808-44764808 on Build GRCh38
rs935646218 Chr.11: 44764572-44764572 on Build GRCh38
rs1001781951 Chr.11: 44764513-44764513 on Build GRCh38
rs77473655 Chr.11: 44764442-44764442 on Build GRCh38
rs912802230 Chr.11: 44764621-44764621 on Build GRCh38
rs1052771196 Chr.11: 44764575-44764575 on Build GRCh38
rs961707566 Chr.11: 44764716-44764716 on Build GRCh38
rs940133005 Chr.11: 44764377-44764377 on Build GRCh38
rs905621997 Chr.11: 44764689-44764689 on Build GRCh38
rs980258711 Chr.11: 44764786-44764786 on Build GRCh38
rs770233818 Chr.11: 44764521-44764521 on Build GRCh38
rs974222229 Chr.11: 44764567-44764567 on Build GRCh38
rs969878987 Chr.11: 44764498-44764498 on Build GRCh38
rs185653930 Chr.11: 44764632-44764632 on Build GRCh38
rs961629834 Chr.11: 44764464-44764464 on Build GRCh38
rs932808164 Chr.11: 44764615-44764615 on Build GRCh38
rs985615994 Chr.11: 44764815-44764815 on Build GRCh38
rs535441120 Chr.11: 44764794-44764794 on Build GRCh38
rs1038928101 Chr.11: 44764391-44764391 on Build GRCh38
rs775693687 Chr.11: 44764635-44764635 on Build GRCh38
rs1021875513 Chr.11: 44764494-44764494 on Build GRCh38
rs992101962 Chr.11: 44764536-44764536 on Build GRCh38
rs932891615 Chr.11: 44764412-44764415 on Build GRCh38
rs573527209 Chr.11: 44764471-44764471 on Build GRCh38
rs531726435 Chr.11: 44764685-44764685 on Build GRCh38
rs1027049258 Chr.11: 44764774-44764774 on Build GRCh38
rs952657370 Chr.11: 44764375-44764375 on Build GRCh38
rs189968578 Chr.11: 44764770-44764770 on Build GRCh38
rs1049968850 Chr.11: 44764422-44764422 on Build GRCh38
rs891485475 Chr.11: 44764595-44764595 on Build GRCh38
rs776468143 Chr.11: 44764607-44764607 on Build GRCh38
rs560586666 Chr.11: 44764539-44764539 on Build GRCh38
rs917404394 Chr.11: 44764522-44764522 on Build GRCh38
rs920139081 Chr.11: 44764396-44764396 on Build GRCh38
rs891541354 Chr.11: 44764476-44764476 on Build GRCh38
rs891507170 Chr.11: 44764671-44764671 on Build GRCh38

View Full Product Details