Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 44917885-44918379 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSPAN18
Entrez Gene ID
90139
Gene Name
tetraspanin 18
Gene Aliases
TSPAN
UniGene
Hs.385634
Gene Chromosome Location
Chr.11: 44727148-44932427 on Build GRCh38
Transcripts
NM_130783.4 XM_005253217.3 XM_006718372.3 XM_006718373.3 XM_011520459.2 XM_017018530.1 XM_017018531.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6810630 c.319A>G p.I107V Chr.11: 44918032-44918032 on Build GRCh38
3447369 c.328G>A p.E110K Chr.11: 44918041-44918041 on Build GRCh38
6551942 c.309C>T p.I103I Chr.11: 44918022-44918022 on Build GRCh38
7423422 c.307A>T p.I103F Chr.11: 44918020-44918020 on Build GRCh38
4958734 c.327G>A p.R109R Chr.11: 44918040-44918040 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs940078441 Chr.11: 44917918-44917918 on Build GRCh38
rs112859596 Chr.11: 44918290-44918290 on Build GRCh38
rs867205264 Chr.11: 44918219-44918219 on Build GRCh38
rs369703331 Chr.11: 44917956-44917956 on Build GRCh38
rs775184541 Chr.11: 44917953-44917953 on Build GRCh38
rs1008585778 Chr.11: 44918166-44918166 on Build GRCh38
rs1037584198 Chr.11: 44918064-44918064 on Build GRCh38
rs34284182 Chr.11: 44918208-44918208 on Build GRCh38
rs750206775 Chr.11: 44917966-44917966 on Build GRCh38
rs563176060 Chr.11: 44918324-44918324 on Build GRCh38
rs760184491 Chr.11: 44917945-44917945 on Build GRCh38
rs372957209 Chr.11: 44918053-44918053 on Build GRCh38
rs779474763 Chr.11: 44918050-44918050 on Build GRCh38
rs372718144 Chr.11: 44918221-44918221 on Build GRCh38
rs983316937 Chr.11: 44918231-44918231 on Build GRCh38
rs557381543 Chr.11: 44918194-44918194 on Build GRCh38
rs749475454 Chr.11: 44917927-44917927 on Build GRCh38
rs753870147 Chr.11: 44918329-44918329 on Build GRCh38
rs377070723 Chr.11: 44918082-44918082 on Build GRCh38
rs776165997 Chr.11: 44918068-44918068 on Build GRCh38
rs779934951 Chr.11: 44917985-44917985 on Build GRCh38
rs749473945 Chr.11: 44918034-44918034 on Build GRCh38
rs772687913 Chr.11: 44917951-44917951 on Build GRCh38
rs199651174 Chr.11: 44918070-44918070 on Build GRCh38
rs3816791 Chr.11: 44918343-44918343 on Build GRCh38
rs1019057574 Chr.11: 44918177-44918177 on Build GRCh38
rs375494499 Chr.11: 44917923-44917923 on Build GRCh38
rs904809689 Chr.11: 44918351-44918351 on Build GRCh38
rs932688244 Chr.11: 44918101-44918101 on Build GRCh38
rs917257668 Chr.11: 44918339-44918339 on Build GRCh38
rs772528545 Chr.11: 44918060-44918060 on Build GRCh38
rs1035345066 Chr.11: 44918314-44918314 on Build GRCh38
rs765009338 Chr.11: 44917964-44917964 on Build GRCh38
rs1049747964 Chr.11: 44918113-44918113 on Build GRCh38
rs540848681 Chr.11: 44918193-44918193 on Build GRCh38
rs975081553 Chr.11: 44918191-44918191 on Build GRCh38
rs775668144 Chr.11: 44917988-44917988 on Build GRCh38
rs534336642 Chr.11: 44918097-44918097 on Build GRCh38
rs142957647 Chr.11: 44917989-44917989 on Build GRCh38
rs762645858 Chr.11: 44918121-44918121 on Build GRCh38
rs907706357 Chr.11: 44918283-44918283 on Build GRCh38
rs577204416 Chr.11: 44917913-44917913 on Build GRCh38
rs1008296349 Chr.11: 44918171-44918171 on Build GRCh38
rs577675140 Chr.11: 44918282-44918282 on Build GRCh38
rs1021863000 Chr.11: 44917919-44917919 on Build GRCh38
rs756169586 Chr.11: 44918010-44918010 on Build GRCh38
rs983235549 Chr.11: 44918330-44918330 on Build GRCh38
rs955831218 Chr.11: 44918115-44918115 on Build GRCh38
rs369062337 Chr.11: 44918020-44918020 on Build GRCh38
rs36104013 Chr.11: 44917945-44917945 on Build GRCh38
rs373104718 Chr.11: 44917960-44917960 on Build GRCh38
rs771210653 Chr.11: 44917938-44917938 on Build GRCh38
rs761463807 Chr.11: 44917958-44917958 on Build GRCh38
rs187088925 Chr.11: 44918348-44918348 on Build GRCh38
rs774707026 Chr.11: 44917941-44917941 on Build GRCh38
rs3816790 Chr.11: 44918331-44918331 on Build GRCh38
rs182505582 Chr.11: 44918294-44918294 on Build GRCh38
rs879380035 Chr.11: 44918218-44918218 on Build GRCh38
rs760931439 Chr.11: 44918173-44918173 on Build GRCh38
rs559088528 Chr.11: 44918205-44918205 on Build GRCh38
rs376320762 Chr.11: 44917975-44917975 on Build GRCh38
rs771176141 Chr.11: 44918049-44918049 on Build GRCh38
rs959814658 Chr.11: 44917962-44917962 on Build GRCh38
rs34998834 Chr.11: 44917918-44917918 on Build GRCh38
rs375283466 Chr.11: 44918188-44918188 on Build GRCh38

View Full Product Details