Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 46369390-46369881 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGKZ
Entrez Gene ID
8525
Gene Name
diacylglycerol kinase zeta
Gene Aliases
DAGK5, DAGK6, DGK-ZETA, hDGKzeta
UniGene
Hs.502461
Gene Chromosome Location
Chr.11: 46332905-46380554 on Build GRCh38
Transcripts
NM_001105540.1 NM_001199266.1 NM_001199267.1 NM_001199268.1 NM_003646.3 NM_201532.2 NM_201533.3 XM_005253181.3 XM_005253182.3 XM_006718354.1 XM_006718355.2 XM_011520422.1 XM_011520423.1 XM_017018455.1 XM_017018456.1 XM_017018457.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
927149 c.1057G>A p.V353I Chr.11: 46369542-46369542 on Build GRCh38
4165791 c.462G>T p.K154N Chr.11: 46369511-46369511 on Build GRCh38
6448598 c.347A>G p.N116S Chr.11: 46369498-46369498 on Build GRCh38
3739063 c.389A>G p.N130S Chr.11: 46369540-46369540 on Build GRCh38
3739062 c.491A>G p.N164S Chr.11: 46369540-46369540 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759019331 Chr.11: 46369425-46369425 on Build GRCh38
rs573795420 Chr.11: 46369697-46369697 on Build GRCh38
rs988356665 Chr.11: 46369658-46369658 on Build GRCh38
rs1014887849 Chr.11: 46369741-46369741 on Build GRCh38
rs958071706 Chr.11: 46369787-46369787 on Build GRCh38
rs537912059 Chr.11: 46369645-46369645 on Build GRCh38
rs371937492 Chr.11: 46369624-46369624 on Build GRCh38
rs901616928 Chr.11: 46369694-46369694 on Build GRCh38
rs759870827 Chr.11: 46369439-46369443 on Build GRCh38
rs936893401 Chr.11: 46369649-46369649 on Build GRCh38
rs540738165 Chr.11: 46369725-46369725 on Build GRCh38
rs1050064989 Chr.11: 46369707-46369707 on Build GRCh38
rs202018019 Chr.11: 46369503-46369503 on Build GRCh38
rs778100454 Chr.11: 46369514-46369514 on Build GRCh38
rs368761340 Chr.11: 46369583-46369583 on Build GRCh38
rs988346837 Chr.11: 46369825-46369825 on Build GRCh38
rs771992578 Chr.11: 46369517-46369517 on Build GRCh38
rs760047840 Chr.11: 46369721-46369721 on Build GRCh38
rs200877028 Chr.11: 46369548-46369548 on Build GRCh38
rs372720290 Chr.11: 46369448-46369448 on Build GRCh38
rs542793219 Chr.11: 46369763-46369763 on Build GRCh38
rs374240578 Chr.11: 46369573-46369573 on Build GRCh38
rs1006468127 Chr.11: 46369827-46369827 on Build GRCh38
rs748179735 Chr.11: 46369504-46369504 on Build GRCh38
rs373519977 Chr.11: 46369615-46369615 on Build GRCh38
rs35267642 Chr.11: 46369547-46369547 on Build GRCh38
rs750116491 Chr.11: 46369593-46369593 on Build GRCh38
rs944244930 Chr.11: 46369683-46369683 on Build GRCh38
rs760134267 Chr.11: 46369562-46369562 on Build GRCh38
rs528630358 Chr.11: 46369691-46369691 on Build GRCh38
rs763682728 Chr.11: 46369445-46369445 on Build GRCh38
rs992647986 Chr.11: 46369612-46369612 on Build GRCh38
rs757041070 Chr.11: 46369449-46369449 on Build GRCh38
rs761568294 Chr.11: 46369582-46369582 on Build GRCh38
rs771373804 Chr.11: 46369420-46369420 on Build GRCh38
rs760328609 Chr.11: 46369431-46369431 on Build GRCh38
rs980817881 Chr.11: 46369641-46369641 on Build GRCh38
rs1013447878 Chr.11: 46369760-46369760 on Build GRCh38
rs887977795 Chr.11: 46369797-46369798 on Build GRCh38
rs897652739 Chr.11: 46369742-46369742 on Build GRCh38
rs557315025 Chr.11: 46369596-46369596 on Build GRCh38
rs887450137 Chr.11: 46369715-46369715 on Build GRCh38
rs774891343 Chr.11: 46369424-46369424 on Build GRCh38
rs151034874 Chr.11: 46369786-46369786 on Build GRCh38
rs767043543 Chr.11: 46369426-46369426 on Build GRCh38
rs976924204 Chr.11: 46369843-46369843 on Build GRCh38
rs778761376 Chr.11: 46369457-46369457 on Build GRCh38
rs781423874 Chr.11: 46369494-46369494 on Build GRCh38
rs1029332877 Chr.11: 46369618-46369618 on Build GRCh38
rs370663840 Chr.11: 46369546-46369546 on Build GRCh38
rs750357277 Chr.11: 46369462-46369462 on Build GRCh38
rs75106374 Chr.11: 46369511-46369511 on Build GRCh38
rs76712654 Chr.11: 46369482-46369482 on Build GRCh38
rs553332708 Chr.11: 46369781-46369781 on Build GRCh38
rs928677317 Chr.11: 46369770-46369770 on Build GRCh38
rs113036131 Chr.11: 46369820-46369820 on Build GRCh38
rs765481690 Chr.11: 46369575-46369576 on Build GRCh38
rs113231110 Chr.11: 46369439-46369439 on Build GRCh38
rs779969600 Chr.11: 46369616-46369616 on Build GRCh38
rs529201013 Chr.11: 46369858-46369858 on Build GRCh38
rs760190097 Chr.11: 46369794-46369794 on Build GRCh38
rs774847235 Chr.11: 46369714-46369714 on Build GRCh38
rs569025084 Chr.11: 46369545-46369545 on Build GRCh38
rs569614750 Chr.11: 46369842-46369842 on Build GRCh38
rs543404986 Chr.11: 46369561-46369561 on Build GRCh38
rs776241005 Chr.11: 46369581-46369581 on Build GRCh38
rs373755319 Chr.11: 46369609-46369609 on Build GRCh38
rs140990488 Chr.11: 46369828-46369828 on Build GRCh38
rs75463289 Chr.11: 46369668-46369668 on Build GRCh38
rs969473869 Chr.11: 46369767-46369767 on Build GRCh38

View Full Product Details