Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 56159380-56159895 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR8K5
Entrez Gene ID
219453
Gene Name
olfactory receptor family 8 subfamily K member 5
Gene Aliases
OR11-174
UniGene
Hs.553626
Gene Chromosome Location
Chr.11: 56159394-56160317 on Build GRCh38
Transcripts
NM_001004058.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
313799 c.479C>G p.T160S Chr.11: 56159839-56159839 on Build GRCh38
6735396 c.843A>G p.V281V Chr.11: 56159475-56159475 on Build GRCh38
928260 c.683T>C p.M228T Chr.11: 56159635-56159635 on Build GRCh38
4799735 c.597C>A p.S199R Chr.11: 56159721-56159721 on Build GRCh38
1321614 c.709G>T p.A237S Chr.11: 56159609-56159609 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754976961 Chr.11: 56159651-56159651 on Build GRCh38
rs776222859 Chr.11: 56159796-56159796 on Build GRCh38
rs927221622 Chr.11: 56159689-56159689 on Build GRCh38
rs752069339 Chr.11: 56159642-56159642 on Build GRCh38
rs371380657 Chr.11: 56159806-56159806 on Build GRCh38
rs984798254 Chr.11: 56159662-56159662 on Build GRCh38
rs199816778 Chr.11: 56159712-56159712 on Build GRCh38
rs1048792978 Chr.11: 56159446-56159446 on Build GRCh38
rs754723153 Chr.11: 56159451-56159451 on Build GRCh38
rs201567440 Chr.11: 56159801-56159801 on Build GRCh38
rs747152095 Chr.11: 56159573-56159573 on Build GRCh38
rs766412336 Chr.11: 56159616-56159616 on Build GRCh38
rs374278118 Chr.11: 56159452-56159452 on Build GRCh38
rs769423741 Chr.11: 56159581-56159581 on Build GRCh38
rs772152342 Chr.11: 56159411-56159411 on Build GRCh38
rs760911007 Chr.11: 56159413-56159413 on Build GRCh38
rs761807276 Chr.11: 56159434-56159434 on Build GRCh38
rs75240067 Chr.11: 56159563-56159563 on Build GRCh38
rs774741509 Chr.11: 56159809-56159809 on Build GRCh38
rs1043395952 Chr.11: 56159816-56159816 on Build GRCh38
rs772114197 Chr.11: 56159776-56159776 on Build GRCh38
rs759607853 Chr.11: 56159621-56159622 on Build GRCh38
rs772325896 Chr.11: 56159435-56159435 on Build GRCh38
rs749204201 Chr.11: 56159582-56159582 on Build GRCh38
rs370391770 Chr.11: 56159835-56159835 on Build GRCh38
rs953374889 Chr.11: 56159583-56159583 on Build GRCh38
rs781208303 Chr.11: 56159577-56159577 on Build GRCh38
rs765488797 Chr.11: 56159630-56159630 on Build GRCh38
rs766764772 Chr.11: 56159723-56159723 on Build GRCh38
rs141718394 Chr.11: 56159408-56159408 on Build GRCh38
rs759256070 Chr.11: 56159532-56159532 on Build GRCh38
rs750908770 Chr.11: 56159550-56159550 on Build GRCh38
rs760134796 Chr.11: 56159730-56159730 on Build GRCh38
rs148293568 Chr.11: 56159614-56159614 on Build GRCh38
rs138516468 Chr.11: 56159480-56159480 on Build GRCh38
rs748437830 Chr.11: 56159578-56159578 on Build GRCh38
rs757496765 Chr.11: 56159538-56159538 on Build GRCh38
rs775649734 Chr.11: 56159412-56159412 on Build GRCh38
rs573633126 Chr.11: 56159629-56159629 on Build GRCh38
rs746467434 Chr.11: 56159783-56159783 on Build GRCh38
rs918381243 Chr.11: 56159668-56159668 on Build GRCh38
rs778081855 Chr.11: 56159758-56159758 on Build GRCh38
rs753735706 Chr.11: 56159620-56159621 on Build GRCh38
rs766014059 Chr.11: 56159445-56159445 on Build GRCh38
rs767058017 Chr.11: 56159619-56159619 on Build GRCh38
rs931272029 Chr.11: 56159444-56159444 on Build GRCh38
rs780105706 Chr.11: 56159560-56159560 on Build GRCh38
rs866693001 Chr.11: 56159601-56159601 on Build GRCh38
rs780157192 Chr.11: 56159496-56159496 on Build GRCh38
rs757077659 Chr.11: 56159763-56159763 on Build GRCh38
rs867472909 Chr.11: 56159471-56159471 on Build GRCh38
rs772978988 Chr.11: 56159522-56159522 on Build GRCh38
rs763239095 Chr.11: 56159720-56159720 on Build GRCh38
rs138593758 Chr.11: 56159406-56159406 on Build GRCh38
rs760697247 Chr.11: 56159614-56159619 on Build GRCh38
rs569114519 Chr.11: 56159870-56159870 on Build GRCh38
rs749151515 Chr.11: 56159671-56159671 on Build GRCh38
rs766816017 Chr.11: 56159641-56159641 on Build GRCh38
rs369678334 Chr.11: 56159527-56159527 on Build GRCh38
rs770861865 Chr.11: 56159676-56159676 on Build GRCh38
rs370002672 Chr.11: 56159576-56159576 on Build GRCh38
rs769640092 Chr.11: 56159512-56159512 on Build GRCh38
rs201996541 Chr.11: 56159497-56159497 on Build GRCh38
rs193229624 Chr.11: 56159598-56159598 on Build GRCh38
rs767461136 Chr.11: 56159732-56159732 on Build GRCh38
rs752886343 Chr.11: 56159738-56159738 on Build GRCh38
rs201041122 Chr.11: 56159724-56159724 on Build GRCh38
rs916261979 Chr.11: 56159734-56159734 on Build GRCh38
rs771461674 Chr.11: 56159682-56159682 on Build GRCh38
rs550530626 Chr.11: 56159635-56159635 on Build GRCh38
rs1018974825 Chr.11: 56159616-56159616 on Build GRCh38
rs546979930 Chr.11: 56159537-56159537 on Build GRCh38
rs374291964 Chr.11: 56159657-56159657 on Build GRCh38
rs778873014 Chr.11: 56159770-56159770 on Build GRCh38
rs376998905 Chr.11: 56159535-56159535 on Build GRCh38
rs776281961 Chr.11: 56159714-56159714 on Build GRCh38
rs141487223 Chr.11: 56159621-56159621 on Build GRCh38
rs745761568 Chr.11: 56159771-56159771 on Build GRCh38
rs200083825 Chr.11: 56159787-56159787 on Build GRCh38
rs776476824 Chr.11: 56159505-56159505 on Build GRCh38
rs143262863 Chr.11: 56159552-56159552 on Build GRCh38
rs756396007 Chr.11: 56159664-56159664 on Build GRCh38
rs757767631 Chr.11: 56159470-56159470 on Build GRCh38
rs751205072 Chr.11: 56159448-56159448 on Build GRCh38
rs754219929 Chr.11: 56159464-56159464 on Build GRCh38
rs778926147 Chr.11: 56159677-56159677 on Build GRCh38
rs765265830 Chr.11: 56159793-56159801 on Build GRCh38
rs747843505 Chr.11: 56159506-56159506 on Build GRCh38
rs376114385 Chr.11: 56159435-56159435 on Build GRCh38
rs773431761 Chr.11: 56159558-56159566 on Build GRCh38
rs61746296 Chr.11: 56159523-56159523 on Build GRCh38
rs199985785 Chr.11: 56159820-56159820 on Build GRCh38
rs778249386 Chr.11: 56159458-56159458 on Build GRCh38
rs185682297 Chr.11: 56159824-56159824 on Build GRCh38
rs781254378 Chr.11: 56159587-56159587 on Build GRCh38
rs779229915 Chr.11: 56159472-56159472 on Build GRCh38
rs984027825 Chr.11: 56159681-56159681 on Build GRCh38
rs74535912 Chr.11: 56159572-56159572 on Build GRCh38
rs754116030 Chr.11: 56159759-56159759 on Build GRCh38
rs373590873 Chr.11: 56159534-56159534 on Build GRCh38
rs765656548 Chr.11: 56159545-56159545 on Build GRCh38
rs1056859901 Chr.11: 56159524-56159524 on Build GRCh38
rs768418948 Chr.11: 56159419-56159419 on Build GRCh38
rs779415220 Chr.11: 56159782-56159782 on Build GRCh38
rs200952940 Chr.11: 56159834-56159834 on Build GRCh38
rs781350633 Chr.11: 56159655-56159655 on Build GRCh38
rs756274085 Chr.11: 56159739-56159739 on Build GRCh38
rs377710232 Chr.11: 56159633-56159633 on Build GRCh38
rs138027817 Chr.11: 56159679-56159679 on Build GRCh38
rs760242150 Chr.11: 56159687-56159687 on Build GRCh38
rs774041609 Chr.11: 56159531-56159531 on Build GRCh38
rs562202949 Chr.11: 56159818-56159818 on Build GRCh38
rs377462769 Chr.11: 56159830-56159830 on Build GRCh38
rs777704496 Chr.11: 56159666-56159666 on Build GRCh38
rs757592955 Chr.11: 56159871-56159871 on Build GRCh38
rs768415577 Chr.11: 56159704-56159704 on Build GRCh38
rs774965266 Chr.11: 56159622-56159622 on Build GRCh38
rs147115733 Chr.11: 56159436-56159436 on Build GRCh38
rs776333511 Chr.11: 56159426-56159426 on Build GRCh38

View Full Product Details