Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 56177214-56177726 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR5J2
Entrez Gene ID
282775
Gene Name
olfactory receptor family 5 subfamily J member 2
Gene Aliases
OR11-266
UniGene
Hs.537145
Gene Chromosome Location
Chr.11: 56176618-56177556 on Build GRCh38
Transcripts
NM_001005492.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6537351 c.836C>G p.T279R Chr.11: 56177453-56177453 on Build GRCh38
7245692 c.680G>C p.R227T Chr.11: 56177297-56177297 on Build GRCh38
313730 c.728C>T p.S243F Chr.11: 56177345-56177345 on Build GRCh38
5819369 c.880A>T p.N294Y Chr.11: 56177497-56177497 on Build GRCh38
6552061 c.758G>T p.G253V Chr.11: 56177375-56177375 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs536548848 Chr.11: 56177339-56177339 on Build GRCh38
rs144489025 Chr.11: 56177550-56177550 on Build GRCh38
rs757880597 Chr.11: 56177376-56177376 on Build GRCh38
rs1045874125 Chr.11: 56177578-56177578 on Build GRCh38
rs558712266 Chr.11: 56177508-56177508 on Build GRCh38
rs142761104 Chr.11: 56177315-56177315 on Build GRCh38
rs760717579 Chr.11: 56177332-56177332 on Build GRCh38
rs745595664 Chr.11: 56177316-56177316 on Build GRCh38
rs776840586 Chr.11: 56177484-56177485 on Build GRCh38
rs530836972 Chr.11: 56177699-56177699 on Build GRCh38
rs755394606 Chr.11: 56177287-56177287 on Build GRCh38
rs961561145 Chr.11: 56177270-56177270 on Build GRCh38
rs982687313 Chr.11: 56177499-56177499 on Build GRCh38
rs752634029 Chr.11: 56177568-56177569 on Build GRCh38
rs941097033 Chr.11: 56177244-56177244 on Build GRCh38
rs534012364 Chr.11: 56177529-56177529 on Build GRCh38
rs757279588 Chr.11: 56177455-56177455 on Build GRCh38
rs753110839 Chr.11: 56177357-56177357 on Build GRCh38
rs760964840 Chr.11: 56177453-56177453 on Build GRCh38
rs766583747 Chr.11: 56177559-56177559 on Build GRCh38
rs745557121 Chr.11: 56177607-56177607 on Build GRCh38
rs751185697 Chr.11: 56177580-56177580 on Build GRCh38
rs780985237 Chr.11: 56177616-56177616 on Build GRCh38
rs766836725 Chr.11: 56177277-56177277 on Build GRCh38
rs139886298 Chr.11: 56177384-56177384 on Build GRCh38
rs563607151 Chr.11: 56177591-56177591 on Build GRCh38
rs199679290 Chr.11: 56177291-56177291 on Build GRCh38
rs757338200 Chr.11: 56177378-56177378 on Build GRCh38
rs762348814 Chr.11: 56177543-56177543 on Build GRCh38
rs768579526 Chr.11: 56177394-56177394 on Build GRCh38
rs868232221 Chr.11: 56177428-56177428 on Build GRCh38
rs763684226 Chr.11: 56177241-56177241 on Build GRCh38
rs746751233 Chr.11: 56177503-56177503 on Build GRCh38
rs765299056 Chr.11: 56177548-56177548 on Build GRCh38
rs781507139 Chr.11: 56177289-56177289 on Build GRCh38
rs767970353 Chr.11: 56177356-56177356 on Build GRCh38
rs777332064 Chr.11: 56177425-56177425 on Build GRCh38
rs776338136 Chr.11: 56177337-56177337 on Build GRCh38
rs866231331 Chr.11: 56177334-56177334 on Build GRCh38
rs1035526095 Chr.11: 56177686-56177686 on Build GRCh38
rs900892601 Chr.11: 56177511-56177511 on Build GRCh38
rs1039452756 Chr.11: 56177700-56177700 on Build GRCh38
rs915845433 Chr.11: 56177340-56177340 on Build GRCh38
rs775502945 Chr.11: 56177237-56177237 on Build GRCh38
rs759660929 Chr.11: 56177541-56177542 on Build GRCh38
rs762502405 Chr.11: 56177514-56177514 on Build GRCh38
rs149835720 Chr.11: 56177405-56177405 on Build GRCh38
rs1029923609 Chr.11: 56177471-56177471 on Build GRCh38
rs758411507 Chr.11: 56177551-56177551 on Build GRCh38
rs376537675 Chr.11: 56177565-56177565 on Build GRCh38
rs780151820 Chr.11: 56177501-56177501 on Build GRCh38
rs916057433 Chr.11: 56177256-56177256 on Build GRCh38
rs548534792 Chr.11: 56177323-56177323 on Build GRCh38
rs756180935 Chr.11: 56177579-56177579 on Build GRCh38
rs139773791 Chr.11: 56177542-56177542 on Build GRCh38
rs994311501 Chr.11: 56177674-56177674 on Build GRCh38
rs752261661 Chr.11: 56177375-56177375 on Build GRCh38
rs745807561 Chr.11: 56177618-56177618 on Build GRCh38
rs772974358 Chr.11: 56177419-56177419 on Build GRCh38
rs867286735 Chr.11: 56177575-56177575 on Build GRCh38
rs142214796 Chr.11: 56177505-56177505 on Build GRCh38
rs748141067 Chr.11: 56177506-56177506 on Build GRCh38
rs757035260 Chr.11: 56177272-56177272 on Build GRCh38
rs893137551 Chr.11: 56177667-56177667 on Build GRCh38
rs909570306 Chr.11: 56177242-56177242 on Build GRCh38
rs779964381 Chr.11: 56177390-56177390 on Build GRCh38
rs541890586 Chr.11: 56177605-56177605 on Build GRCh38
rs748768888 Chr.11: 56177585-56177585 on Build GRCh38
rs764523350 Chr.11: 56177454-56177454 on Build GRCh38
rs868086623 Chr.11: 56177526-56177526 on Build GRCh38
rs760890519 Chr.11: 56177341-56177342 on Build GRCh38
rs189953005 Chr.11: 56177562-56177562 on Build GRCh38
rs140282757 Chr.11: 56177303-56177303 on Build GRCh38
rs1017185038 Chr.11: 56177603-56177603 on Build GRCh38
rs756713357 Chr.11: 56177368-56177368 on Build GRCh38
rs762873170 Chr.11: 56177584-56177603 on Build GRCh38
rs760064405 Chr.11: 56177240-56177240 on Build GRCh38
rs1055499203 Chr.11: 56177245-56177245 on Build GRCh38
rs753399221 Chr.11: 56177265-56177265 on Build GRCh38
rs765314988 Chr.11: 56177548-56177549 on Build GRCh38
rs771934315 Chr.11: 56177321-56177321 on Build GRCh38
rs750127531 Chr.11: 56177343-56177343 on Build GRCh38
rs903404369 Chr.11: 56177294-56177294 on Build GRCh38
rs755244934 Chr.11: 56177354-56177354 on Build GRCh38
rs868722629 Chr.11: 56177293-56177293 on Build GRCh38
rs376303545 Chr.11: 56177417-56177417 on Build GRCh38
rs865779239 Chr.11: 56177475-56177475 on Build GRCh38
rs750615086 Chr.11: 56177490-56177490 on Build GRCh38
rs201477399 Chr.11: 56177338-56177338 on Build GRCh38
rs150737659 Chr.11: 56177371-56177371 on Build GRCh38
rs554524924 Chr.11: 56177383-56177383 on Build GRCh38
rs984356551 Chr.11: 56177247-56177247 on Build GRCh38
rs756122621 Chr.11: 56177301-56177301 on Build GRCh38
rs771204046 Chr.11: 56177312-56177312 on Build GRCh38
rs748091693 Chr.11: 56177571-56177571 on Build GRCh38
rs751914437 Chr.11: 56177280-56177280 on Build GRCh38
rs753166632 Chr.11: 56177448-56177448 on Build GRCh38
rs147148092 Chr.11: 56177302-56177302 on Build GRCh38
rs763505261 Chr.11: 56177333-56177333 on Build GRCh38
rs542520753 Chr.11: 56177587-56177587 on Build GRCh38
rs568231265 Chr.11: 56177327-56177327 on Build GRCh38
rs370730952 Chr.11: 56177450-56177450 on Build GRCh38
rs758702708 Chr.11: 56177500-56177500 on Build GRCh38
rs777870080 Chr.11: 56177582-56177582 on Build GRCh38
rs764464449 Chr.11: 56177538-56177538 on Build GRCh38
rs576997052 Chr.11: 56177515-56177515 on Build GRCh38
rs757025813 Chr.11: 56177613-56177613 on Build GRCh38
rs140193705 Chr.11: 56177412-56177412 on Build GRCh38
rs141667991 Chr.11: 56177615-56177615 on Build GRCh38
rs541303401 Chr.11: 56177525-56177525 on Build GRCh38
rs1046323612 Chr.11: 56177264-56177264 on Build GRCh38
rs950287067 Chr.11: 56177263-56177263 on Build GRCh38
rs762704004 Chr.11: 56177426-56177426 on Build GRCh38
rs771048735 Chr.11: 56177389-56177389 on Build GRCh38
rs779014585 Chr.11: 56177486-56177486 on Build GRCh38

View Full Product Details