Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 57649607-57650104 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YPEL4
Entrez Gene ID
219539
Gene Name
yippee like 4
Gene Aliases
-
UniGene
Hs.135978
Gene Chromosome Location
Chr.11: 57645087-57649944 on Build GRCh38
Transcripts
NM_145008.2 XM_017017318.1

SNP Detail

SNP ID Chromosome Location
rs572799184 Chr.11: 57649887-57649887 on Build GRCh38
rs560704216 Chr.11: 57649867-57649867 on Build GRCh38
rs879465994 Chr.11: 57649978-57649978 on Build GRCh38
rs906261937 Chr.11: 57650086-57650086 on Build GRCh38
rs958769273 Chr.11: 57649727-57649734 on Build GRCh38
rs956225305 Chr.11: 57649714-57649714 on Build GRCh38
rs1022018889 Chr.11: 57650047-57650047 on Build GRCh38
rs983265668 Chr.11: 57649913-57649913 on Build GRCh38
rs1048495082 Chr.11: 57649925-57649925 on Build GRCh38
rs780019076 Chr.11: 57650084-57650084 on Build GRCh38
rs1006449013 Chr.11: 57649962-57649962 on Build GRCh38
rs981548006 Chr.11: 57649722-57649722 on Build GRCh38
rs888052448 Chr.11: 57649958-57649958 on Build GRCh38
rs560879279 Chr.11: 57649966-57649966 on Build GRCh38
rs536557120 Chr.11: 57649968-57649968 on Build GRCh38
rs567611183 Chr.11: 57649864-57649864 on Build GRCh38
rs964702791 Chr.11: 57649797-57649797 on Build GRCh38
rs1021839919 Chr.11: 57649788-57649788 on Build GRCh38
rs943853787 Chr.11: 57649874-57649874 on Build GRCh38
rs990332694 Chr.11: 57649757-57649757 on Build GRCh38
rs560875737 Chr.11: 57649972-57649972 on Build GRCh38
rs1047973557 Chr.11: 57649803-57649803 on Build GRCh38
rs962133762 Chr.11: 57649717-57649717 on Build GRCh38
rs929334041 Chr.11: 57649791-57649791 on Build GRCh38
rs1006396222 Chr.11: 57649715-57649718 on Build GRCh38
rs941858280 Chr.11: 57650051-57650051 on Build GRCh38
rs1039192041 Chr.11: 57649822-57649822 on Build GRCh38
rs1035774839 Chr.11: 57649726-57649733 on Build GRCh38
rs370368614 Chr.11: 57649715-57649716 on Build GRCh38
rs933431279 Chr.11: 57649911-57649911 on Build GRCh38
rs959005015 Chr.11: 57650018-57650018 on Build GRCh38
rs564809349 Chr.11: 57650074-57650074 on Build GRCh38
rs991865858 Chr.11: 57649728-57649728 on Build GRCh38
rs532195755 Chr.11: 57650083-57650083 on Build GRCh38
rs888050932 Chr.11: 57649807-57649807 on Build GRCh38
rs907688252 Chr.11: 57650031-57650031 on Build GRCh38
rs543298066 Chr.11: 57649959-57649959 on Build GRCh38
rs975188529 Chr.11: 57649883-57649883 on Build GRCh38
rs991526395 Chr.11: 57650023-57650023 on Build GRCh38
rs977241157 Chr.11: 57649868-57649868 on Build GRCh38
rs549651959 Chr.11: 57649966-57649966 on Build GRCh38
rs775533080 Chr.11: 57649656-57649656 on Build GRCh38
rs996911143 Chr.11: 57649714-57649715 on Build GRCh38
rs914499447 Chr.11: 57649740-57649740 on Build GRCh38
rs543592898 Chr.11: 57650008-57650008 on Build GRCh38
rs756038084 Chr.11: 57650068-57650068 on Build GRCh38
rs1015075825 Chr.11: 57649715-57649722 on Build GRCh38
rs187958279 Chr.11: 57649992-57649992 on Build GRCh38
rs1035788259 Chr.11: 57650009-57650009 on Build GRCh38
rs1037590655 Chr.11: 57650065-57650065 on Build GRCh38
rs942191346 Chr.11: 57649814-57649814 on Build GRCh38
rs932341624 Chr.11: 57649916-57649917 on Build GRCh38

View Full Product Details