Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 57740585-57741121 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMX2
Entrez Gene ID
51075
Gene Name
thioredoxin related transmembrane protein 2
Gene Aliases
CGI-31, PDIA12, PIG26, TXNDC14
UniGene
Hs.654965
Gene Chromosome Location
Chr.11: 57712523-57740973 on Build GRCh38
Transcripts
NM_001144012.2 NM_015959.3 NR_026593.2 NR_037645.1 NR_037646.1

Gene Symbol
TMX2-CTNND1
Entrez Gene ID
100528016
Gene Name
TMX2-CTNND1 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.166011
Gene Chromosome Location
Chr.11: 57712523-57819180 on Build GRCh38
Transcripts
NM_001144012.2 NM_015959.3 NR_026593.2 NR_037645.1 NR_037646.1

SNP Detail

SNP ID Chromosome Location
rs1007093643 Chr.11: 57740838-57740838 on Build GRCh38
rs865889414 Chr.11: 57740927-57740927 on Build GRCh38
rs1058118 Chr.11: 57740910-57740910 on Build GRCh38
rs1019658738 Chr.11: 57740655-57740655 on Build GRCh38
rs943064842 Chr.11: 57740926-57740926 on Build GRCh38
rs1032361144 Chr.11: 57741055-57741055 on Build GRCh38
rs569918685 Chr.11: 57740711-57740711 on Build GRCh38
rs1025655820 Chr.11: 57740876-57740876 on Build GRCh38
rs1041472259 Chr.11: 57740806-57740806 on Build GRCh38
rs1041498933 Chr.11: 57740973-57740973 on Build GRCh38
rs1025470554 Chr.11: 57740779-57740779 on Build GRCh38
rs565879085 Chr.11: 57740938-57740938 on Build GRCh38
rs969454358 Chr.11: 57740800-57740800 on Build GRCh38
rs1025731767 Chr.11: 57741098-57741098 on Build GRCh38
rs933115440 Chr.11: 57740901-57740901 on Build GRCh38
rs972784712 Chr.11: 57740674-57740674 on Build GRCh38
rs184381132 Chr.11: 57741076-57741076 on Build GRCh38
rs887233949 Chr.11: 57740774-57740774 on Build GRCh38
rs1041262277 Chr.11: 57741010-57741010 on Build GRCh38
rs918448264 Chr.11: 57740706-57740706 on Build GRCh38
rs761908688 Chr.11: 57740941-57740941 on Build GRCh38
rs965336861 Chr.11: 57740845-57740845 on Build GRCh38
rs937025734 Chr.11: 57740746-57740746 on Build GRCh38
rs531058332 Chr.11: 57740737-57740737 on Build GRCh38
rs187788066 Chr.11: 57740707-57740707 on Build GRCh38
rs972815767 Chr.11: 57740853-57740853 on Build GRCh38
rs552684929 Chr.11: 57740773-57740773 on Build GRCh38
rs1002258764 Chr.11: 57741022-57741022 on Build GRCh38
rs1013677043 Chr.11: 57740775-57740776 on Build GRCh38
rs952583690 Chr.11: 57740900-57740900 on Build GRCh38
rs1056859412 Chr.11: 57740755-57740755 on Build GRCh38
rs936512087 Chr.11: 57740664-57740664 on Build GRCh38
rs535732444 Chr.11: 57740944-57740944 on Build GRCh38
rs965472323 Chr.11: 57740667-57740667 on Build GRCh38
rs1001410718 Chr.11: 57740822-57740822 on Build GRCh38
rs1039974792 Chr.11: 57740733-57740733 on Build GRCh38
rs115411262 Chr.11: 57740907-57740907 on Build GRCh38
rs931566629 Chr.11: 57740943-57740943 on Build GRCh38
rs192167900 Chr.11: 57741074-57741074 on Build GRCh38
rs954083330 Chr.11: 57740642-57740642 on Build GRCh38
rs957985771 Chr.11: 57741073-57741073 on Build GRCh38
rs888796145 Chr.11: 57740712-57740712 on Build GRCh38
rs557422744 Chr.11: 57741045-57741045 on Build GRCh38
rs14996 Chr.11: 57740848-57740848 on Build GRCh38
rs188669680 Chr.11: 57740983-57740983 on Build GRCh38
rs765154468 Chr.11: 57740783-57740784 on Build GRCh38
rs112095568 Chr.11: 57740968-57740968 on Build GRCh38
rs768975715 Chr.11: 57740748-57740748 on Build GRCh38
rs1032981320 Chr.11: 57740622-57740622 on Build GRCh38
rs1047267919 Chr.11: 57740945-57740945 on Build GRCh38
rs369566827 Chr.11: 57740745-57740745 on Build GRCh38
rs969758576 Chr.11: 57740629-57740629 on Build GRCh38
rs761227841 Chr.11: 57741028-57741028 on Build GRCh38
rs931117057 Chr.11: 57740715-57740715 on Build GRCh38
rs1054401934 Chr.11: 57740980-57740980 on Build GRCh38
rs933104361 Chr.11: 57740703-57740703 on Build GRCh38
rs927146778 Chr.11: 57740921-57740921 on Build GRCh38
rs913707975 Chr.11: 57740668-57740668 on Build GRCh38
rs553748058 Chr.11: 57740843-57740843 on Build GRCh38
rs192084385 Chr.11: 57740809-57740809 on Build GRCh38
rs11547016 Chr.11: 57740645-57740645 on Build GRCh38
rs1001146171 Chr.11: 57740619-57740619 on Build GRCh38
rs12577827 Chr.11: 57740657-57740657 on Build GRCh38
rs556932045 Chr.11: 57740977-57740977 on Build GRCh38
rs925188174 Chr.11: 57740658-57740658 on Build GRCh38
rs988034094 Chr.11: 57740847-57740847 on Build GRCh38
rs868106380 Chr.11: 57740828-57740828 on Build GRCh38
rs184113182 Chr.11: 57740824-57740824 on Build GRCh38
rs910292698 Chr.11: 57740917-57740917 on Build GRCh38
rs895477324 Chr.11: 57740788-57740788 on Build GRCh38
rs945669636 Chr.11: 57740954-57740954 on Build GRCh38
rs143808914 Chr.11: 57740978-57740978 on Build GRCh38
rs984068707 Chr.11: 57740736-57740736 on Build GRCh38
rs994095389 Chr.11: 57741079-57741079 on Build GRCh38
rs750938228 Chr.11: 57740992-57740992 on Build GRCh38
rs943021702 Chr.11: 57740716-57740716 on Build GRCh38
rs945810399 Chr.11: 57740804-57740804 on Build GRCh38

View Full Product Details