Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 58610820-58611322 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFP91
Entrez Gene ID
80829
Gene Name
ZFP91 zinc finger protein
Gene Aliases
DMS-8, DSM-8, FKSG11, PZF, ZFP-91, ZNF757
UniGene
Hs.524920
Gene Chromosome Location
Chr.11: 58579114-58621550 on Build GRCh38
Transcripts
NM_001197051.1 NM_053023.4 NR_024091.1

Gene Symbol
ZFP91-CNTF
Entrez Gene ID
386607
Gene Name
ZFP91-CNTF readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.524920
Gene Chromosome Location
Chr.11: 58579114-58625733 on Build GRCh38
Transcripts
NM_001197051.1 NM_053023.4 NR_024091.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6069417 c.646G>A p.E216K Chr.11: 58610978-58610978 on Build GRCh38
1270893 c.678C>T p.F226F Chr.11: 58611010-58611010 on Build GRCh38
3752521 c.619A>G p.S207G Chr.11: 58610951-58610951 on Build GRCh38
3752520 c.619A>G p.S207G Chr.11: 58610951-58610951 on Build GRCh38
6133536 c.664G>C p.E222Q Chr.11: 58610996-58610996 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs550921562 Chr.11: 58611114-58611114 on Build GRCh38
rs764643103 Chr.11: 58611080-58611080 on Build GRCh38
rs113748078 Chr.11: 58610979-58610979 on Build GRCh38
rs757638760 Chr.11: 58611100-58611100 on Build GRCh38
rs28362517 Chr.11: 58610883-58610883 on Build GRCh38
rs638467 Chr.11: 58610861-58610861 on Build GRCh38
rs575765077 Chr.11: 58610924-58610924 on Build GRCh38
rs12282490 Chr.11: 58610847-58610847 on Build GRCh38
rs762623323 Chr.11: 58611143-58611143 on Build GRCh38
rs757613959 Chr.11: 58610935-58610935 on Build GRCh38
rs780295977 Chr.11: 58611076-58611076 on Build GRCh38
rs768611318 Chr.11: 58610988-58610988 on Build GRCh38
rs546652104 Chr.11: 58611263-58611263 on Build GRCh38
rs76596241 Chr.11: 58611204-58611204 on Build GRCh38
rs1030812558 Chr.11: 58611283-58611283 on Build GRCh38
rs895777724 Chr.11: 58611248-58611248 on Build GRCh38
rs772910103 Chr.11: 58611045-58611045 on Build GRCh38
rs758232182 Chr.11: 58611119-58611123 on Build GRCh38
rs766050804 Chr.11: 58610912-58610912 on Build GRCh38
rs12282535 Chr.11: 58610921-58610921 on Build GRCh38
rs746045614 Chr.11: 58611110-58611110 on Build GRCh38
rs757892072 Chr.11: 58610911-58610911 on Build GRCh38
rs111240529 Chr.11: 58610982-58610982 on Build GRCh38
rs767969643 Chr.11: 58610947-58610949 on Build GRCh38
rs758624125 Chr.11: 58611111-58611111 on Build GRCh38
rs756850146 Chr.11: 58611171-58611174 on Build GRCh38
rs112181818 Chr.11: 58610890-58610890 on Build GRCh38
rs746145793 Chr.11: 58610943-58610943 on Build GRCh38
rs943809528 Chr.11: 58611101-58611101 on Build GRCh38
rs756320458 Chr.11: 58611152-58611152 on Build GRCh38
rs754358290 Chr.11: 58611222-58611222 on Build GRCh38
rs746882540 Chr.11: 58610964-58610964 on Build GRCh38
rs8373 Chr.11: 58610951-58610951 on Build GRCh38
rs557105778 Chr.11: 58611254-58611254 on Build GRCh38
rs546361411 Chr.11: 58610908-58610908 on Build GRCh38
rs533022106 Chr.11: 58611189-58611189 on Build GRCh38
rs79135825 Chr.11: 58610949-58610949 on Build GRCh38
rs1055256717 Chr.11: 58611180-58611180 on Build GRCh38
rs779205356 Chr.11: 58611104-58611104 on Build GRCh38
rs776239529 Chr.11: 58610998-58610998 on Build GRCh38
rs2245938 Chr.11: 58610909-58610909 on Build GRCh38
rs776489568 Chr.11: 58610902-58610902 on Build GRCh38
rs1013518563 Chr.11: 58610857-58610857 on Build GRCh38
rs910859536 Chr.11: 58611034-58611034 on Build GRCh38
rs979249548 Chr.11: 58610894-58610894 on Build GRCh38
rs777543700 Chr.11: 58611124-58611124 on Build GRCh38
rs539900935 Chr.11: 58610934-58610934 on Build GRCh38
rs1048684733 Chr.11: 58611184-58611184 on Build GRCh38
rs562756552 Chr.11: 58611134-58611134 on Build GRCh38
rs1055820076 Chr.11: 58611236-58611236 on Build GRCh38
rs573747696 Chr.11: 58611298-58611298 on Build GRCh38
rs751150130 Chr.11: 58611074-58611074 on Build GRCh38
rs766623386 Chr.11: 58610963-58610965 on Build GRCh38
rs765925242 Chr.11: 58611070-58611070 on Build GRCh38
rs372594868 Chr.11: 58610916-58610916 on Build GRCh38
rs535130896 Chr.11: 58611260-58611260 on Build GRCh38
rs746725435 Chr.11: 58611160-58611160 on Build GRCh38
rs966561849 Chr.11: 58610886-58610886 on Build GRCh38
rs752659728 Chr.11: 58611100-58611101 on Build GRCh38
rs753793734 Chr.11: 58611097-58611098 on Build GRCh38
rs767005374 Chr.11: 58611205-58611205 on Build GRCh38
rs780067763 Chr.11: 58611113-58611113 on Build GRCh38
rs1014358800 Chr.11: 58611253-58611253 on Build GRCh38
rs1020915320 Chr.11: 58611228-58611228 on Build GRCh38
rs765572050 Chr.11: 58611265-58611265 on Build GRCh38
rs754975822 Chr.11: 58611099-58611100 on Build GRCh38
rs764978662 Chr.11: 58611004-58611004 on Build GRCh38
rs1020861248 Chr.11: 58610860-58610860 on Build GRCh38
rs866608478 Chr.11: 58610993-58610993 on Build GRCh38
rs762032340 Chr.11: 58610940-58610945 on Build GRCh38
rs781151120 Chr.11: 58611118-58611118 on Build GRCh38
rs748099207 Chr.11: 58611128-58611128 on Build GRCh38
rs756698042 Chr.11: 58610919-58610919 on Build GRCh38
rs773606671 Chr.11: 58610954-58610959 on Build GRCh38
rs769777354 Chr.11: 58610974-58610974 on Build GRCh38
rs757420355 Chr.11: 58610917-58610917 on Build GRCh38
rs779167680 Chr.11: 58610938-58610938 on Build GRCh38
rs769615486 Chr.11: 58611130-58611130 on Build GRCh38
rs754344527 Chr.11: 58611099-58611099 on Build GRCh38
rs908122460 Chr.11: 58610957-58610957 on Build GRCh38
rs1013653318 Chr.11: 58611209-58611210 on Build GRCh38
rs557201388 Chr.11: 58610850-58610850 on Build GRCh38
rs1033582901 Chr.11: 58611267-58611267 on Build GRCh38
rs929865220 Chr.11: 58611177-58611177 on Build GRCh38
rs762749591 Chr.11: 58611056-58611056 on Build GRCh38
rs760973725 Chr.11: 58610960-58610962 on Build GRCh38

View Full Product Details