Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 60179752-60180252 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MS4A6A
Entrez Gene ID
64231
Gene Name
membrane spanning 4-domains A6A
Gene Aliases
4SPAN3, 4SPAN3.2, CD20L3, CDA01, MS4A6, MST090, MSTP090
UniGene
Hs.523702
Gene Chromosome Location
Chr.11: 60171607-60184666 on Build GRCh38
Transcripts
NM_001247999.1 NM_022349.3 NM_152851.2 NM_152852.2 XM_005274177.2 XM_006718658.2 XM_006718660.2 XM_006718661.2 XM_011545209.1 XM_017018123.1 XM_017018124.1 XM_017018125.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5604118 c.218C>T p.P73L Chr.11: 60179895-60179895 on Build GRCh38
4911716 c.207T>C p.A69A Chr.11: 60179906-60179906 on Build GRCh38
5341626 c.268T>C p.L90L Chr.11: 60179929-60179929 on Build GRCh38
344029 c.215C>G p.S72C Chr.11: 60179898-60179898 on Build GRCh38
3450395 c.276C>T p.I92I Chr.11: 60179921-60179921 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs948002699 Chr.11: 60179991-60179991 on Build GRCh38
rs755881843 Chr.11: 60179830-60179831 on Build GRCh38
rs747966380 Chr.11: 60179955-60179955 on Build GRCh38
rs767248647 Chr.11: 60179821-60179822 on Build GRCh38
rs583791 Chr.11: 60179779-60179779 on Build GRCh38
rs375008083 Chr.11: 60180071-60180071 on Build GRCh38
rs201705826 Chr.11: 60179870-60179870 on Build GRCh38
rs994927036 Chr.11: 60180066-60180066 on Build GRCh38
rs201491942 Chr.11: 60179868-60179868 on Build GRCh38
rs557660195 Chr.11: 60180040-60180040 on Build GRCh38
rs746012399 Chr.11: 60179813-60179813 on Build GRCh38
rs532459647 Chr.11: 60179807-60179807 on Build GRCh38
rs536424213 Chr.11: 60180037-60180037 on Build GRCh38
rs779090668 Chr.11: 60179914-60179914 on Build GRCh38
rs757973221 Chr.11: 60179781-60179781 on Build GRCh38
rs780245885 Chr.11: 60180015-60180015 on Build GRCh38
rs774194882 Chr.11: 60179978-60179978 on Build GRCh38
rs751125562 Chr.11: 60179793-60179793 on Build GRCh38
rs758814336 Chr.11: 60180013-60180013 on Build GRCh38
rs758888269 Chr.11: 60179968-60179968 on Build GRCh38
rs142502787 Chr.11: 60180030-60180030 on Build GRCh38
rs1002795299 Chr.11: 60180138-60180138 on Build GRCh38
rs777912985 Chr.11: 60180047-60180047 on Build GRCh38
rs142312942 Chr.11: 60179925-60179925 on Build GRCh38
rs559668079 Chr.11: 60180224-60180224 on Build GRCh38
rs549670217 Chr.11: 60179831-60179831 on Build GRCh38
rs750757798 Chr.11: 60180012-60180012 on Build GRCh38
rs577283728 Chr.11: 60180168-60180168 on Build GRCh38
rs751072418 Chr.11: 60179890-60179890 on Build GRCh38
rs753488437 Chr.11: 60179952-60179952 on Build GRCh38
rs779636346 Chr.11: 60179789-60179789 on Build GRCh38
rs760790218 Chr.11: 60179808-60179808 on Build GRCh38
rs987787037 Chr.11: 60179943-60179943 on Build GRCh38
rs766097790 Chr.11: 60179972-60179972 on Build GRCh38
rs867210808 Chr.11: 60179841-60179841 on Build GRCh38
rs988383326 Chr.11: 60180171-60180171 on Build GRCh38
rs1022249832 Chr.11: 60180056-60180056 on Build GRCh38
rs768785446 Chr.11: 60179933-60179933 on Build GRCh38
rs375980883 Chr.11: 60179836-60179836 on Build GRCh38
rs747293288 Chr.11: 60180016-60180016 on Build GRCh38
rs750234193 Chr.11: 60179873-60179873 on Build GRCh38
rs776485053 Chr.11: 60179851-60179851 on Build GRCh38
rs147611482 Chr.11: 60179882-60179882 on Build GRCh38
rs532381110 Chr.11: 60179912-60179912 on Build GRCh38
rs954313523 Chr.11: 60180070-60180070 on Build GRCh38
rs758278290 Chr.11: 60180044-60180044 on Build GRCh38
rs558996807 Chr.11: 60180157-60180157 on Build GRCh38
rs368555139 Chr.11: 60179922-60179922 on Build GRCh38
rs868512640 Chr.11: 60180055-60180055 on Build GRCh38
rs200749132 Chr.11: 60179970-60179970 on Build GRCh38
rs969548379 Chr.11: 60180088-60180088 on Build GRCh38
rs772244377 Chr.11: 60179919-60179919 on Build GRCh38
rs757658575 Chr.11: 60179992-60179992 on Build GRCh38
rs749633289 Chr.11: 60179810-60179810 on Build GRCh38
rs965421643 Chr.11: 60179846-60179846 on Build GRCh38
rs766149183 Chr.11: 60179884-60179884 on Build GRCh38
rs751391419 Chr.11: 60180179-60180179 on Build GRCh38
rs972763456 Chr.11: 60180198-60180198 on Build GRCh38
rs756638678 Chr.11: 60179794-60179794 on Build GRCh38
rs775502191 Chr.11: 60179825-60179825 on Build GRCh38
rs754370880 Chr.11: 60179903-60179903 on Build GRCh38
rs541350839 Chr.11: 60180206-60180206 on Build GRCh38
rs746255438 Chr.11: 60179917-60179917 on Build GRCh38
rs779192277 Chr.11: 60179812-60179812 on Build GRCh38
rs766872413 Chr.11: 60179982-60179982 on Build GRCh38
rs201014833 Chr.11: 60179832-60179832 on Build GRCh38
rs769781965 Chr.11: 60179959-60179959 on Build GRCh38
rs766394874 Chr.11: 60179857-60179857 on Build GRCh38
rs772297224 Chr.11: 60179821-60179821 on Build GRCh38
rs546886853 Chr.11: 60180057-60180057 on Build GRCh38
rs151122444 Chr.11: 60180017-60180017 on Build GRCh38
rs2305431 Chr.11: 60179902-60179902 on Build GRCh38
rs913564364 Chr.11: 60180182-60180182 on Build GRCh38
rs529814098 Chr.11: 60179932-60179932 on Build GRCh38
rs189938105 Chr.11: 60180029-60180029 on Build GRCh38
rs776622879 Chr.11: 60179944-60179944 on Build GRCh38
rs185080144 Chr.11: 60179907-60179907 on Build GRCh38
rs202162706 Chr.11: 60179993-60179993 on Build GRCh38
rs754574003 Chr.11: 60179898-60179898 on Build GRCh38
rs530571432 Chr.11: 60179987-60179987 on Build GRCh38
rs200072504 Chr.11: 60179981-60179981 on Build GRCh38
rs377193282 Chr.11: 60180031-60180031 on Build GRCh38

View Full Product Details