Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 60300859-60301360 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MS4A4A
Entrez Gene ID
51338
Gene Name
membrane spanning 4-domains A4A
Gene Aliases
4SPAN1, CD20-L1, CD20L1, HDCME31P, MS4A4, MS4A7
UniGene
Hs.325960
Gene Chromosome Location
Chr.11: 60280541-60308972 on Build GRCh38
Transcripts
NM_001243266.1 NM_024021.3 NM_148975.2 XM_017017909.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6365904 c.286G>A p.G96R Chr.11: 60301013-60301013 on Build GRCh38
4420401 c.387G>C p.L129L Chr.11: 60301057-60301057 on Build GRCh38
689458 c.383G>C p.G128A Chr.11: 60301053-60301053 on Build GRCh38
929174 c.310A>C p.I104L Chr.11: 60301037-60301037 on Build GRCh38
5441020 c.277A>T p.I93F Chr.11: 60301004-60301004 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749413854 Chr.11: 60301015-60301015 on Build GRCh38
rs370792005 Chr.11: 60300946-60300946 on Build GRCh38
rs766985783 Chr.11: 60301082-60301082 on Build GRCh38
rs973695853 Chr.11: 60301058-60301058 on Build GRCh38
rs914564388 Chr.11: 60301331-60301331 on Build GRCh38
rs996922093 Chr.11: 60300918-60300918 on Build GRCh38
rs879476315 Chr.11: 60301025-60301025 on Build GRCh38
rs774450435 Chr.11: 60300987-60300988 on Build GRCh38
rs781621575 Chr.11: 60300969-60300969 on Build GRCh38
rs547479495 Chr.11: 60301052-60301052 on Build GRCh38
rs756263672 Chr.11: 60301101-60301101 on Build GRCh38
rs990555771 Chr.11: 60301262-60301263 on Build GRCh38
rs979656925 Chr.11: 60301201-60301201 on Build GRCh38
rs751632293 Chr.11: 60301051-60301052 on Build GRCh38
rs761680706 Chr.11: 60301001-60301001 on Build GRCh38
rs752043205 Chr.11: 60301084-60301084 on Build GRCh38
rs781365824 Chr.11: 60301097-60301097 on Build GRCh38
rs924809593 Chr.11: 60301287-60301287 on Build GRCh38
rs995368214 Chr.11: 60301100-60301100 on Build GRCh38
rs1015430121 Chr.11: 60301211-60301211 on Build GRCh38
rs973004266 Chr.11: 60301306-60301306 on Build GRCh38
rs143864976 Chr.11: 60301253-60301253 on Build GRCh38
rs766152690 Chr.11: 60301085-60301086 on Build GRCh38
rs960908651 Chr.11: 60301147-60301147 on Build GRCh38
rs147625120 Chr.11: 60301054-60301054 on Build GRCh38
rs772993181 Chr.11: 60301072-60301073 on Build GRCh38
rs920943248 Chr.11: 60301203-60301203 on Build GRCh38
rs556084438 Chr.11: 60301125-60301125 on Build GRCh38
rs574833277 Chr.11: 60301295-60301295 on Build GRCh38
rs770080559 Chr.11: 60300982-60300982 on Build GRCh38
rs112999646 Chr.11: 60301168-60301168 on Build GRCh38
rs200174846 Chr.11: 60301018-60301018 on Build GRCh38
rs35437255 Chr.11: 60301275-60301276 on Build GRCh38
rs748263014 Chr.11: 60300973-60300973 on Build GRCh38
rs912404313 Chr.11: 60301044-60301044 on Build GRCh38
rs897031408 Chr.11: 60301213-60301213 on Build GRCh38
rs755520504 Chr.11: 60301087-60301087 on Build GRCh38
rs748580665 Chr.11: 60301098-60301098 on Build GRCh38
rs1045910681 Chr.11: 60301112-60301112 on Build GRCh38
rs979126778 Chr.11: 60300908-60300908 on Build GRCh38
rs112460988 Chr.11: 60301267-60301267 on Build GRCh38
rs756881817 Chr.11: 60301075-60301075 on Build GRCh38
rs372861848 Chr.11: 60301330-60301330 on Build GRCh38
rs968287536 Chr.11: 60300901-60300901 on Build GRCh38
rs759323164 Chr.11: 60301047-60301047 on Build GRCh38
rs939775773 Chr.11: 60301283-60301283 on Build GRCh38
rs565962862 Chr.11: 60301268-60301268 on Build GRCh38
rs753504775 Chr.11: 60301104-60301105 on Build GRCh38
rs932722341 Chr.11: 60301167-60301167 on Build GRCh38
rs970519695 Chr.11: 60301274-60301274 on Build GRCh38
rs7949560 Chr.11: 60301141-60301141 on Build GRCh38
rs865899955 Chr.11: 60300895-60300895 on Build GRCh38
rs116580535 Chr.11: 60301184-60301184 on Build GRCh38
rs968312232 Chr.11: 60301197-60301197 on Build GRCh38
rs779242000 Chr.11: 60300902-60300902 on Build GRCh38
rs763846897 Chr.11: 60301063-60301063 on Build GRCh38
rs536402796 Chr.11: 60301116-60301116 on Build GRCh38
rs760533307 Chr.11: 60301086-60301086 on Build GRCh38
rs920912822 Chr.11: 60300945-60300945 on Build GRCh38
rs778106015 Chr.11: 60301103-60301103 on Build GRCh38
rs934930544 Chr.11: 60301136-60301136 on Build GRCh38
rs368745795 Chr.11: 60300910-60300910 on Build GRCh38
rs749561361 Chr.11: 60301146-60301149 on Build GRCh38
rs995457426 Chr.11: 60301248-60301248 on Build GRCh38
rs753375659 Chr.11: 60301068-60301068 on Build GRCh38
rs369641774 Chr.11: 60301109-60301109 on Build GRCh38
rs986567780 Chr.11: 60301255-60301255 on Build GRCh38
rs140807080 Chr.11: 60301059-60301059 on Build GRCh38
rs767540265 Chr.11: 60301007-60301008 on Build GRCh38
rs934913795 Chr.11: 60301292-60301292 on Build GRCh38
rs905181644 Chr.11: 60301119-60301119 on Build GRCh38
rs774331114 Chr.11: 60301022-60301022 on Build GRCh38
rs572081659 Chr.11: 60301208-60301208 on Build GRCh38
rs200557697 Chr.11: 60301011-60301011 on Build GRCh38

View Full Product Details