Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 60871125-60871620 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZP1
Entrez Gene ID
22917
Gene Name
zona pellucida glycoprotein 1
Gene Aliases
HEL163, OOMD, OOMD1
UniGene
Hs.172130
Gene Chromosome Location
Chr.11: 60867542-60875693 on Build GRCh38
Transcripts
NM_207341.3 XM_011544852.2 XM_011544853.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5838928 c.1027C>T p.Q343* Chr.11: 60871229-60871229 on Build GRCh38
2036569 c.1033A>G p.I345V Chr.11: 60871235-60871235 on Build GRCh38
689378 c.1022G>T p.G341V Chr.11: 60871224-60871224 on Build GRCh38
5477720 c.1103G>A p.S368N Chr.11: 60871305-60871305 on Build GRCh38
5460476 c.1032C>A p.L344L Chr.11: 60871234-60871234 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200345260 Chr.11: 60871297-60871297 on Build GRCh38
rs529750157 Chr.11: 60871374-60871374 on Build GRCh38
rs117566381 Chr.11: 60871280-60871280 on Build GRCh38
rs114192813 Chr.11: 60871391-60871391 on Build GRCh38
rs987812398 Chr.11: 60871200-60871200 on Build GRCh38
rs377106329 Chr.11: 60871248-60871248 on Build GRCh38
rs772595609 Chr.11: 60871250-60871250 on Build GRCh38
rs964589635 Chr.11: 60871189-60871189 on Build GRCh38
rs775667106 Chr.11: 60871254-60871254 on Build GRCh38
rs948942236 Chr.11: 60871373-60871373 on Build GRCh38
rs377224060 Chr.11: 60871153-60871153 on Build GRCh38
rs764256632 Chr.11: 60871154-60871154 on Build GRCh38
rs1011666223 Chr.11: 60871451-60871451 on Build GRCh38
rs560242743 Chr.11: 60871365-60871365 on Build GRCh38
rs964588365 Chr.11: 60871591-60871591 on Build GRCh38
rs370046295 Chr.11: 60871156-60871156 on Build GRCh38
rs539465410 Chr.11: 60871483-60871483 on Build GRCh38
rs766796564 Chr.11: 60871152-60871157 on Build GRCh38
rs1003359726 Chr.11: 60871455-60871455 on Build GRCh38
rs199843782 Chr.11: 60871305-60871305 on Build GRCh38
rs758197113 Chr.11: 60871194-60871194 on Build GRCh38
rs755688586 Chr.11: 60871336-60871336 on Build GRCh38
rs1000378159 Chr.11: 60871579-60871579 on Build GRCh38
rs765433489 Chr.11: 60871362-60871363 on Build GRCh38
rs112226679 Chr.11: 60871213-60871213 on Build GRCh38
rs780369403 Chr.11: 60871155-60871155 on Build GRCh38
rs372297696 Chr.11: 60871226-60871226 on Build GRCh38
rs751251211 Chr.11: 60871205-60871205 on Build GRCh38
rs908573291 Chr.11: 60871319-60871319 on Build GRCh38
rs997415671 Chr.11: 60871477-60871477 on Build GRCh38
rs114636442 Chr.11: 60871584-60871584 on Build GRCh38
rs777376296 Chr.11: 60871338-60871338 on Build GRCh38
rs754640634 Chr.11: 60871218-60871218 on Build GRCh38
rs773167967 Chr.11: 60871289-60871289 on Build GRCh38
rs775771568 Chr.11: 60871576-60871592 on Build GRCh38
rs573773151 Chr.11: 60871434-60871434 on Build GRCh38
rs767240451 Chr.11: 60871167-60871167 on Build GRCh38
rs1044932739 Chr.11: 60871411-60871411 on Build GRCh38
rs765236048 Chr.11: 60871287-60871287 on Build GRCh38
rs747398422 Chr.11: 60871353-60871353 on Build GRCh38
rs747727685 Chr.11: 60871225-60871225 on Build GRCh38
rs1047090804 Chr.11: 60871376-60871376 on Build GRCh38
rs552066309 Chr.11: 60871437-60871437 on Build GRCh38
rs747377357 Chr.11: 60871256-60871256 on Build GRCh38
rs540349918 Chr.11: 60871345-60871345 on Build GRCh38
rs759371225 Chr.11: 60871299-60871299 on Build GRCh38
rs1021879678 Chr.11: 60871571-60871571 on Build GRCh38
rs964641945 Chr.11: 60871476-60871476 on Build GRCh38
rs754193132 Chr.11: 60871233-60871236 on Build GRCh38
rs146542167 Chr.11: 60871296-60871296 on Build GRCh38
rs909801730 Chr.11: 60871387-60871387 on Build GRCh38
rs779579016 Chr.11: 60871240-60871240 on Build GRCh38
rs780866828 Chr.11: 60871224-60871224 on Build GRCh38
rs112964743 Chr.11: 60871431-60871431 on Build GRCh38
rs768796578 Chr.11: 60871257-60871257 on Build GRCh38
rs376630078 Chr.11: 60871358-60871358 on Build GRCh38
rs535323884 Chr.11: 60871152-60871152 on Build GRCh38
rs866126006 Chr.11: 60871416-60871416 on Build GRCh38
rs777982724 Chr.11: 60871457-60871457 on Build GRCh38
rs373846692 Chr.11: 60871465-60871465 on Build GRCh38
rs751351771 Chr.11: 60871298-60871298 on Build GRCh38
rs745798186 Chr.11: 60871177-60871177 on Build GRCh38
rs765223404 Chr.11: 60871176-60871176 on Build GRCh38
rs998650925 Chr.11: 60871429-60871429 on Build GRCh38
rs916169439 Chr.11: 60871369-60871369 on Build GRCh38
rs767134275 Chr.11: 60871303-60871303 on Build GRCh38
rs549353768 Chr.11: 60871392-60871392 on Build GRCh38
rs531917813 Chr.11: 60871430-60871430 on Build GRCh38
rs776904131 Chr.11: 60871263-60871263 on Build GRCh38
rs761847157 Chr.11: 60871169-60871169 on Build GRCh38
rs750345575 Chr.11: 60871177-60871177 on Build GRCh38
rs555527981 Chr.11: 60871196-60871196 on Build GRCh38
rs563096087 Chr.11: 60871414-60871414 on Build GRCh38
rs11230511 Chr.11: 60871340-60871340 on Build GRCh38
rs1009228448 Chr.11: 60871528-60871528 on Build GRCh38
rs990123537 Chr.11: 60871159-60871159 on Build GRCh38
rs755345891 Chr.11: 60871355-60871363 on Build GRCh38

View Full Product Details