Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 60922883-60923395 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM109
Entrez Gene ID
79073
Gene Name
transmembrane protein 109
Gene Aliases
-
UniGene
Hs.13662
Gene Chromosome Location
Chr.11: 60913899-60923443 on Build GRCh38
Transcripts
NM_024092.2

SNP Detail

SNP ID Chromosome Location
rs1032751159 Chr.11: 60923070-60923070 on Build GRCh38
rs7116175 Chr.11: 60923108-60923108 on Build GRCh38
rs893778230 Chr.11: 60923251-60923251 on Build GRCh38
rs978835596 Chr.11: 60923021-60923021 on Build GRCh38
rs977153343 Chr.11: 60923057-60923057 on Build GRCh38
rs549294313 Chr.11: 60923158-60923158 on Build GRCh38
rs949516207 Chr.11: 60923187-60923187 on Build GRCh38
rs909519293 Chr.11: 60923159-60923159 on Build GRCh38
rs965171599 Chr.11: 60923080-60923080 on Build GRCh38
rs984421181 Chr.11: 60923358-60923358 on Build GRCh38
rs909765629 Chr.11: 60923344-60923344 on Build GRCh38
rs540732391 Chr.11: 60922962-60922962 on Build GRCh38
rs1046547991 Chr.11: 60923201-60923201 on Build GRCh38
rs939070984 Chr.11: 60923119-60923119 on Build GRCh38
rs73493043 Chr.11: 60923263-60923263 on Build GRCh38
rs554083577 Chr.11: 60923052-60923052 on Build GRCh38
rs578038931 Chr.11: 60922913-60922913 on Build GRCh38
rs998302943 Chr.11: 60923313-60923313 on Build GRCh38
rs189007301 Chr.11: 60923343-60923343 on Build GRCh38
rs1054068695 Chr.11: 60923219-60923219 on Build GRCh38
rs1055492400 Chr.11: 60922954-60922954 on Build GRCh38
rs1013720096 Chr.11: 60922990-60922990 on Build GRCh38
rs1018329931 Chr.11: 60923300-60923300 on Build GRCh38
rs546533756 Chr.11: 60923196-60923197 on Build GRCh38
rs867855603 Chr.11: 60923266-60923266 on Build GRCh38
rs902716849 Chr.11: 60923202-60923202 on Build GRCh38
rs1046598098 Chr.11: 60923030-60923030 on Build GRCh38
rs1018391719 Chr.11: 60923075-60923075 on Build GRCh38
rs968830814 Chr.11: 60922984-60922984 on Build GRCh38
rs935570513 Chr.11: 60923212-60923212 on Build GRCh38
rs937040335 Chr.11: 60923161-60923161 on Build GRCh38
rs1020399323 Chr.11: 60922963-60922963 on Build GRCh38
rs34697422 Chr.11: 60923126-60923127 on Build GRCh38
rs577436170 Chr.11: 60923177-60923177 on Build GRCh38
rs922566917 Chr.11: 60923059-60923060 on Build GRCh38
rs1056516328 Chr.11: 60923141-60923141 on Build GRCh38
rs976582713 Chr.11: 60923089-60923089 on Build GRCh38
rs1007369606 Chr.11: 60923074-60923074 on Build GRCh38
rs1010460007 Chr.11: 60923176-60923176 on Build GRCh38
rs1025468238 Chr.11: 60923335-60923335 on Build GRCh38
rs901253362 Chr.11: 60923309-60923309 on Build GRCh38
rs999821153 Chr.11: 60923056-60923056 on Build GRCh38
rs140318423 Chr.11: 60923127-60923127 on Build GRCh38
rs61899181 Chr.11: 60923231-60923231 on Build GRCh38
rs951366177 Chr.11: 60923126-60923126 on Build GRCh38
rs958095464 Chr.11: 60923071-60923071 on Build GRCh38
rs560563333 Chr.11: 60923118-60923118 on Build GRCh38
rs937072514 Chr.11: 60922940-60922940 on Build GRCh38
rs1055441048 Chr.11: 60923162-60923162 on Build GRCh38
rs895294910 Chr.11: 60922979-60922979 on Build GRCh38
rs531933456 Chr.11: 60923269-60923269 on Build GRCh38
rs929867917 Chr.11: 60923066-60923066 on Build GRCh38

View Full Product Details