Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61122807-61123305 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD5
Entrez Gene ID
921
Gene Name
CD5 molecule
Gene Aliases
LEU1, T1
UniGene
Hs.58685
Gene Chromosome Location
Chr.11: 61102458-61127852 on Build GRCh38
Transcripts
NM_014207.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7214987 c.1185C>T p.V395V Chr.11: 61122992-61122992 on Build GRCh38
6519116 c.1116C>A p.P372P Chr.11: 61122923-61122923 on Build GRCh38
2037076 c.1137G>A p.T379T Chr.11: 61122944-61122944 on Build GRCh38
1704108 c.1172T>A p.V391E Chr.11: 61122979-61122979 on Build GRCh38
1355180 c.1155G>A p.L385L Chr.11: 61122962-61122962 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768180897 Chr.11: 61122892-61122892 on Build GRCh38
rs745961025 Chr.11: 61122909-61122909 on Build GRCh38
rs138495063 Chr.11: 61122998-61122998 on Build GRCh38
rs140393887 Chr.11: 61122864-61122864 on Build GRCh38
rs373847040 Chr.11: 61123044-61123044 on Build GRCh38
rs778308890 Chr.11: 61123102-61123102 on Build GRCh38
rs370905519 Chr.11: 61123043-61123043 on Build GRCh38
rs765411444 Chr.11: 61122964-61122965 on Build GRCh38
rs764258889 Chr.11: 61122952-61122952 on Build GRCh38
rs939767574 Chr.11: 61123179-61123179 on Build GRCh38
rs771893006 Chr.11: 61123067-61123067 on Build GRCh38
rs117646548 Chr.11: 61122936-61122936 on Build GRCh38
rs746706273 Chr.11: 61123001-61123001 on Build GRCh38
rs1045107399 Chr.11: 61122931-61122931 on Build GRCh38
rs953269176 Chr.11: 61123186-61123186 on Build GRCh38
rs759076498 Chr.11: 61123182-61123182 on Build GRCh38
rs775961418 Chr.11: 61122901-61122901 on Build GRCh38
rs552756645 Chr.11: 61123110-61123110 on Build GRCh38
rs559340518 Chr.11: 61122944-61122944 on Build GRCh38
rs1034877449 Chr.11: 61122843-61122843 on Build GRCh38
rs192428474 Chr.11: 61122864-61122864 on Build GRCh38
rs771363007 Chr.11: 61122916-61122916 on Build GRCh38
rs1003172979 Chr.11: 61123147-61123147 on Build GRCh38
rs1002669004 Chr.11: 61122842-61122842 on Build GRCh38
rs370584277 Chr.11: 61123060-61123060 on Build GRCh38
rs979295117 Chr.11: 61123109-61123109 on Build GRCh38
rs575783885 Chr.11: 61122855-61122855 on Build GRCh38
rs117045863 Chr.11: 61123028-61123028 on Build GRCh38
rs183289659 Chr.11: 61122869-61122869 on Build GRCh38
rs765259650 Chr.11: 61122990-61122990 on Build GRCh38
rs199607524 Chr.11: 61123049-61123049 on Build GRCh38
rs753805856 Chr.11: 61122960-61122960 on Build GRCh38
rs948972530 Chr.11: 61123128-61123128 on Build GRCh38
rs11230602 Chr.11: 61123136-61123136 on Build GRCh38
rs748167810 Chr.11: 61123047-61123047 on Build GRCh38
rs753848066 Chr.11: 61122867-61122867 on Build GRCh38
rs570621363 Chr.11: 61123089-61123089 on Build GRCh38
rs374622426 Chr.11: 61123112-61123112 on Build GRCh38
rs764848026 Chr.11: 61123271-61123271 on Build GRCh38
rs760772701 Chr.11: 61122945-61122945 on Build GRCh38
rs757097043 Chr.11: 61122868-61122868 on Build GRCh38
rs1018841810 Chr.11: 61123250-61123250 on Build GRCh38
rs930899028 Chr.11: 61123215-61123215 on Build GRCh38
rs573198698 Chr.11: 61123175-61123175 on Build GRCh38
rs35740410 Chr.11: 61122992-61122992 on Build GRCh38
rs1036478860 Chr.11: 61122834-61122834 on Build GRCh38
rs774765989 Chr.11: 61122924-61122924 on Build GRCh38
rs920933079 Chr.11: 61123111-61123111 on Build GRCh38
rs199785003 Chr.11: 61122993-61122993 on Build GRCh38
rs149639963 Chr.11: 61122999-61122999 on Build GRCh38
rs529365857 Chr.11: 61123236-61123238 on Build GRCh38
rs760968026 Chr.11: 61123068-61123068 on Build GRCh38
rs372280969 Chr.11: 61122912-61122912 on Build GRCh38
rs568377511 Chr.11: 61123234-61123234 on Build GRCh38
rs1052110633 Chr.11: 61123162-61123162 on Build GRCh38
rs4245224 Chr.11: 61123091-61123091 on Build GRCh38
rs1007820498 Chr.11: 61123243-61123243 on Build GRCh38
rs200326779 Chr.11: 61122877-61122877 on Build GRCh38
rs986474513 Chr.11: 61123137-61123137 on Build GRCh38
rs752819439 Chr.11: 61123082-61123083 on Build GRCh38
rs760404081 Chr.11: 61123103-61123103 on Build GRCh38
rs757219518 Chr.11: 61122975-61122975 on Build GRCh38
rs746275565 Chr.11: 61123059-61123059 on Build GRCh38
rs767776548 Chr.11: 61122939-61122939 on Build GRCh38
rs759713555 Chr.11: 61122889-61122890 on Build GRCh38
rs1020731514 Chr.11: 61123099-61123099 on Build GRCh38
rs1013205027 Chr.11: 61122891-61122891 on Build GRCh38
rs780760300 Chr.11: 61123024-61123024 on Build GRCh38
rs919393392 Chr.11: 61123184-61123184 on Build GRCh38
rs756199996 Chr.11: 61122863-61122863 on Build GRCh38
rs755797613 Chr.11: 61122869-61122869 on Build GRCh38
rs144236116 Chr.11: 61123020-61123020 on Build GRCh38

View Full Product Details