Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61902351-61902862 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB3IL1
Entrez Gene ID
5866
Gene Name
RAB3A interacting protein like 1
Gene Aliases
GRAB
UniGene
Hs.13759
Gene Chromosome Location
Chr.11: 61897296-61946312 on Build GRCh38
Transcripts
NM_001271686.1 NM_013401.3 XM_005274140.4 XM_006718647.3 XM_011545196.2 XM_011545197.2 XM_011545198.2 XM_011545199.2 XM_011545200.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4427227 c.900-1G>C p.? Chr.11: 61902542-61902542 on Build GRCh38
3739177 c.967C>T p.H323Y Chr.11: 61902474-61902474 on Build GRCh38
5965109 c.951C>T p.L317L Chr.11: 61902490-61902490 on Build GRCh38
1676089 c.925C>T p.R309C Chr.11: 61902516-61902516 on Build GRCh38
5458248 c.935G>A p.R312H Chr.11: 61902506-61902506 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs892977024 Chr.11: 61902454-61902454 on Build GRCh38
rs750341680 Chr.11: 61902496-61902496 on Build GRCh38
rs751174758 Chr.11: 61902687-61902687 on Build GRCh38
rs139628915 Chr.11: 61902455-61902455 on Build GRCh38
rs923984837 Chr.11: 61902409-61902409 on Build GRCh38
rs771492421 Chr.11: 61902560-61902560 on Build GRCh38
rs773889636 Chr.11: 61902476-61902476 on Build GRCh38
rs369063181 Chr.11: 61902516-61902516 on Build GRCh38
rs538759980 Chr.11: 61902775-61902775 on Build GRCh38
rs376882014 Chr.11: 61902460-61902460 on Build GRCh38
rs750154210 Chr.11: 61902487-61902487 on Build GRCh38
rs889734778 Chr.11: 61902465-61902465 on Build GRCh38
rs111293479 Chr.11: 61902816-61902816 on Build GRCh38
rs116610257 Chr.11: 61902664-61902664 on Build GRCh38
rs778167885 Chr.11: 61902673-61902673 on Build GRCh38
rs529887109 Chr.11: 61902669-61902669 on Build GRCh38
rs1034853673 Chr.11: 61902665-61902665 on Build GRCh38
rs909432127 Chr.11: 61902709-61902709 on Build GRCh38
rs527256903 Chr.11: 61902538-61902538 on Build GRCh38
rs771399474 Chr.11: 61902706-61902706 on Build GRCh38
rs918298136 Chr.11: 61902767-61902767 on Build GRCh38
rs781557779 Chr.11: 61902523-61902523 on Build GRCh38
rs756048663 Chr.11: 61902515-61902515 on Build GRCh38
rs752794464 Chr.11: 61902500-61902500 on Build GRCh38
rs747610934 Chr.11: 61902466-61902468 on Build GRCh38
rs772320713 Chr.11: 61902450-61902450 on Build GRCh38
rs549702716 Chr.11: 61902736-61902736 on Build GRCh38
rs935412787 Chr.11: 61902422-61902425 on Build GRCh38
rs911098101 Chr.11: 61902798-61902798 on Build GRCh38
rs766231559 Chr.11: 61902578-61902578 on Build GRCh38
rs374549951 Chr.11: 61902419-61902419 on Build GRCh38
rs748187334 Chr.11: 61902631-61902631 on Build GRCh38
rs1032852204 Chr.11: 61902581-61902581 on Build GRCh38
rs762549633 Chr.11: 61902639-61902639 on Build GRCh38
rs889537703 Chr.11: 61902814-61902814 on Build GRCh38
rs779084807 Chr.11: 61902422-61902422 on Build GRCh38
rs763818912 Chr.11: 61902683-61902683 on Build GRCh38
rs767072056 Chr.11: 61902823-61902823 on Build GRCh38
rs369427588 Chr.11: 61902416-61902416 on Build GRCh38
rs267603069 Chr.11: 61902461-61902461 on Build GRCh38
rs753666686 Chr.11: 61902587-61902587 on Build GRCh38
rs758474840 Chr.11: 61902507-61902507 on Build GRCh38
rs376769767 Chr.11: 61902431-61902431 on Build GRCh38
rs145011989 Chr.11: 61902494-61902494 on Build GRCh38
rs148671281 Chr.11: 61902495-61902495 on Build GRCh38
rs970973479 Chr.11: 61902603-61902603 on Build GRCh38
rs3815045 Chr.11: 61902474-61902474 on Build GRCh38
rs761369585 Chr.11: 61902553-61902553 on Build GRCh38
rs746277976 Chr.11: 61902525-61902525 on Build GRCh38
rs370497170 Chr.11: 61902492-61902492 on Build GRCh38
rs1020273365 Chr.11: 61902645-61902645 on Build GRCh38
rs139626798 Chr.11: 61902378-61902378 on Build GRCh38
rs773973219 Chr.11: 61902452-61902453 on Build GRCh38
rs760065197 Chr.11: 61902471-61902471 on Build GRCh38
rs761269708 Chr.11: 61902481-61902481 on Build GRCh38
rs971871809 Chr.11: 61902661-61902661 on Build GRCh38
rs893072976 Chr.11: 61902623-61902623 on Build GRCh38
rs949359437 Chr.11: 61902375-61902375 on Build GRCh38
rs989951287 Chr.11: 61902695-61902695 on Build GRCh38
rs537143844 Chr.11: 61902804-61902804 on Build GRCh38
rs1019241353 Chr.11: 61902482-61902482 on Build GRCh38
rs778042635 Chr.11: 61902513-61902513 on Build GRCh38
rs760432020 Chr.11: 61902566-61902566 on Build GRCh38
rs113099489 Chr.11: 61902542-61902542 on Build GRCh38
rs569794201 Chr.11: 61902765-61902765 on Build GRCh38
rs142156400 Chr.11: 61902540-61902540 on Build GRCh38
rs756826943 Chr.11: 61902689-61902689 on Build GRCh38
rs767190966 Chr.11: 61902485-61902485 on Build GRCh38
rs777175196 Chr.11: 61902727-61902727 on Build GRCh38
rs174471 Chr.11: 61902565-61902565 on Build GRCh38
rs770303582 Chr.11: 61902526-61902526 on Build GRCh38
rs760486887 Chr.11: 61902490-61902490 on Build GRCh38

View Full Product Details