Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62614866-62615360 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GAT3
Entrez Gene ID
26229
Gene Name
beta-1,3-glucuronyltransferase 3
Gene Aliases
GLCATI, JDSCD, glcUAT-I
UniGene
Hs.502759
Gene Chromosome Location
Chr.11: 62615296-62622175 on Build GRCh38
Transcripts
NM_001288721.1 NM_001288722.1 NM_001288723.1 NM_012200.3 NR_109991.1 NM_000327.3

Gene Symbol
ROM1
Entrez Gene ID
6094
Gene Name
retinal outer segment membrane protein 1
Gene Aliases
ROM, ROSP1, RP7, TSPAN23
UniGene
Hs.281564
Gene Chromosome Location
Chr.11: 62612741-62615120 on Build GRCh38
Transcripts
NM_001288721.1 NM_001288722.1 NM_001288723.1 NM_012200.3 NR_109991.1 NM_000327.3

SNP Detail

SNP ID Chromosome Location
rs774001343 Chr.11: 62615212-62615212 on Build GRCh38
rs886903981 Chr.11: 62614922-62614922 on Build GRCh38
rs751175681 Chr.11: 62615235-62615235 on Build GRCh38
rs923256139 Chr.11: 62615144-62615144 on Build GRCh38
rs372630965 Chr.11: 62615128-62615128 on Build GRCh38
rs570679350 Chr.11: 62614993-62614993 on Build GRCh38
rs147684283 Chr.11: 62615094-62615094 on Build GRCh38
rs566443401 Chr.11: 62615103-62615103 on Build GRCh38
rs531890375 Chr.11: 62614911-62614911 on Build GRCh38
rs904058340 Chr.11: 62615300-62615300 on Build GRCh38
rs1001922183 Chr.11: 62615238-62615238 on Build GRCh38
rs376237139 Chr.11: 62615129-62615129 on Build GRCh38
rs918775545 Chr.11: 62614953-62614953 on Build GRCh38
rs1002271199 Chr.11: 62614980-62614980 on Build GRCh38
rs942974841 Chr.11: 62615279-62615279 on Build GRCh38
rs886048441 Chr.11: 62615018-62615018 on Build GRCh38
rs895096098 Chr.11: 62615192-62615192 on Build GRCh38
rs116942133 Chr.11: 62615224-62615224 on Build GRCh38
rs942694594 Chr.11: 62615007-62615007 on Build GRCh38
rs535324390 Chr.11: 62615221-62615221 on Build GRCh38
rs1046931760 Chr.11: 62614915-62614915 on Build GRCh38
rs1005326614 Chr.11: 62614931-62614931 on Build GRCh38
rs1021091629 Chr.11: 62614996-62614996 on Build GRCh38
rs757950484 Chr.11: 62614948-62614948 on Build GRCh38
rs1054496079 Chr.11: 62615018-62615018 on Build GRCh38
rs778890533 Chr.11: 62615215-62615216 on Build GRCh38
rs1042216520 Chr.11: 62615110-62615110 on Build GRCh38
rs893185054 Chr.11: 62615026-62615026 on Build GRCh38
rs1000979000 Chr.11: 62615308-62615308 on Build GRCh38
rs780975259 Chr.11: 62615299-62615299 on Build GRCh38
rs1017727079 Chr.11: 62615323-62615323 on Build GRCh38
rs551577435 Chr.11: 62614958-62614958 on Build GRCh38
rs757033824 Chr.11: 62615254-62615254 on Build GRCh38
rs946131219 Chr.11: 62615089-62615089 on Build GRCh38
rs902332009 Chr.11: 62615113-62615113 on Build GRCh38
rs186029493 Chr.11: 62615266-62615266 on Build GRCh38
rs568508647 Chr.11: 62615234-62615234 on Build GRCh38
rs976485177 Chr.11: 62615139-62615139 on Build GRCh38
rs972471934 Chr.11: 62615240-62615240 on Build GRCh38
rs962415024 Chr.11: 62615237-62615237 on Build GRCh38
rs934233533 Chr.11: 62614974-62614974 on Build GRCh38
rs763702278 Chr.11: 62615074-62615074 on Build GRCh38
rs933850794 Chr.11: 62614897-62614897 on Build GRCh38
rs987446333 Chr.11: 62614988-62614988 on Build GRCh38
rs1035343919 Chr.11: 62615168-62615168 on Build GRCh38
rs1021143862 Chr.11: 62615295-62615295 on Build GRCh38
rs910612847 Chr.11: 62615065-62615065 on Build GRCh38
rs911456558 Chr.11: 62615275-62615275 on Build GRCh38
rs908400992 Chr.11: 62615216-62615216 on Build GRCh38
rs1003399067 Chr.11: 62615146-62615146 on Build GRCh38
rs570657269 Chr.11: 62614969-62614969 on Build GRCh38
rs1025422952 Chr.11: 62615244-62615244 on Build GRCh38
rs896126861 Chr.11: 62615249-62615249 on Build GRCh38
rs964773980 Chr.11: 62615071-62615071 on Build GRCh38
rs1035318941 Chr.11: 62614981-62614981 on Build GRCh38

View Full Product Details