Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62876492-62877024 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC3A2
Entrez Gene ID
6520
Gene Name
solute carrier family 3 member 2
Gene Aliases
4F2, 4F2HC, 4T2HC, CD98, CD98HC, MDU1, NACAE
UniGene
Hs.502769
Gene Chromosome Location
Chr.11: 62856012-62888883 on Build GRCh38
Transcripts
NM_001012662.2 NM_001012664.2 NM_002394.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6397748 c.356C>T p.P119L Chr.11: 62876844-62876844 on Build GRCh38
3396038 c.319A>G p.S107G Chr.11: 62876807-62876807 on Build GRCh38
5970257 c.365C>T p.S122L Chr.11: 62876853-62876853 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs944574194 Chr.11: 62876763-62876763 on Build GRCh38
rs543183034 Chr.11: 62876748-62876748 on Build GRCh38
rs183937541 Chr.11: 62876605-62876605 on Build GRCh38
rs191811946 Chr.11: 62876592-62876592 on Build GRCh38
rs1039177397 Chr.11: 62876664-62876664 on Build GRCh38
rs915732351 Chr.11: 62876803-62876803 on Build GRCh38
rs761903894 Chr.11: 62876631-62876631 on Build GRCh38
rs953035159 Chr.11: 62876701-62876701 on Build GRCh38
rs535184420 Chr.11: 62876982-62876982 on Build GRCh38
rs989181207 Chr.11: 62876675-62876675 on Build GRCh38
rs117818261 Chr.11: 62876742-62876742 on Build GRCh38
rs1042647551 Chr.11: 62876954-62876954 on Build GRCh38
rs895258628 Chr.11: 62876990-62876990 on Build GRCh38
rs758482915 Chr.11: 62876860-62876860 on Build GRCh38
rs566686093 Chr.11: 62876980-62876980 on Build GRCh38
rs1003717513 Chr.11: 62876999-62876999 on Build GRCh38
rs760541875 Chr.11: 62876790-62876790 on Build GRCh38
rs1035143984 Chr.11: 62876749-62876749 on Build GRCh38
rs774984910 Chr.11: 62876610-62876611 on Build GRCh38
rs985865554 Chr.11: 62876724-62876724 on Build GRCh38
rs751399468 Chr.11: 62876578-62876578 on Build GRCh38
rs1007718761 Chr.11: 62876663-62876663 on Build GRCh38
rs192948215 Chr.11: 62876693-62876693 on Build GRCh38
rs539759537 Chr.11: 62876861-62876861 on Build GRCh38
rs116133114 Chr.11: 62876882-62876882 on Build GRCh38
rs369806448 Chr.11: 62876956-62876956 on Build GRCh38
rs893991232 Chr.11: 62876895-62876895 on Build GRCh38
rs957137109 Chr.11: 62876633-62876633 on Build GRCh38
rs1011007871 Chr.11: 62876949-62876949 on Build GRCh38
rs760538271 Chr.11: 62876872-62876872 on Build GRCh38
rs1003585137 Chr.11: 62876979-62876979 on Build GRCh38
rs941905912 Chr.11: 62876532-62876532 on Build GRCh38
rs561348101 Chr.11: 62876527-62876527 on Build GRCh38
rs1011101061 Chr.11: 62876697-62876697 on Build GRCh38
rs919127428 Chr.11: 62876820-62876820 on Build GRCh38
rs763959166 Chr.11: 62876572-62876572 on Build GRCh38
rs552409745 Chr.11: 62876886-62876886 on Build GRCh38
rs1037397621 Chr.11: 62876554-62876554 on Build GRCh38
rs774545432 Chr.11: 62876539-62876539 on Build GRCh38
rs188134162 Chr.11: 62876682-62876682 on Build GRCh38
rs973361554 Chr.11: 62876814-62876814 on Build GRCh38
rs1045209073 Chr.11: 62876865-62876865 on Build GRCh38
rs540420663 Chr.11: 62876645-62876645 on Build GRCh38
rs959440959 Chr.11: 62876784-62876784 on Build GRCh38
rs918940972 Chr.11: 62876562-62876562 on Build GRCh38
rs146764790 Chr.11: 62876611-62876611 on Build GRCh38
rs190424203 Chr.11: 62876838-62876838 on Build GRCh38
rs902755498 Chr.11: 62876969-62876969 on Build GRCh38
rs752972544 Chr.11: 62876849-62876849 on Build GRCh38
rs778806373 Chr.11: 62876810-62876810 on Build GRCh38
rs1048960364 Chr.11: 62876842-62876842 on Build GRCh38
rs1051527128 Chr.11: 62876616-62876616 on Build GRCh38
rs185098712 Chr.11: 62876815-62876815 on Build GRCh38
rs966754926 Chr.11: 62876747-62876747 on Build GRCh38
rs928939044 Chr.11: 62876567-62876567 on Build GRCh38

View Full Product Details