Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62883882-62884386 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC3A2
Entrez Gene ID
6520
Gene Name
solute carrier family 3 member 2
Gene Aliases
4F2, 4F2HC, 4T2HC, CD98, CD98HC, MDU1, NACAE
UniGene
Hs.502769
Gene Chromosome Location
Chr.11: 62856012-62888883 on Build GRCh38
Transcripts
NM_001012662.2 NM_001012664.2 NM_001013251.2 NM_002394.5 NR_037193.1

SNP Detail

SNP ID Chromosome Location
rs528983097 Chr.11: 62884213-62884213 on Build GRCh38
rs980914653 Chr.11: 62884164-62884164 on Build GRCh38
rs915489354 Chr.11: 62884113-62884113 on Build GRCh38
rs755193639 Chr.11: 62884013-62884013 on Build GRCh38
rs182617055 Chr.11: 62883971-62883971 on Build GRCh38
rs987152980 Chr.11: 62884056-62884056 on Build GRCh38
rs973039415 Chr.11: 62883992-62883992 on Build GRCh38
rs566701475 Chr.11: 62884176-62884176 on Build GRCh38
rs1016960599 Chr.11: 62884083-62884083 on Build GRCh38
rs779190152 Chr.11: 62884069-62884069 on Build GRCh38
rs902484622 Chr.11: 62884185-62884185 on Build GRCh38
rs950363657 Chr.11: 62884362-62884362 on Build GRCh38
rs190713396 Chr.11: 62883955-62883955 on Build GRCh38
rs1039702761 Chr.11: 62884096-62884096 on Build GRCh38
rs546815231 Chr.11: 62884119-62884119 on Build GRCh38
rs368223843 Chr.11: 62884348-62884348 on Build GRCh38
rs115750608 Chr.11: 62884336-62884336 on Build GRCh38
rs145642653 Chr.11: 62883920-62883920 on Build GRCh38
rs549115567 Chr.11: 62884255-62884255 on Build GRCh38
rs946749835 Chr.11: 62884131-62884131 on Build GRCh38
rs1003645178 Chr.11: 62884226-62884226 on Build GRCh38
rs1026253886 Chr.11: 62884052-62884052 on Build GRCh38
rs1022846470 Chr.11: 62883970-62883970 on Build GRCh38
rs901709861 Chr.11: 62884021-62884021 on Build GRCh38
rs912065977 Chr.11: 62884068-62884068 on Build GRCh38
rs1013250246 Chr.11: 62883969-62883969 on Build GRCh38
rs1043368063 Chr.11: 62883918-62883918 on Build GRCh38
rs564418350 Chr.11: 62883989-62883989 on Build GRCh38
rs187831298 Chr.11: 62884070-62884070 on Build GRCh38
rs950458073 Chr.11: 62884045-62884045 on Build GRCh38
rs562487002 Chr.11: 62884284-62884284 on Build GRCh38
rs568711495 Chr.11: 62884302-62884302 on Build GRCh38
rs960866436 Chr.11: 62884253-62884253 on Build GRCh38
rs1042395471 Chr.11: 62884133-62884133 on Build GRCh38
rs73487880 Chr.11: 62883981-62883981 on Build GRCh38
rs542745783 Chr.11: 62883942-62883942 on Build GRCh38
rs963306867 Chr.11: 62884339-62884339 on Build GRCh38
rs969793027 Chr.11: 62884145-62884145 on Build GRCh38
rs914166365 Chr.11: 62884272-62884272 on Build GRCh38
rs752318350 Chr.11: 62883928-62883928 on Build GRCh38
rs1025950308 Chr.11: 62884358-62884358 on Build GRCh38
rs781024514 Chr.11: 62883935-62883935 on Build GRCh38
rs1012744159 Chr.11: 62884320-62884320 on Build GRCh38

View Full Product Details