Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 63614280-63614737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLA2G16
Entrez Gene ID
11145
Gene Name
phospholipase A2 group XVI
Gene Aliases
AdPLA, H-REV107-1, HRASLS3, HREV107, HREV107-1, HREV107-3, HRSL3
UniGene
Hs.502775
Gene Chromosome Location
Chr.11: 63574462-63616926 on Build GRCh38
Transcripts
NM_001128203.1 NM_007069.3 XM_006718426.1 XM_011544741.1

SNP Detail

SNP ID Chromosome Location
rs138447550 Chr.11: 63614404-63614404 on Build GRCh38
rs914855871 Chr.11: 63614614-63614614 on Build GRCh38
rs994598210 Chr.11: 63614576-63614576 on Build GRCh38
rs962394896 Chr.11: 63614506-63614506 on Build GRCh38
rs12277033 Chr.11: 63614601-63614601 on Build GRCh38
rs1025663275 Chr.11: 63614584-63614584 on Build GRCh38
rs571261289 Chr.11: 63614354-63614354 on Build GRCh38
rs1037520498 Chr.11: 63614688-63614688 on Build GRCh38
rs34785126 Chr.11: 63614448-63614448 on Build GRCh38
rs899703396 Chr.11: 63614458-63614458 on Build GRCh38
rs917491111 Chr.11: 63614373-63614373 on Build GRCh38
rs147354386 Chr.11: 63614629-63614629 on Build GRCh38
rs1054640094 Chr.11: 63614421-63614421 on Build GRCh38
rs934988307 Chr.11: 63614650-63614659 on Build GRCh38
rs1055590514 Chr.11: 63614660-63614660 on Build GRCh38
rs386754290 Chr.11: 63614404-63614405 on Build GRCh38
rs897630721 Chr.11: 63614696-63614696 on Build GRCh38
rs987267462 Chr.11: 63614394-63614394 on Build GRCh38
rs914826213 Chr.11: 63614456-63614456 on Build GRCh38
rs2030730 Chr.11: 63614375-63614375 on Build GRCh38
rs182578761 Chr.11: 63614656-63614656 on Build GRCh38
rs3809072 Chr.11: 63614504-63614504 on Build GRCh38
rs560440210 Chr.11: 63614556-63614556 on Build GRCh38
rs3809073 Chr.11: 63614405-63614405 on Build GRCh38
rs977687520 Chr.11: 63614595-63614595 on Build GRCh38
rs35902999 Chr.11: 63614588-63614588 on Build GRCh38
rs765330280 Chr.11: 63614489-63614489 on Build GRCh38
rs962743565 Chr.11: 63614335-63614335 on Build GRCh38
rs537066103 Chr.11: 63614467-63614467 on Build GRCh38
rs955529446 Chr.11: 63614585-63614585 on Build GRCh38
rs149254878 Chr.11: 63614609-63614609 on Build GRCh38
rs921312991 Chr.11: 63614649-63614649 on Build GRCh38
rs556755395 Chr.11: 63614478-63614478 on Build GRCh38
rs553191897 Chr.11: 63614569-63614569 on Build GRCh38
rs958898810 Chr.11: 63614560-63614560 on Build GRCh38
rs541697676 Chr.11: 63614596-63614596 on Build GRCh38
rs566335892 Chr.11: 63614346-63614346 on Build GRCh38
rs1029590957 Chr.11: 63614477-63614477 on Build GRCh38
rs894221627 Chr.11: 63614662-63614662 on Build GRCh38
rs938456463 Chr.11: 63614490-63614490 on Build GRCh38
rs945739140 Chr.11: 63614473-63614473 on Build GRCh38
rs911491660 Chr.11: 63614563-63614563 on Build GRCh38
rs1016147902 Chr.11: 63614313-63614313 on Build GRCh38
rs753886668 Chr.11: 63614380-63614380 on Build GRCh38
rs918695666 Chr.11: 63614348-63614348 on Build GRCh38
rs1016223233 Chr.11: 63614552-63614552 on Build GRCh38
rs7482863 Chr.11: 63614530-63614530 on Build GRCh38
rs552752297 Chr.11: 63614711-63614711 on Build GRCh38
rs867957201 Chr.11: 63614672-63614672 on Build GRCh38
rs75668261 Chr.11: 63614706-63614706 on Build GRCh38
rs1018275530 Chr.11: 63614600-63614600 on Build GRCh38
rs376205228 Chr.11: 63614623-63614623 on Build GRCh38
rs1055516860 Chr.11: 63614498-63614498 on Build GRCh38
rs35941673 Chr.11: 63614469-63614470 on Build GRCh38
rs978046355 Chr.11: 63614655-63614655 on Build GRCh38
rs2030729 Chr.11: 63614551-63614551 on Build GRCh38
rs938306051 Chr.11: 63614659-63614659 on Build GRCh38

View Full Product Details