Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 63988247-63988756 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OTUB1
Entrez Gene ID
55611
Gene Name
OTU deubiquitinase, ubiquitin aldehyde binding 1
Gene Aliases
HSPC263, OTB1, OTU1
UniGene
Hs.473788
Gene Chromosome Location
Chr.11: 63985853-63998420 on Build GRCh38
Transcripts
NM_017670.2 NR_003089.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5510514 c.123C>T p.D41D Chr.11: 63988629-63988629 on Build GRCh38
5649683 c.269_270insGCG p.E90_L91insR Chr.11: 63988691-63988692 on Build GRCh38
4941090 c.106C>A p.R36R Chr.11: 63988384-63988384 on Build GRCh38
5649682 c.158_159insGCG p.E53_L54insR Chr.11: 63988691-63988692 on Build GRCh38
5649684 c.185_186insGCG p.E62_L63insR Chr.11: 63988691-63988692 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766516104 Chr.11: 63988601-63988601 on Build GRCh38
rs942232967 Chr.11: 63988713-63988713 on Build GRCh38
rs765392727 Chr.11: 63988693-63988693 on Build GRCh38
rs908572133 Chr.11: 63988556-63988556 on Build GRCh38
rs917169479 Chr.11: 63988490-63988490 on Build GRCh38
rs146537845 Chr.11: 63988355-63988355 on Build GRCh38
rs774306824 Chr.11: 63988418-63988418 on Build GRCh38
rs1004900791 Chr.11: 63988332-63988332 on Build GRCh38
rs559126240 Chr.11: 63988467-63988467 on Build GRCh38
rs144215400 Chr.11: 63988353-63988353 on Build GRCh38
rs968429585 Chr.11: 63988417-63988417 on Build GRCh38
rs996980060 Chr.11: 63988368-63988368 on Build GRCh38
rs918292936 Chr.11: 63988671-63988671 on Build GRCh38
rs778277838 Chr.11: 63988621-63988621 on Build GRCh38
rs761783927 Chr.11: 63988448-63988448 on Build GRCh38
rs748789116 Chr.11: 63988630-63988630 on Build GRCh38
rs748068070 Chr.11: 63988640-63988640 on Build GRCh38
rs369002057 Chr.11: 63988685-63988685 on Build GRCh38
rs753897462 Chr.11: 63988602-63988602 on Build GRCh38
rs760884620 Chr.11: 63988600-63988600 on Build GRCh38
rs765398197 Chr.11: 63988616-63988616 on Build GRCh38
rs752889774 Chr.11: 63988698-63988698 on Build GRCh38
rs543774091 Chr.11: 63988557-63988557 on Build GRCh38
rs773051756 Chr.11: 63988599-63988599 on Build GRCh38
rs758905157 Chr.11: 63988700-63988700 on Build GRCh38
rs744864 Chr.11: 63988489-63988489 on Build GRCh38
rs746644686 Chr.11: 63988588-63988591 on Build GRCh38
rs759850958 Chr.11: 63988691-63988691 on Build GRCh38
rs369249906 Chr.11: 63988628-63988628 on Build GRCh38
rs566264770 Chr.11: 63988478-63988478 on Build GRCh38
rs373268158 Chr.11: 63988622-63988622 on Build GRCh38
rs768585519 Chr.11: 63988383-63988383 on Build GRCh38
rs1028085841 Chr.11: 63988392-63988392 on Build GRCh38
rs528268983 Chr.11: 63988701-63988701 on Build GRCh38
rs773902347 Chr.11: 63988634-63988634 on Build GRCh38
rs374524419 Chr.11: 63988270-63988270 on Build GRCh38
rs576458828 Chr.11: 63988626-63988626 on Build GRCh38
rs139788271 Chr.11: 63988689-63988689 on Build GRCh38
rs759030305 Chr.11: 63988618-63988618 on Build GRCh38
rs554865823 Chr.11: 63988510-63988510 on Build GRCh38
rs772066149 Chr.11: 63988641-63988641 on Build GRCh38
rs943719379 Chr.11: 63988321-63988321 on Build GRCh38
rs760399947 Chr.11: 63988663-63988663 on Build GRCh38
rs1048352311 Chr.11: 63988304-63988304 on Build GRCh38
rs112636655 Chr.11: 63988570-63988570 on Build GRCh38
rs887385950 Chr.11: 63988331-63988331 on Build GRCh38
rs1035660666 Chr.11: 63988534-63988534 on Build GRCh38
rs1048460674 Chr.11: 63988547-63988547 on Build GRCh38
rs745500946 Chr.11: 63988334-63988334 on Build GRCh38
rs183970848 Chr.11: 63988682-63988682 on Build GRCh38
rs904675514 Chr.11: 63988720-63988720 on Build GRCh38
rs748757526 Chr.11: 63988379-63988379 on Build GRCh38
rs758583806 Chr.11: 63988484-63988484 on Build GRCh38
rs374859386 Chr.11: 63988587-63988587 on Build GRCh38
rs151160538 Chr.11: 63988631-63988631 on Build GRCh38
rs17850645 Chr.11: 63988715-63988715 on Build GRCh38
rs575035360 Chr.11: 63988527-63988527 on Build GRCh38
rs931373656 Chr.11: 63988540-63988540 on Build GRCh38
rs1002879037 Chr.11: 63988531-63988531 on Build GRCh38
rs375265756 Chr.11: 63988644-63988644 on Build GRCh38
rs549302473 Chr.11: 63988611-63988611 on Build GRCh38
rs778329122 Chr.11: 63988326-63988326 on Build GRCh38
rs1031775238 Chr.11: 63988466-63988466 on Build GRCh38
rs969866305 Chr.11: 63988503-63988503 on Build GRCh38
rs755036323 Chr.11: 63988612-63988612 on Build GRCh38
rs181133600 Chr.11: 63988558-63988558 on Build GRCh38
rs767280029 Chr.11: 63988578-63988578 on Build GRCh38
rs926722437 Chr.11: 63988516-63988516 on Build GRCh38
rs943750383 Chr.11: 63988623-63988623 on Build GRCh38
rs541962168 Chr.11: 63988704-63988704 on Build GRCh38
rs149786795 Chr.11: 63988722-63988722 on Build GRCh38

View Full Product Details